Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 158293940-158294476 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD1B
Entrez Gene ID
910
Gene Name
CD1b molecule
Gene Aliases
CD1, CD1A, R1
UniGene
Hs.1310
Gene Chromosome Location
Chr.1: 158286841-158332038 on Build GRCh38
Transcripts
XM_017002785.1 NM_001765.2 XM_005245579.4

Gene Symbol
CD1C
Entrez Gene ID
911
Gene Name
CD1c molecule
Gene Aliases
BDCA1, CD1, CD1A, R7
UniGene
Hs.132448
Gene Chromosome Location
Chr.1: 158289773-158294774 on Build GRCh38
Transcripts
XM_017002785.1 NM_001765.2 XM_005245579.4

SNP Detail

SNP ID Chromosome Location
rs528013348 Chr.1: 158294122-158294122 on Build GRCh38
rs1006171794 Chr.1: 158294005-158294005 on Build GRCh38
rs755336138 Chr.1: 158294394-158294394 on Build GRCh38
rs994187375 Chr.1: 158294443-158294443 on Build GRCh38
rs555856405 Chr.1: 158294246-158294246 on Build GRCh38
rs1031700499 Chr.1: 158294034-158294034 on Build GRCh38
rs943272035 Chr.1: 158294292-158294292 on Build GRCh38
rs760670077 Chr.1: 158294417-158294417 on Build GRCh38
rs1025736844 Chr.1: 158294077-158294077 on Build GRCh38
rs57287745 Chr.1: 158293965-158293965 on Build GRCh38
rs113892954 Chr.1: 158294071-158294071 on Build GRCh38
rs980749829 Chr.1: 158294424-158294424 on Build GRCh38
rs898539247 Chr.1: 158294067-158294067 on Build GRCh38
rs1037657272 Chr.1: 158294393-158294393 on Build GRCh38
rs77773003 Chr.1: 158294366-158294366 on Build GRCh38
rs914793170 Chr.1: 158294165-158294165 on Build GRCh38
rs779439717 Chr.1: 158294418-158294418 on Build GRCh38
rs918934653 Chr.1: 158294250-158294250 on Build GRCh38
rs73023949 Chr.1: 158294230-158294230 on Build GRCh38
rs1011545084 Chr.1: 158294380-158294380 on Build GRCh38
rs567662422 Chr.1: 158294154-158294154 on Build GRCh38
rs891810145 Chr.1: 158294373-158294373 on Build GRCh38
rs958965508 Chr.1: 158294135-158294135 on Build GRCh38
rs553535194 Chr.1: 158294441-158294441 on Build GRCh38
rs971947347 Chr.1: 158294111-158294111 on Build GRCh38
rs567776493 Chr.1: 158294364-158294364 on Build GRCh38
rs565746411 Chr.1: 158294413-158294413 on Build GRCh38
rs1038894831 Chr.1: 158294312-158294312 on Build GRCh38
rs887733066 Chr.1: 158294271-158294271 on Build GRCh38
rs992300896 Chr.1: 158294243-158294243 on Build GRCh38
rs538008167 Chr.1: 158294098-158294098 on Build GRCh38
rs939146245 Chr.1: 158294242-158294242 on Build GRCh38
rs1048967236 Chr.1: 158294270-158294270 on Build GRCh38
rs967024806 Chr.1: 158294255-158294255 on Build GRCh38
rs941925574 Chr.1: 158294377-158294377 on Build GRCh38
rs970635084 Chr.1: 158294181-158294181 on Build GRCh38
rs570982901 Chr.1: 158294130-158294130 on Build GRCh38
rs966186238 Chr.1: 158293967-158293967 on Build GRCh38
rs1021633817 Chr.1: 158294406-158294406 on Build GRCh38
rs3138107 Chr.1: 158293976-158293976 on Build GRCh38
rs114449264 Chr.1: 158294123-158294123 on Build GRCh38
rs970371130 Chr.1: 158294421-158294421 on Build GRCh38
rs566853666 Chr.1: 158294073-158294073 on Build GRCh38
rs928938013 Chr.1: 158294265-158294265 on Build GRCh38
rs563525747 Chr.1: 158294128-158294131 on Build GRCh38
rs1016929225 Chr.1: 158294047-158294047 on Build GRCh38
rs1035889358 Chr.1: 158294428-158294428 on Build GRCh38
rs928789820 Chr.1: 158294234-158294234 on Build GRCh38
rs901654861 Chr.1: 158294356-158294356 on Build GRCh38
rs762968561 Chr.1: 158294222-158294222 on Build GRCh38
rs981644986 Chr.1: 158294121-158294121 on Build GRCh38

View Full Product Details