Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161190937-161191440 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS4
Entrez Gene ID
9507
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 4
Gene Aliases
ADAMTS-2, ADAMTS-4, ADMP-1
UniGene
Hs.211604
Gene Chromosome Location
Chr.1: 161189725-161199080 on Build GRCh38
Transcripts
NM_001320336.1 NM_005099.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1689013 c.2236C>T p.P746S Chr.1: 161191416-161191416 on Build GRCh38
5477727 c.2415G>A p.P805P Chr.1: 161191237-161191237 on Build GRCh38
2086883 c.2389C>T p.R797* Chr.1: 161191263-161191263 on Build GRCh38
4902221 c.2264G>A p.G755E Chr.1: 161191388-161191388 on Build GRCh38
6059049 c.2325G>A p.G775G Chr.1: 161191327-161191327 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs927775299 Chr.1: 161191167-161191167 on Build GRCh38
rs373248605 Chr.1: 161191341-161191341 on Build GRCh38
rs112325225 Chr.1: 161191257-161191257 on Build GRCh38
rs979493536 Chr.1: 161191066-161191066 on Build GRCh38
rs747846256 Chr.1: 161191096-161191096 on Build GRCh38
rs201242393 Chr.1: 161191307-161191307 on Build GRCh38
rs201146448 Chr.1: 161191128-161191128 on Build GRCh38
rs776872102 Chr.1: 161191173-161191173 on Build GRCh38
rs750256063 Chr.1: 161191126-161191126 on Build GRCh38
rs1009344636 Chr.1: 161191052-161191052 on Build GRCh38
rs894771559 Chr.1: 161191008-161191008 on Build GRCh38
rs200302452 Chr.1: 161191184-161191184 on Build GRCh38
rs766357712 Chr.1: 161191133-161191133 on Build GRCh38
rs568647916 Chr.1: 161190993-161190993 on Build GRCh38
rs781559105 Chr.1: 161191269-161191269 on Build GRCh38
rs41270037 Chr.1: 161191043-161191043 on Build GRCh38
rs138218745 Chr.1: 161191383-161191383 on Build GRCh38
rs879128704 Chr.1: 161191399-161191399 on Build GRCh38
rs538312174 Chr.1: 161191055-161191055 on Build GRCh38
rs553901778 Chr.1: 161191025-161191025 on Build GRCh38
rs41270039 Chr.1: 161191226-161191226 on Build GRCh38
rs745735551 Chr.1: 161191296-161191296 on Build GRCh38
rs377576787 Chr.1: 161191163-161191163 on Build GRCh38
rs926625483 Chr.1: 161191070-161191070 on Build GRCh38
rs749440061 Chr.1: 161191391-161191391 on Build GRCh38
rs760634103 Chr.1: 161191256-161191256 on Build GRCh38
rs568445358 Chr.1: 161191238-161191238 on Build GRCh38
rs139491371 Chr.1: 161191373-161191373 on Build GRCh38
rs770065512 Chr.1: 161191170-161191170 on Build GRCh38
rs1027595025 Chr.1: 161191397-161191397 on Build GRCh38
rs955219383 Chr.1: 161191047-161191047 on Build GRCh38
rs114600134 Chr.1: 161191151-161191151 on Build GRCh38
rs776532659 Chr.1: 161191113-161191113 on Build GRCh38
rs534230869 Chr.1: 161191024-161191024 on Build GRCh38
rs753727996 Chr.1: 161190960-161190960 on Build GRCh38
rs780587893 Chr.1: 161191225-161191225 on Build GRCh38
rs755682745 Chr.1: 161191214-161191214 on Build GRCh38
rs115479312 Chr.1: 161191248-161191248 on Build GRCh38
rs146525824 Chr.1: 161191161-161191161 on Build GRCh38
rs114728887 Chr.1: 161191164-161191164 on Build GRCh38
rs748315188 Chr.1: 161191237-161191237 on Build GRCh38
rs781544063 Chr.1: 161191234-161191234 on Build GRCh38
rs771097446 Chr.1: 161191242-161191242 on Build GRCh38
rs753559369 Chr.1: 161191221-161191221 on Build GRCh38
rs752025981 Chr.1: 161191268-161191268 on Build GRCh38
rs771778596 Chr.1: 161191297-161191297 on Build GRCh38
rs111348569 Chr.1: 161191236-161191236 on Build GRCh38
rs774434720 Chr.1: 161191293-161191293 on Build GRCh38
rs757018849 Chr.1: 161191262-161191262 on Build GRCh38
rs200032286 Chr.1: 161191150-161191150 on Build GRCh38
rs767338527 Chr.1: 161191199-161191199 on Build GRCh38
rs749640297 Chr.1: 161191279-161191280 on Build GRCh38
rs200129460 Chr.1: 161191193-161191193 on Build GRCh38
rs11807350 Chr.1: 161191145-161191145 on Build GRCh38
rs776137620 Chr.1: 161191105-161191105 on Build GRCh38
rs750055404 Chr.1: 161191366-161191366 on Build GRCh38
rs527950908 Chr.1: 161191241-161191241 on Build GRCh38
rs200466524 Chr.1: 161191135-161191135 on Build GRCh38
rs764246778 Chr.1: 161191112-161191112 on Build GRCh38
rs749352206 Chr.1: 161191282-161191282 on Build GRCh38
rs370882335 Chr.1: 161191174-161191174 on Build GRCh38
rs1038435908 Chr.1: 161190992-161190992 on Build GRCh38
rs780506474 Chr.1: 161191094-161191094 on Build GRCh38
rs774881907 Chr.1: 161191098-161191098 on Build GRCh38
rs997001091 Chr.1: 161191009-161191009 on Build GRCh38
rs761401967 Chr.1: 161191347-161191347 on Build GRCh38
rs770089604 Chr.1: 161191240-161191240 on Build GRCh38
rs769245494 Chr.1: 161191228-161191228 on Build GRCh38
rs769295102 Chr.1: 161191097-161191097 on Build GRCh38
rs751276435 Chr.1: 161191186-161191186 on Build GRCh38
rs759910530 Chr.1: 161191183-161191183 on Build GRCh38
rs752293807 Chr.1: 161191213-161191213 on Build GRCh38
rs778397651 Chr.1: 161191263-161191263 on Build GRCh38
rs950769847 Chr.1: 161191377-161191377 on Build GRCh38
rs554591180 Chr.1: 161191125-161191125 on Build GRCh38
rs746215424 Chr.1: 161191101-161191101 on Build GRCh38
rs760726172 Chr.1: 161191326-161191326 on Build GRCh38
rs776320045 Chr.1: 161191331-161191331 on Build GRCh38
rs113278384 Chr.1: 161191396-161191396 on Build GRCh38
rs149404061 Chr.1: 161191166-161191166 on Build GRCh38
rs779637375 Chr.1: 161191056-161191056 on Build GRCh38
rs771185545 Chr.1: 161191285-161191285 on Build GRCh38
rs148170603 Chr.1: 161191220-161191220 on Build GRCh38
rs376147938 Chr.1: 161191115-161191115 on Build GRCh38
rs145548254 Chr.1: 161191380-161191380 on Build GRCh38
rs202054746 Chr.1: 161191102-161191102 on Build GRCh38
rs752772603 Chr.1: 161191209-161191209 on Build GRCh38
rs150535186 Chr.1: 161191272-161191272 on Build GRCh38
rs757571443 Chr.1: 161191326-161191327 on Build GRCh38
rs778324760 Chr.1: 161191278-161191279 on Build GRCh38
rs34884997 Chr.1: 161191082-161191082 on Build GRCh38
rs1011162222 Chr.1: 161191022-161191022 on Build GRCh38
rs777539144 Chr.1: 161191158-161191158 on Build GRCh38
rs773407497 Chr.1: 161191171-161191171 on Build GRCh38
rs1005259594 Chr.1: 161190965-161190965 on Build GRCh38
rs546046388 Chr.1: 161191160-161191160 on Build GRCh38
rs767560823 Chr.1: 161191251-161191251 on Build GRCh38
rs376187908 Chr.1: 161191364-161191364 on Build GRCh38
rs116068231 Chr.1: 161191249-161191249 on Build GRCh38
rs750317755 Chr.1: 161191106-161191106 on Build GRCh38
rs756656653 Chr.1: 161191242-161191242 on Build GRCh38
rs777856617 Chr.1: 161191281-161191281 on Build GRCh38
rs756904302 Chr.1: 161191223-161191223 on Build GRCh38
rs369928976 Chr.1: 161191378-161191378 on Build GRCh38
rs182544563 Chr.1: 161191374-161191374 on Build GRCh38
rs114152196 Chr.1: 161191254-161191254 on Build GRCh38
rs770241246 Chr.1: 161191276-161191276 on Build GRCh38
rs757273170 Chr.1: 161191401-161191401 on Build GRCh38
rs891266079 Chr.1: 161191360-161191360 on Build GRCh38

View Full Product Details