Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 116854053-116854581 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAD21
Entrez Gene ID
5885
Gene Name
RAD21 cohesin complex component
Gene Aliases
CDLS4, HR21, HRAD21, MCD1, NXP1, SCC1, hHR21
UniGene
Hs.81848
Gene Chromosome Location
Chr.8: 116845934-116874866 on Build GRCh38
Transcripts
NM_006265.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6961900 c.1151G>A p.R384K Chr.8: 116854255-116854255 on Build GRCh38
1488937 c.1130C>G p.A377G Chr.8: 116854276-116854276 on Build GRCh38
5945152 c.1161+1G>A p.? Chr.8: 116854244-116854244 on Build GRCh38
6964430 c.1105G>A p.V369I Chr.8: 116854301-116854301 on Build GRCh38
5945153 c.1142G>A p.W381* Chr.8: 116854264-116854264 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761874801 Chr.8: 116854241-116854241 on Build GRCh38
rs200333487 Chr.8: 116854479-116854479 on Build GRCh38
rs1007740879 Chr.8: 116854183-116854183 on Build GRCh38
rs142066051 Chr.8: 116854126-116854128 on Build GRCh38
rs765764891 Chr.8: 116854511-116854511 on Build GRCh38
rs755883894 Chr.8: 116854358-116854358 on Build GRCh38
rs201090747 Chr.8: 116854187-116854188 on Build GRCh38
rs202233096 Chr.8: 116854341-116854341 on Build GRCh38
rs917737700 Chr.8: 116854556-116854556 on Build GRCh38
rs780794833 Chr.8: 116854213-116854213 on Build GRCh38
rs375317261 Chr.8: 116854214-116854214 on Build GRCh38
rs964642413 Chr.8: 116854088-116854088 on Build GRCh38
rs200852626 Chr.8: 116854240-116854240 on Build GRCh38
rs1046539043 Chr.8: 116854360-116854360 on Build GRCh38
rs779990116 Chr.8: 116854364-116854364 on Build GRCh38
rs762219753 Chr.8: 116854495-116854495 on Build GRCh38
rs748990686 Chr.8: 116854309-116854309 on Build GRCh38
rs749183142 Chr.8: 116854366-116854366 on Build GRCh38
rs767878994 Chr.8: 116854519-116854520 on Build GRCh38
rs375637600 Chr.8: 116854491-116854491 on Build GRCh38
rs372737701 Chr.8: 116854329-116854329 on Build GRCh38
rs758001515 Chr.8: 116854278-116854278 on Build GRCh38
rs1033099909 Chr.8: 116854153-116854153 on Build GRCh38
rs78584904 Chr.8: 116854101-116854101 on Build GRCh38
rs748245537 Chr.8: 116854465-116854465 on Build GRCh38
rs761113004 Chr.8: 116854270-116854270 on Build GRCh38
rs387907212 Chr.8: 116854279-116854279 on Build GRCh38
rs757084262 Chr.8: 116854211-116854211 on Build GRCh38
rs771370772 Chr.8: 116854490-116854490 on Build GRCh38
rs772246627 Chr.8: 116854477-116854477 on Build GRCh38
rs1045802969 Chr.8: 116854115-116854115 on Build GRCh38
rs140766458 Chr.8: 116854080-116854080 on Build GRCh38
rs201146181 Chr.8: 116854392-116854392 on Build GRCh38
rs768674798 Chr.8: 116854238-116854238 on Build GRCh38
rs751240893 Chr.8: 116854326-116854326 on Build GRCh38
rs377667234 Chr.8: 116854509-116854509 on Build GRCh38
rs774690551 Chr.8: 116854482-116854482 on Build GRCh38
rs768725843 Chr.8: 116854407-116854407 on Build GRCh38
rs938268975 Chr.8: 116854420-116854420 on Build GRCh38
rs764414089 Chr.8: 116854222-116854222 on Build GRCh38
rs921269485 Chr.8: 116854134-116854134 on Build GRCh38
rs772262381 Chr.8: 116854239-116854239 on Build GRCh38
rs267601737 Chr.8: 116854372-116854372 on Build GRCh38
rs931343064 Chr.8: 116854176-116854176 on Build GRCh38
rs775377566 Chr.8: 116854216-116854216 on Build GRCh38
rs998823213 Chr.8: 116854142-116854142 on Build GRCh38
rs550840858 Chr.8: 116854236-116854236 on Build GRCh38
rs908977328 Chr.8: 116854514-116854514 on Build GRCh38
rs761355472 Chr.8: 116854515-116854515 on Build GRCh38
rs185554904 Chr.8: 116854178-116854178 on Build GRCh38
rs199944918 Chr.8: 116854258-116854258 on Build GRCh38
rs143272532 Chr.8: 116854261-116854261 on Build GRCh38
rs893432919 Chr.8: 116854499-116854499 on Build GRCh38
rs10101469 Chr.8: 116854541-116854541 on Build GRCh38
rs778800442 Chr.8: 116854437-116854437 on Build GRCh38
rs761168258 Chr.8: 116854486-116854486 on Build GRCh38
rs746605400 Chr.8: 116854197-116854197 on Build GRCh38
rs547750022 Chr.8: 116854275-116854275 on Build GRCh38
rs754371615 Chr.8: 116854277-116854277 on Build GRCh38
rs377195087 Chr.8: 116854335-116854335 on Build GRCh38
rs565975762 Chr.8: 116854328-116854328 on Build GRCh38
rs902290392 Chr.8: 116854548-116854548 on Build GRCh38
rs372984542 Chr.8: 116854128-116854130 on Build GRCh38
rs879723853 Chr.8: 116854219-116854219 on Build GRCh38

View Full Product Details