Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 143694398-143694888 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF707
Entrez Gene ID
286075
Gene Name
zinc finger protein 707
Gene Aliases
-
UniGene
Hs.521922
Gene Chromosome Location
Chr.8: 143684452-143695633 on Build GRCh38
Transcripts
NM_001100598.1 NM_001100599.1 NM_001288805.1 NM_001288806.1 NM_001288807.1 NM_001288808.1 NM_001288809.1 NM_173831.3 NR_110190.1 NR_110191.1 NR_110192.1 NR_110193.1 XM_005250883.3 XM_006716542.3 XM_006716543.3 XM_011516977.2 XM_011516979.2 XM_017013335.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6832583 c.1061G>A p.R354H Chr.8: 143694475-143694475 on Build GRCh38
4138636 c.1079C>T p.T360M Chr.8: 143694493-143694493 on Build GRCh38
1097002 c.1104C>T p.H368H Chr.8: 143694518-143694518 on Build GRCh38
380022 c.1039G>T p.G347C Chr.8: 143694453-143694453 on Build GRCh38
7202977 c.1032C>T p.Y344Y Chr.8: 143694446-143694446 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs782716133 Chr.8: 143694418-143694418 on Build GRCh38
rs754197074 Chr.8: 143694497-143694497 on Build GRCh38
rs750449242 Chr.8: 143694754-143694754 on Build GRCh38
rs1041409339 Chr.8: 143694706-143694706 on Build GRCh38
rs781886091 Chr.8: 143694483-143694483 on Build GRCh38
rs56213419 Chr.8: 143694737-143694737 on Build GRCh38
rs782183106 Chr.8: 143694427-143694427 on Build GRCh38
rs367801774 Chr.8: 143694623-143694623 on Build GRCh38
rs781939649 Chr.8: 143694452-143694452 on Build GRCh38
rs202174797 Chr.8: 143694535-143694535 on Build GRCh38
rs782663817 Chr.8: 143694433-143694433 on Build GRCh38
rs373473246 Chr.8: 143694538-143694538 on Build GRCh38
rs549568894 Chr.8: 143694768-143694768 on Build GRCh38
rs782715175 Chr.8: 143694474-143694474 on Build GRCh38
rs914362222 Chr.8: 143694659-143694659 on Build GRCh38
rs937219977 Chr.8: 143694625-143694625 on Build GRCh38
rs782805732 Chr.8: 143694461-143694461 on Build GRCh38
rs1055562021 Chr.8: 143694446-143694446 on Build GRCh38
rs559876557 Chr.8: 143694453-143694453 on Build GRCh38
rs534396047 Chr.8: 143694496-143694496 on Build GRCh38
rs902873805 Chr.8: 143694724-143694724 on Build GRCh38
rs782678907 Chr.8: 143694564-143694564 on Build GRCh38
rs550364821 Chr.8: 143694463-143694463 on Build GRCh38
rs562546726 Chr.8: 143694736-143694736 on Build GRCh38
rs782250250 Chr.8: 143694437-143694437 on Build GRCh38
rs782810361 Chr.8: 143694549-143694549 on Build GRCh38
rs947144862 Chr.8: 143694688-143694688 on Build GRCh38
rs369140192 Chr.8: 143694536-143694536 on Build GRCh38
rs781838187 Chr.8: 143694479-143694479 on Build GRCh38
rs371296133 Chr.8: 143694518-143694518 on Build GRCh38
rs943628360 Chr.8: 143694812-143694812 on Build GRCh38
rs577662333 Chr.8: 143694539-143694539 on Build GRCh38
rs782046557 Chr.8: 143694616-143694616 on Build GRCh38
rs202032154 Chr.8: 143694440-143694440 on Build GRCh38
rs554441219 Chr.8: 143694670-143694670 on Build GRCh38
rs531435183 Chr.8: 143694806-143694806 on Build GRCh38
rs782241373 Chr.8: 143694534-143694534 on Build GRCh38
rs1037921281 Chr.8: 143694831-143694831 on Build GRCh38
rs201437689 Chr.8: 143694531-143694531 on Build GRCh38
rs971098511 Chr.8: 143694547-143694547 on Build GRCh38
rs115859646 Chr.8: 143694571-143694571 on Build GRCh38
rs782121498 Chr.8: 143694469-143694469 on Build GRCh38
rs928491857 Chr.8: 143694421-143694421 on Build GRCh38
rs970768151 Chr.8: 143694761-143694761 on Build GRCh38
rs553159699 Chr.8: 143694527-143694527 on Build GRCh38
rs781955256 Chr.8: 143694445-143694445 on Build GRCh38
rs7464270 Chr.8: 143694543-143694543 on Build GRCh38
rs782429639 Chr.8: 143694813-143694813 on Build GRCh38
rs953583830 Chr.8: 143694494-143694494 on Build GRCh38
rs527949882 Chr.8: 143694763-143694763 on Build GRCh38
rs137980493 Chr.8: 143694805-143694805 on Build GRCh38
rs375504308 Chr.8: 143694480-143694480 on Build GRCh38
rs782498822 Chr.8: 143694512-143694512 on Build GRCh38
rs914409396 Chr.8: 143694788-143694788 on Build GRCh38
rs560816234 Chr.8: 143694741-143694741 on Build GRCh38
rs542787194 Chr.8: 143694648-143694648 on Build GRCh38
rs782318302 Chr.8: 143694448-143694448 on Build GRCh38
rs899351048 Chr.8: 143694836-143694836 on Build GRCh38
rs781791350 Chr.8: 143694556-143694556 on Build GRCh38
rs370290602 Chr.8: 143694808-143694808 on Build GRCh38
rs115259396 Chr.8: 143694811-143694811 on Build GRCh38
rs782455127 Chr.8: 143694563-143694563 on Build GRCh38
rs567002350 Chr.8: 143694475-143694475 on Build GRCh38
rs369906843 Chr.8: 143694508-143694508 on Build GRCh38
rs782746265 Chr.8: 143694555-143694555 on Build GRCh38
rs782141283 Chr.8: 143694548-143694548 on Build GRCh38
rs375874764 Chr.8: 143694521-143694521 on Build GRCh38
rs993586250 Chr.8: 143694748-143694748 on Build GRCh38
rs1011220555 Chr.8: 143694464-143694464 on Build GRCh38
rs899300187 Chr.8: 143694747-143694747 on Build GRCh38
rs782325191 Chr.8: 143694428-143694428 on Build GRCh38
rs782498027 Chr.8: 143694574-143694574 on Build GRCh38
rs997780409 Chr.8: 143694490-143694490 on Build GRCh38
rs782406291 Chr.8: 143694525-143694525 on Build GRCh38
rs781800394 Chr.8: 143694776-143694776 on Build GRCh38
rs977153057 Chr.8: 143694797-143694797 on Build GRCh38
rs781873993 Chr.8: 143694552-143694552 on Build GRCh38
rs190577044 Chr.8: 143694575-143694575 on Build GRCh38
rs958869492 Chr.8: 143694772-143694772 on Build GRCh38
rs574097304 Chr.8: 143694720-143694720 on Build GRCh38
rs991315966 Chr.8: 143694785-143694785 on Build GRCh38
rs568885751 Chr.8: 143694725-143694725 on Build GRCh38
rs902822906 Chr.8: 143694473-143694473 on Build GRCh38
rs544262330 Chr.8: 143694729-143694729 on Build GRCh38
rs1056017125 Chr.8: 143694627-143694627 on Build GRCh38
rs997562299 Chr.8: 143694727-143694727 on Build GRCh38

View Full Product Details