Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 34400646-34401139 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf24
Entrez Gene ID
84688
Gene Name
chromosome 9 open reading frame 24
Gene Aliases
CBE1, NYD-SP22, SMRP1, bA573M23.4
UniGene
Hs.50334
Gene Chromosome Location
Chr.9: 34379019-34400833 on Build GRCh38
Transcripts
XM_005251616.4 NM_001184940.1 NM_001184941.1 NM_001184942.1 NM_001184943.1 NM_001184945.1 NM_147202.1 XM_005251402.4 XM_011517797.2 XM_011517798.2 XM_011517799.2

Gene Symbol
FAM219A
Entrez Gene ID
203259
Gene Name
family with sequence similarity 219 member A
Gene Aliases
C9orf25
UniGene
Hs.493771
Gene Chromosome Location
Chr.9: 34398184-34458610 on Build GRCh38
Transcripts
XM_005251616.4 NM_001184940.1 NM_001184941.1 NM_001184942.1 NM_001184943.1 NM_001184945.1 NM_147202.1 XM_005251402.4 XM_011517797.2 XM_011517798.2 XM_011517799.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4338998 c.438delC p.N147fs*8 Chr.9: 34401030-34401030 on Build GRCh38
3413621 c.415G>A p.D139N Chr.9: 34401056-34401056 on Build GRCh38
3907015 c.387C>T p.D129D Chr.9: 34401084-34401084 on Build GRCh38
4815113 c.368T>C p.I123T Chr.9: 34401100-34401100 on Build GRCh38
3907014 c.475G>A p.A159T Chr.9: 34400996-34400996 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762366217 Chr.9: 34400917-34400917 on Build GRCh38
rs373803647 Chr.9: 34401054-34401054 on Build GRCh38
rs772829038 Chr.9: 34400930-34400952 on Build GRCh38
rs928330722 Chr.9: 34400883-34400883 on Build GRCh38
rs368703121 Chr.9: 34401060-34401060 on Build GRCh38
rs773670695 Chr.9: 34400981-34400981 on Build GRCh38
rs761028377 Chr.9: 34400930-34400930 on Build GRCh38
rs943803099 Chr.9: 34400961-34400961 on Build GRCh38
rs117759827 Chr.9: 34400982-34400982 on Build GRCh38
rs948358341 Chr.9: 34400877-34400877 on Build GRCh38
rs752635356 Chr.9: 34400943-34400943 on Build GRCh38
rs907881187 Chr.9: 34400921-34400921 on Build GRCh38
rs748725441 Chr.9: 34400959-34400959 on Build GRCh38
rs747512027 Chr.9: 34401084-34401084 on Build GRCh38
rs940732817 Chr.9: 34400929-34400929 on Build GRCh38
rs996326791 Chr.9: 34400709-34400709 on Build GRCh38
rs557660642 Chr.9: 34400749-34400749 on Build GRCh38
rs778166447 Chr.9: 34401076-34401076 on Build GRCh38
rs957922520 Chr.9: 34400869-34400869 on Build GRCh38
rs751416711 Chr.9: 34401032-34401032 on Build GRCh38
rs990685336 Chr.9: 34400882-34400882 on Build GRCh38
rs755817741 Chr.9: 34401057-34401057 on Build GRCh38
rs371984739 Chr.9: 34400944-34400944 on Build GRCh38
rs980283508 Chr.9: 34401041-34401041 on Build GRCh38
rs1032003881 Chr.9: 34401021-34401021 on Build GRCh38
rs201618351 Chr.9: 34400958-34400958 on Build GRCh38
rs1053477160 Chr.9: 34400916-34400916 on Build GRCh38
rs755713470 Chr.9: 34400955-34400955 on Build GRCh38
rs971223009 Chr.9: 34400900-34400900 on Build GRCh38
rs764008191 Chr.9: 34400947-34400947 on Build GRCh38
rs781107277 Chr.9: 34400816-34400816 on Build GRCh38
rs776821716 Chr.9: 34400935-34400935 on Build GRCh38
rs767301145 Chr.9: 34401053-34401053 on Build GRCh38
rs1032289178 Chr.9: 34400862-34400862 on Build GRCh38
rs202019820 Chr.9: 34401030-34401030 on Build GRCh38
rs1006903644 Chr.9: 34400756-34400756 on Build GRCh38
rs180843824 Chr.9: 34400687-34400687 on Build GRCh38
rs966379082 Chr.9: 34400828-34400828 on Build GRCh38
rs766480938 Chr.9: 34400933-34400933 on Build GRCh38
rs374848079 Chr.9: 34400952-34400952 on Build GRCh38
rs953987782 Chr.9: 34400707-34400707 on Build GRCh38
rs748780577 Chr.9: 34401067-34401067 on Build GRCh38
rs969779459 Chr.9: 34401031-34401031 on Build GRCh38
rs984503208 Chr.9: 34400898-34400898 on Build GRCh38
rs200178804 Chr.9: 34401016-34401016 on Build GRCh38
rs771206276 Chr.9: 34401093-34401093 on Build GRCh38
rs745473472 Chr.9: 34400954-34400954 on Build GRCh38
rs954156803 Chr.9: 34400806-34400806 on Build GRCh38
rs757554867 Chr.9: 34400739-34400739 on Build GRCh38
rs1020065823 Chr.9: 34400759-34400759 on Build GRCh38
rs765262433 Chr.9: 34400941-34400941 on Build GRCh38
rs1012132464 Chr.9: 34401056-34401056 on Build GRCh38
rs772706250 Chr.9: 34400963-34400963 on Build GRCh38
rs953004266 Chr.9: 34400841-34400841 on Build GRCh38
rs916385007 Chr.9: 34400884-34400884 on Build GRCh38
rs1007942548 Chr.9: 34400760-34400760 on Build GRCh38
rs776995760 Chr.9: 34401114-34401114 on Build GRCh38
rs543779902 Chr.9: 34400706-34400706 on Build GRCh38
rs773437600 Chr.9: 34400922-34400922 on Build GRCh38
rs772581463 Chr.9: 34400919-34400919 on Build GRCh38
rs1007018142 Chr.9: 34400940-34400940 on Build GRCh38
rs757021291 Chr.9: 34401036-34401036 on Build GRCh38
rs751307029 Chr.9: 34400948-34400948 on Build GRCh38
rs372298576 Chr.9: 34401072-34401072 on Build GRCh38
rs574243829 Chr.9: 34400863-34400863 on Build GRCh38
rs918748185 Chr.9: 34400833-34400833 on Build GRCh38
rs1040769512 Chr.9: 34400962-34400962 on Build GRCh38
rs775532816 Chr.9: 34401024-34401024 on Build GRCh38
rs1011655104 Chr.9: 34400673-34400673 on Build GRCh38
rs1044034282 Chr.9: 34400880-34400884 on Build GRCh38
rs759689001 Chr.9: 34401015-34401015 on Build GRCh38
rs776751257 Chr.9: 34400984-34400984 on Build GRCh38
rs1037890980 Chr.9: 34400686-34400686 on Build GRCh38
rs760074479 Chr.9: 34401029-34401030 on Build GRCh38
rs982196425 Chr.9: 34400911-34400911 on Build GRCh38
rs999862132 Chr.9: 34400844-34400844 on Build GRCh38

View Full Product Details