Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 30237251-30237793 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAGEB3
Entrez Gene ID
4114
Gene Name
MAGE family member B3
Gene Aliases
CT3.5
UniGene
Hs.113290
Gene Chromosome Location
Chr.X: 30230436-30237493 on Build GRCh38
Transcripts
NM_002365.4 XM_011545513.2 XM_011545514.2

SNP Detail

SNP ID Chromosome Location
rs1004152502 Chr.X: 30237308-30237308 on Build GRCh38
rs930736904 Chr.X: 30237498-30237498 on Build GRCh38
rs762057351 Chr.X: 30237433-30237433 on Build GRCh38
rs1004122809 Chr.X: 30237582-30237582 on Build GRCh38
rs966384239 Chr.X: 30237732-30237732 on Build GRCh38
rs1028587506 Chr.X: 30237690-30237690 on Build GRCh38
rs928182550 Chr.X: 30237760-30237760 on Build GRCh38
rs866306553 Chr.X: 30237306-30237306 on Build GRCh38
rs184341363 Chr.X: 30237435-30237435 on Build GRCh38
rs925897946 Chr.X: 30237532-30237532 on Build GRCh38
rs147318419 Chr.X: 30237331-30237331 on Build GRCh38
rs2529554 Chr.X: 30237403-30237403 on Build GRCh38
rs987522812 Chr.X: 30237617-30237617 on Build GRCh38
rs560625752 Chr.X: 30237470-30237470 on Build GRCh38
rs914004762 Chr.X: 30237373-30237373 on Build GRCh38
rs901091286 Chr.X: 30237634-30237634 on Build GRCh38
rs980014875 Chr.X: 30237525-30237525 on Build GRCh38
rs959623820 Chr.X: 30237771-30237771 on Build GRCh38
rs1011095674 Chr.X: 30237726-30237726 on Build GRCh38
rs1023944549 Chr.X: 30237491-30237492 on Build GRCh38
rs770846385 Chr.X: 30237536-30237536 on Build GRCh38
rs913081187 Chr.X: 30237614-30237614 on Build GRCh38
rs755825726 Chr.X: 30237366-30237366 on Build GRCh38
rs957787121 Chr.X: 30237720-30237720 on Build GRCh38
rs1041020431 Chr.X: 30237632-30237632 on Build GRCh38
rs1039696317 Chr.X: 30237432-30237432 on Build GRCh38
rs776766351 Chr.X: 30237564-30237564 on Build GRCh38
rs767916315 Chr.X: 30237458-30237458 on Build GRCh38
rs957217094 Chr.X: 30237545-30237545 on Build GRCh38
rs899328312 Chr.X: 30237434-30237434 on Build GRCh38
rs938222272 Chr.X: 30237481-30237481 on Build GRCh38
rs935874120 Chr.X: 30237347-30237347 on Build GRCh38
rs759809495 Chr.X: 30237662-30237662 on Build GRCh38
rs890468595 Chr.X: 30237420-30237423 on Build GRCh38
rs774120030 Chr.X: 30237406-30237406 on Build GRCh38
rs943411779 Chr.X: 30237627-30237627 on Build GRCh38
rs886616279 Chr.X: 30237756-30237756 on Build GRCh38
rs749072751 Chr.X: 30237390-30237390 on Build GRCh38
rs866942688 Chr.X: 30237381-30237381 on Build GRCh38
rs529811313 Chr.X: 30237321-30237321 on Build GRCh38
rs780055880 Chr.X: 30237387-30237387 on Build GRCh38
rs751411890 Chr.X: 30237424-30237424 on Build GRCh38
rs886464766 Chr.X: 30237579-30237579 on Build GRCh38
rs750513895 Chr.X: 30237483-30237483 on Build GRCh38
rs780911680 Chr.X: 30237286-30237286 on Build GRCh38
rs866339261 Chr.X: 30237604-30237604 on Build GRCh38
rs975156208 Chr.X: 30237359-30237359 on Build GRCh38
rs930809116 Chr.X: 30237746-30237746 on Build GRCh38
rs188034121 Chr.X: 30237749-30237749 on Build GRCh38
rs995448646 Chr.X: 30237436-30237436 on Build GRCh38
rs920657803 Chr.X: 30237489-30237489 on Build GRCh38
rs911853098 Chr.X: 30237622-30237622 on Build GRCh38
rs1020605194 Chr.X: 30237727-30237727 on Build GRCh38
rs945431827 Chr.X: 30237615-30237615 on Build GRCh38

View Full Product Details