Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 111689908-111690406 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALG13
Entrez Gene ID
79868
Gene Name
ALG13, UDP-N-acetylglucosaminyltransferase subunit
Gene Aliases
CDG1S, CXorf45, EIEE36, GLT28D1, MDS031, TDRD13, YGL047W
UniGene
Hs.443061
Gene Chromosome Location
Chr.X: 111680741-111764164 on Build GRCh38
Transcripts
NM_001099922.2 NM_001257230.1 NM_001257231.1 NM_001257234.1 NM_001257237.1 NM_001324292.1 NM_001324293.1 XM_006724693.2 XM_006724697.3 XM_006724698.3 XM_011531028.2 XM_011531030.1 XM_011531031.1 XM_011531032.1 XM_011531033.2 XM_011531034.2 XM_017029846.1 XM_017029847.1 XM_017029848.1 XM_017029849.1 XM_017029850.1 XM_017029851.1

SNP Detail

SNP ID Chromosome Location
rs1015384992 Chr.X: 111690039-111690039 on Build GRCh38
rs774290637 Chr.X: 111690317-111690318 on Build GRCh38
rs965813210 Chr.X: 111690195-111690195 on Build GRCh38
rs768436536 Chr.X: 111690135-111690135 on Build GRCh38
rs886064652 Chr.X: 111690130-111690130 on Build GRCh38
rs113017653 Chr.X: 111690333-111690333 on Build GRCh38
rs181356795 Chr.X: 111690246-111690246 on Build GRCh38
rs973924813 Chr.X: 111690125-111690127 on Build GRCh38
rs375772513 Chr.X: 111690083-111690083 on Build GRCh38
rs1044585507 Chr.X: 111690331-111690331 on Build GRCh38
rs906087927 Chr.X: 111690332-111690332 on Build GRCh38
rs960695122 Chr.X: 111689998-111689998 on Build GRCh38
rs145446865 Chr.X: 111690037-111690037 on Build GRCh38
rs1003970593 Chr.X: 111690014-111690014 on Build GRCh38
rs1018676592 Chr.X: 111690142-111690142 on Build GRCh38
rs914683788 Chr.X: 111690296-111690296 on Build GRCh38
rs967761048 Chr.X: 111690357-111690357 on Build GRCh38
rs981481298 Chr.X: 111690361-111690361 on Build GRCh38
rs898746161 Chr.X: 111690018-111690018 on Build GRCh38
rs1015332858 Chr.X: 111690365-111690365 on Build GRCh38
rs987182696 Chr.X: 111690213-111690213 on Build GRCh38
rs189001263 Chr.X: 111689948-111689948 on Build GRCh38
rs753756636 Chr.X: 111690326-111690326 on Build GRCh38
rs778874509 Chr.X: 111690236-111690236 on Build GRCh38
rs766868432 Chr.X: 111690051-111690051 on Build GRCh38
rs1047316392 Chr.X: 111690123-111690123 on Build GRCh38
rs962532480 Chr.X: 111690087-111690087 on Build GRCh38
rs1056497488 Chr.X: 111690349-111690349 on Build GRCh38
rs780111545 Chr.X: 111690291-111690291 on Build GRCh38
rs369240278 Chr.X: 111690292-111690292 on Build GRCh38
rs138165282 Chr.X: 111690285-111690285 on Build GRCh38
rs921166041 Chr.X: 111690133-111690133 on Build GRCh38
rs779293733 Chr.X: 111690344-111690344 on Build GRCh38
rs913151563 Chr.X: 111690354-111690354 on Build GRCh38

View Full Product Details