Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 197153590-197154126 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZBTB41
Entrez Gene ID
360023
Gene Name
zinc finger and BTB domain containing 41
Gene Aliases
FRBZ1, ZNF924
UniGene
Hs.529439
Gene Chromosome Location
Chr.1: 197153682-197200542 on Build GRCh38
Transcripts
NM_194314.2 NR_135153.1

SNP Detail

SNP ID Chromosome Location
rs982152735 Chr.1: 197153827-197153827 on Build GRCh38
rs571203631 Chr.1: 197154037-197154037 on Build GRCh38
rs547903323 Chr.1: 197153833-197153833 on Build GRCh38
rs941163625 Chr.1: 197153928-197153928 on Build GRCh38
rs998084286 Chr.1: 197153960-197153960 on Build GRCh38
rs1022004031 Chr.1: 197154027-197154027 on Build GRCh38
rs944896093 Chr.1: 197154022-197154022 on Build GRCh38
rs1044478269 Chr.1: 197153849-197153849 on Build GRCh38
rs537935128 Chr.1: 197153763-197153763 on Build GRCh38
rs1050786576 Chr.1: 197153989-197153989 on Build GRCh38
rs1127661 Chr.1: 197153913-197153913 on Build GRCh38
rs988186870 Chr.1: 197153851-197153851 on Build GRCh38
rs1050676341 Chr.1: 197153975-197153975 on Build GRCh38
rs946913777 Chr.1: 197153958-197153958 on Build GRCh38
rs1036968934 Chr.1: 197153941-197153941 on Build GRCh38
rs897102803 Chr.1: 197153949-197153949 on Build GRCh38
rs952632078 Chr.1: 197153646-197153646 on Build GRCh38
rs969307311 Chr.1: 197154029-197154029 on Build GRCh38
rs889450244 Chr.1: 197154014-197154014 on Build GRCh38
rs924840276 Chr.1: 197153697-197153697 on Build GRCh38
rs865816580 Chr.1: 197154066-197154066 on Build GRCh38
rs1031789681 Chr.1: 197154079-197154079 on Build GRCh38
rs900951269 Chr.1: 197154057-197154057 on Build GRCh38
rs376745824 Chr.1: 197154077-197154077 on Build GRCh38
rs1025971031 Chr.1: 197153627-197153627 on Build GRCh38
rs984020099 Chr.1: 197153654-197153654 on Build GRCh38
rs1000289961 Chr.1: 197154041-197154041 on Build GRCh38
rs35022190 Chr.1: 197153637-197153637 on Build GRCh38
rs1040797985 Chr.1: 197154023-197154023 on Build GRCh38
rs139903729 Chr.1: 197153830-197153830 on Build GRCh38
rs956055552 Chr.1: 197153812-197153812 on Build GRCh38
rs948857983 Chr.1: 197153840-197153841 on Build GRCh38
rs956316532 Chr.1: 197154096-197154096 on Build GRCh38
rs574443642 Chr.1: 197153737-197153737 on Build GRCh38
rs145970451 Chr.1: 197153675-197153675 on Build GRCh38
rs935308625 Chr.1: 197153836-197153836 on Build GRCh38
rs868007148 Chr.1: 197153821-197153821 on Build GRCh38
rs538221452 Chr.1: 197154063-197154063 on Build GRCh38
rs889281937 Chr.1: 197153995-197153995 on Build GRCh38

View Full Product Details