Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 228175794-228176287 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IBA57
Entrez Gene ID
200205
Gene Name
IBA57 homolog, iron-sulfur cluster assembly
Gene Aliases
C1orf69, MMDS3, SPG74
UniGene
Hs.237017
Gene Chromosome Location
Chr.1: 228165808-228182257 on Build GRCh38
Transcripts
NM_001010867.3 NM_001310327.1

SNP Detail

SNP ID Chromosome Location
rs527945729 Chr.1: 228176174-228176174 on Build GRCh38
rs922809270 Chr.1: 228175976-228175976 on Build GRCh38
rs544331995 Chr.1: 228176055-228176055 on Build GRCh38
rs73095429 Chr.1: 228176173-228176173 on Build GRCh38
rs975707127 Chr.1: 228175975-228175975 on Build GRCh38
rs1044971996 Chr.1: 228176045-228176045 on Build GRCh38
rs915949448 Chr.1: 228176205-228176205 on Build GRCh38
rs895066457 Chr.1: 228175888-228175888 on Build GRCh38
rs759597339 Chr.1: 228175946-228175946 on Build GRCh38
rs773128253 Chr.1: 228176180-228176181 on Build GRCh38
rs558008552 Chr.1: 228175824-228175824 on Build GRCh38
rs963747278 Chr.1: 228176119-228176119 on Build GRCh38
rs187699240 Chr.1: 228175914-228175914 on Build GRCh38
rs571433754 Chr.1: 228175871-228175871 on Build GRCh38
rs996479997 Chr.1: 228176137-228176137 on Build GRCh38
rs538035093 Chr.1: 228175821-228175821 on Build GRCh38
rs561901621 Chr.1: 228175816-228175816 on Build GRCh38
rs964018766 Chr.1: 228175943-228175943 on Build GRCh38
rs1029324367 Chr.1: 228176155-228176155 on Build GRCh38
rs982981896 Chr.1: 228175982-228175982 on Build GRCh38
rs545175202 Chr.1: 228176094-228176094 on Build GRCh38
rs534107767 Chr.1: 228175882-228175882 on Build GRCh38
rs894495607 Chr.1: 228176018-228176018 on Build GRCh38
rs934155187 Chr.1: 228175980-228175980 on Build GRCh38
rs974187751 Chr.1: 228176191-228176191 on Build GRCh38
rs906342346 Chr.1: 228175928-228175928 on Build GRCh38
rs941484757 Chr.1: 228176006-228176006 on Build GRCh38
rs948690316 Chr.1: 228176038-228176038 on Build GRCh38
rs1012179248 Chr.1: 228175829-228175829 on Build GRCh38
rs1055730689 Chr.1: 228176089-228176089 on Build GRCh38
rs1023485147 Chr.1: 228175846-228175846 on Build GRCh38
rs888205402 Chr.1: 228176190-228176190 on Build GRCh38
rs146154211 Chr.1: 228176009-228176009 on Build GRCh38
rs1038397706 Chr.1: 228175820-228175820 on Build GRCh38
rs183545324 Chr.1: 228176061-228176061 on Build GRCh38
rs946143372 Chr.1: 228175815-228175815 on Build GRCh38
rs778640099 Chr.1: 228175896-228175896 on Build GRCh38
rs1021989018 Chr.1: 228175930-228175930 on Build GRCh38
rs963180005 Chr.1: 228176181-228176181 on Build GRCh38
rs1003414709 Chr.1: 228176067-228176067 on Build GRCh38
rs372467645 Chr.1: 228175925-228175925 on Build GRCh38
rs1046333168 Chr.1: 228176124-228176124 on Build GRCh38
rs747548450 Chr.1: 228176148-228176148 on Build GRCh38
rs892279397 Chr.1: 228176091-228176091 on Build GRCh38
rs776902374 Chr.1: 228176098-228176098 on Build GRCh38
rs113813514 Chr.1: 228175915-228175915 on Build GRCh38
rs1045749428 Chr.1: 228176246-228176246 on Build GRCh38
rs1010668322 Chr.1: 228176097-228176097 on Build GRCh38
rs11555655 Chr.1: 228176049-228176049 on Build GRCh38
rs776707795 Chr.1: 228176231-228176231 on Build GRCh38
rs187731849 Chr.1: 228176260-228176260 on Build GRCh38
rs1030818156 Chr.1: 228176078-228176078 on Build GRCh38
rs191426321 Chr.1: 228176002-228176002 on Build GRCh38
rs955646512 Chr.1: 228176169-228176169 on Build GRCh38
rs989615197 Chr.1: 228175926-228175926 on Build GRCh38
rs964305042 Chr.1: 228175987-228175987 on Build GRCh38
rs1022437594 Chr.1: 228176099-228176099 on Build GRCh38
rs772460020 Chr.1: 228175993-228175993 on Build GRCh38

View Full Product Details