Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 24591790-24592335 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP21
Entrez Gene ID
57584
Gene Name
Rho GTPase activating protein 21
Gene Aliases
ARHGAP10
UniGene
Hs.524195
Gene Chromosome Location
Chr.10: 24583609-24725421 on Build GRCh38
Transcripts
NM_020824.3 XM_005252542.3 XM_005252544.4 XM_011519602.2 XM_011519603.2 XM_011519604.2 XM_011519605.2 XM_011519606.2 XM_011519607.2 XM_017016459.1 XM_017016460.1 XM_017016461.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1128080 c.3978T>C p.I1326I Chr.10: 24591908-24591908 on Build GRCh38
7311911 c.3301A>T p.M1101L Chr.10: 24591949-24591949 on Build GRCh38
917152 c.3919C>T p.R1307* Chr.10: 24591967-24591967 on Build GRCh38
4870029 c.3273C>A p.P1091P Chr.10: 24591977-24591977 on Build GRCh38
4546123 c.3931G>A p.D1311N Chr.10: 24591955-24591955 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs981473260 Chr.10: 24592254-24592254 on Build GRCh38
rs1036874560 Chr.10: 24592099-24592099 on Build GRCh38
rs573886778 Chr.10: 24592288-24592288 on Build GRCh38
rs376841445 Chr.10: 24591878-24591878 on Build GRCh38
rs751183813 Chr.10: 24592062-24592062 on Build GRCh38
rs1036358609 Chr.10: 24592283-24592283 on Build GRCh38
rs766850163 Chr.10: 24592158-24592159 on Build GRCh38
rs759702894 Chr.10: 24591995-24591995 on Build GRCh38
rs528954452 Chr.10: 24592199-24592199 on Build GRCh38
rs767250188 Chr.10: 24592061-24592062 on Build GRCh38
rs535710372 Chr.10: 24592132-24592132 on Build GRCh38
rs759825841 Chr.10: 24592157-24592159 on Build GRCh38
rs979379701 Chr.10: 24591931-24591931 on Build GRCh38
rs754115677 Chr.10: 24591844-24591864 on Build GRCh38
rs879046932 Chr.10: 24592155-24592155 on Build GRCh38
rs1009909908 Chr.10: 24592184-24592184 on Build GRCh38
rs11014168 Chr.10: 24592305-24592305 on Build GRCh38
rs940655430 Chr.10: 24592253-24592253 on Build GRCh38
rs748390271 Chr.10: 24592251-24592251 on Build GRCh38
rs7904331 Chr.10: 24592242-24592242 on Build GRCh38
rs373696545 Chr.10: 24591932-24591932 on Build GRCh38
rs997212843 Chr.10: 24592200-24592200 on Build GRCh38
rs748010884 Chr.10: 24591912-24591912 on Build GRCh38
rs754432618 Chr.10: 24592030-24592030 on Build GRCh38
rs561488306 Chr.10: 24592065-24592065 on Build GRCh38
rs1020384412 Chr.10: 24592249-24592249 on Build GRCh38
rs550394307 Chr.10: 24592025-24592025 on Build GRCh38
rs774503483 Chr.10: 24591855-24591855 on Build GRCh38
rs570002645 Chr.10: 24592104-24592104 on Build GRCh38
rs577432756 Chr.10: 24592219-24592220 on Build GRCh38
rs764779664 Chr.10: 24591870-24591870 on Build GRCh38
rs937121346 Chr.10: 24592131-24592138 on Build GRCh38
rs201592348 Chr.10: 24592222-24592223 on Build GRCh38
rs371918980 Chr.10: 24591873-24591873 on Build GRCh38
rs769573845 Chr.10: 24591843-24591844 on Build GRCh38
rs780813035 Chr.10: 24592257-24592258 on Build GRCh38
rs754752303 Chr.10: 24591899-24591899 on Build GRCh38
rs761444076 Chr.10: 24592060-24592061 on Build GRCh38
rs369040406 Chr.10: 24591879-24591879 on Build GRCh38
rs766610619 Chr.10: 24591844-24591844 on Build GRCh38
rs776004603 Chr.10: 24592002-24592002 on Build GRCh38
rs891386540 Chr.10: 24592226-24592226 on Build GRCh38
rs148522047 Chr.10: 24591887-24591887 on Build GRCh38
rs1035456080 Chr.10: 24592156-24592156 on Build GRCh38
rs922032807 Chr.10: 24591940-24591940 on Build GRCh38
rs542955760 Chr.10: 24591825-24591825 on Build GRCh38
rs753210280 Chr.10: 24591866-24591866 on Build GRCh38
rs763783858 Chr.10: 24592039-24592039 on Build GRCh38
rs767519589 Chr.10: 24591821-24591821 on Build GRCh38
rs1048736610 Chr.10: 24592129-24592129 on Build GRCh38
rs775525101 Chr.10: 24592015-24592016 on Build GRCh38
rs768161562 Chr.10: 24592032-24592048 on Build GRCh38
rs940710546 Chr.10: 24592078-24592078 on Build GRCh38
rs765828488 Chr.10: 24592057-24592057 on Build GRCh38
rs764691475 Chr.10: 24592028-24592028 on Build GRCh38
rs759918243 Chr.10: 24591857-24591857 on Build GRCh38
rs918501090 Chr.10: 24592289-24592289 on Build GRCh38
rs756722137 Chr.10: 24591869-24591869 on Build GRCh38
rs780878754 Chr.10: 24591908-24591908 on Build GRCh38
rs995753207 Chr.10: 24591815-24591815 on Build GRCh38
rs1020547004 Chr.10: 24592187-24592187 on Build GRCh38
rs767886454 Chr.10: 24592001-24592001 on Build GRCh38
rs191776185 Chr.10: 24591841-24591841 on Build GRCh38
rs533340791 Chr.10: 24591949-24591949 on Build GRCh38
rs369840767 Chr.10: 24591858-24591858 on Build GRCh38
rs1046037195 Chr.10: 24592122-24592122 on Build GRCh38
rs78989166 Chr.10: 24592221-24592222 on Build GRCh38
rs1005965625 Chr.10: 24592151-24592151 on Build GRCh38
rs183466982 Chr.10: 24592248-24592248 on Build GRCh38
rs201654486 Chr.10: 24592181-24592181 on Build GRCh38
rs142895527 Chr.10: 24591914-24591914 on Build GRCh38
rs759134728 Chr.10: 24591816-24591816 on Build GRCh38
rs928501122 Chr.10: 24592258-24592260 on Build GRCh38
rs527710987 Chr.10: 24591864-24591864 on Build GRCh38
rs1036144441 Chr.10: 24592156-24592157 on Build GRCh38
rs1051699559 Chr.10: 24592006-24592006 on Build GRCh38
rs377468496 Chr.10: 24591944-24591944 on Build GRCh38
rs773920607 Chr.10: 24592058-24592059 on Build GRCh38
rs568662271 Chr.10: 24592149-24592149 on Build GRCh38
rs57846258 Chr.10: 24592181-24592181 on Build GRCh38
rs995346665 Chr.10: 24592123-24592123 on Build GRCh38
rs543243539 Chr.10: 24592235-24592235 on Build GRCh38
rs369212427 Chr.10: 24592214-24592214 on Build GRCh38
rs61735164 Chr.10: 24591941-24591941 on Build GRCh38
rs932088147 Chr.10: 24591997-24591997 on Build GRCh38
rs911871259 Chr.10: 24592068-24592068 on Build GRCh38
rs151034402 Chr.10: 24592209-24592209 on Build GRCh38
rs748742571 Chr.10: 24592031-24592047 on Build GRCh38
rs577714588 Chr.10: 24592232-24592232 on Build GRCh38
rs534649433 Chr.10: 24592192-24592192 on Build GRCh38
rs970249073 Chr.10: 24591819-24591819 on Build GRCh38
rs879093531 Chr.10: 24592139-24592139 on Build GRCh38

View Full Product Details