Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 102676424-102676933 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL3
Entrez Gene ID
403
Gene Name
ADP ribosylation factor like GTPase 3
Gene Aliases
ARFL3
UniGene
Hs.182215
Gene Chromosome Location
Chr.10: 102673727-102714433 on Build GRCh38
Transcripts
NM_004311.3 XM_017016260.1

SNP Detail

SNP ID Chromosome Location
rs768616247 Chr.10: 102676849-102676849 on Build GRCh38
rs562965136 Chr.10: 102676791-102676791 on Build GRCh38
rs951369113 Chr.10: 102676841-102676841 on Build GRCh38
rs565272228 Chr.10: 102676724-102676724 on Build GRCh38
rs757742992 Chr.10: 102676536-102676538 on Build GRCh38
rs374666203 Chr.10: 102676495-102676495 on Build GRCh38
rs761758077 Chr.10: 102676863-102676863 on Build GRCh38
rs574465819 Chr.10: 102676531-102676531 on Build GRCh38
rs8354 Chr.10: 102676884-102676884 on Build GRCh38
rs1054030329 Chr.10: 102676603-102676603 on Build GRCh38
rs990712196 Chr.10: 102676642-102676642 on Build GRCh38
rs376548476 Chr.10: 102676908-102676908 on Build GRCh38
rs940181014 Chr.10: 102676820-102676820 on Build GRCh38
rs889466299 Chr.10: 102676485-102676485 on Build GRCh38
rs745485828 Chr.10: 102676885-102676885 on Build GRCh38
rs571756029 Chr.10: 102676707-102676707 on Build GRCh38
rs765711720 Chr.10: 102676873-102676873 on Build GRCh38
rs139241561 Chr.10: 102676708-102676708 on Build GRCh38
rs758778515 Chr.10: 102676881-102676881 on Build GRCh38
rs981334479 Chr.10: 102676876-102676876 on Build GRCh38
rs764162525 Chr.10: 102676819-102676819 on Build GRCh38
rs917992800 Chr.10: 102676857-102676857 on Build GRCh38
rs960624127 Chr.10: 102676618-102676618 on Build GRCh38
rs41308675 Chr.10: 102676762-102676762 on Build GRCh38
rs748788457 Chr.10: 102676848-102676848 on Build GRCh38
rs910001826 Chr.10: 102676804-102676804 on Build GRCh38
rs1022899074 Chr.10: 102676696-102676697 on Build GRCh38
rs966930476 Chr.10: 102676529-102676532 on Build GRCh38
rs884550 Chr.10: 102676856-102676856 on Build GRCh38
rs548554855 Chr.10: 102676840-102676840 on Build GRCh38
rs1010635979 Chr.10: 102676508-102676508 on Build GRCh38
rs562021997 Chr.10: 102676533-102676539 on Build GRCh38
rs1009913311 Chr.10: 102676645-102676645 on Build GRCh38
rs532717608 Chr.10: 102676761-102676761 on Build GRCh38
rs905437142 Chr.10: 102676470-102676470 on Build GRCh38
rs1019997449 Chr.10: 102676672-102676672 on Build GRCh38
rs559376946 Chr.10: 102676525-102676530 on Build GRCh38
rs755030182 Chr.10: 102676864-102676864 on Build GRCh38
rs769940148 Chr.10: 102676615-102676615 on Build GRCh38
rs112537748 Chr.10: 102676851-102676851 on Build GRCh38
rs566866967 Chr.10: 102676850-102676850 on Build GRCh38
rs936952532 Chr.10: 102676484-102676484 on Build GRCh38
rs145965633 Chr.10: 102676799-102676800 on Build GRCh38
rs541771688 Chr.10: 102676544-102676544 on Build GRCh38
rs553356389 Chr.10: 102676654-102676654 on Build GRCh38
rs539749953 Chr.10: 102676446-102676446 on Build GRCh38
rs530144293 Chr.10: 102676814-102676814 on Build GRCh38
rs775565493 Chr.10: 102676729-102676729 on Build GRCh38

View Full Product Details