Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 1753913-1754333 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTSD
Entrez Gene ID
1509
Gene Name
cathepsin D
Gene Aliases
CLN10, CPSD, HEL-S-130P
UniGene
Hs.654447
Gene Chromosome Location
Chr.11: 1752752-1763992 on Build GRCh38
Transcripts
NM_001909.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6637781 c.938A>C p.K313T Chr.11: 1754028-1754028 on Build GRCh38
7226662 c.951C>T p.A317A Chr.11: 1754015-1754015 on Build GRCh38
5818236 c.908G>T p.G303V Chr.11: 1754058-1754058 on Build GRCh38
6768742 c.957G>A p.P319P Chr.11: 1754009-1754009 on Build GRCh38
925521 c.956C>T p.P319L Chr.11: 1754010-1754010 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373699222 Chr.11: 1754038-1754038 on Build GRCh38
rs1014674908 Chr.11: 1754117-1754117 on Build GRCh38
rs376034984 Chr.11: 1753996-1753996 on Build GRCh38
rs765320531 Chr.11: 1753988-1753988 on Build GRCh38
rs749749338 Chr.11: 1753958-1753969 on Build GRCh38
rs112021026 Chr.11: 1754120-1754120 on Build GRCh38
rs760348359 Chr.11: 1753951-1753951 on Build GRCh38
rs762444592 Chr.11: 1754143-1754143 on Build GRCh38
rs780328613 Chr.11: 1754055-1754055 on Build GRCh38
rs886048065 Chr.11: 1754026-1754026 on Build GRCh38
rs375565948 Chr.11: 1754170-1754170 on Build GRCh38
rs748541999 Chr.11: 1754170-1754170 on Build GRCh38
rs950725235 Chr.11: 1754065-1754065 on Build GRCh38
rs556326445 Chr.11: 1754186-1754186 on Build GRCh38
rs78306946 Chr.11: 1754015-1754015 on Build GRCh38
rs763652137 Chr.11: 1754145-1754145 on Build GRCh38
rs569157908 Chr.11: 1754269-1754269 on Build GRCh38
rs772347858 Chr.11: 1754091-1754091 on Build GRCh38
rs772363414 Chr.11: 1754174-1754175 on Build GRCh38
rs913643339 Chr.11: 1754164-1754164 on Build GRCh38
rs1010417289 Chr.11: 1754210-1754210 on Build GRCh38
rs770520646 Chr.11: 1753957-1753958 on Build GRCh38
rs773433974 Chr.11: 1754192-1754193 on Build GRCh38
rs956390748 Chr.11: 1754203-1754203 on Build GRCh38
rs758724821 Chr.11: 1754274-1754274 on Build GRCh38
rs1051758705 Chr.11: 1754063-1754063 on Build GRCh38
rs764766572 Chr.11: 1754020-1754020 on Build GRCh38
rs761701186 Chr.11: 1753945-1753945 on Build GRCh38
rs149427480 Chr.11: 1754124-1754124 on Build GRCh38
rs766377846 Chr.11: 1754262-1754263 on Build GRCh38
rs764812657 Chr.11: 1754135-1754135 on Build GRCh38
rs373170074 Chr.11: 1754010-1754010 on Build GRCh38
rs759180698 Chr.11: 1754128-1754128 on Build GRCh38
rs755183661 Chr.11: 1754075-1754075 on Build GRCh38
rs752172496 Chr.11: 1754021-1754021 on Build GRCh38
rs759808749 Chr.11: 1754246-1754246 on Build GRCh38
rs940679941 Chr.11: 1754308-1754308 on Build GRCh38
rs913538989 Chr.11: 1753981-1753981 on Build GRCh38
rs748185051 Chr.11: 1754089-1754089 on Build GRCh38
rs762642229 Chr.11: 1754214-1754214 on Build GRCh38
rs905901195 Chr.11: 1754307-1754308 on Build GRCh38
rs756650281 Chr.11: 1754153-1754153 on Build GRCh38
rs750270880 Chr.11: 1753950-1753950 on Build GRCh38
rs757738328 Chr.11: 1753982-1753982 on Build GRCh38
rs969307077 Chr.11: 1754026-1754026 on Build GRCh38
rs1014376434 Chr.11: 1754279-1754279 on Build GRCh38
rs778999077 Chr.11: 1754084-1754084 on Build GRCh38
rs970547833 Chr.11: 1753949-1753949 on Build GRCh38
rs922157971 Chr.11: 1754253-1754253 on Build GRCh38
rs780882378 Chr.11: 1753958-1753963 on Build GRCh38
rs773878052 Chr.11: 1754217-1754217 on Build GRCh38
rs774327757 Chr.11: 1753936-1753936 on Build GRCh38
rs754437898 Chr.11: 1754238-1754238 on Build GRCh38
rs78735768 Chr.11: 1754155-1754155 on Build GRCh38
rs745653205 Chr.11: 1754198-1754198 on Build GRCh38
rs774332555 Chr.11: 1753997-1753997 on Build GRCh38
rs531682785 Chr.11: 1753984-1753984 on Build GRCh38
rs766136679 Chr.11: 1753959-1753959 on Build GRCh38
rs778336438 Chr.11: 1753976-1753976 on Build GRCh38
rs140238987 Chr.11: 1754054-1754054 on Build GRCh38
rs1047769384 Chr.11: 1754245-1754245 on Build GRCh38
rs1053497172 Chr.11: 1753968-1753968 on Build GRCh38
rs776324764 Chr.11: 1754014-1754014 on Build GRCh38
rs553734441 Chr.11: 1754234-1754234 on Build GRCh38
rs752075136 Chr.11: 1753980-1753980 on Build GRCh38
rs369991128 Chr.11: 1754073-1754073 on Build GRCh38
rs746243061 Chr.11: 1753985-1753985 on Build GRCh38
rs778040768 Chr.11: 1754177-1754177 on Build GRCh38
rs752940637 Chr.11: 1754270-1754270 on Build GRCh38
rs750903034 Chr.11: 1754148-1754148 on Build GRCh38
rs776321697 Chr.11: 1754123-1754123 on Build GRCh38
rs754501335 Chr.11: 1753971-1753971 on Build GRCh38
rs35604250 Chr.11: 1754139-1754139 on Build GRCh38
rs377228580 Chr.11: 1754156-1754156 on Build GRCh38
rs748443280 Chr.11: 1754162-1754162 on Build GRCh38
rs749491748 Chr.11: 1754062-1754062 on Build GRCh38
rs945026250 Chr.11: 1754169-1754169 on Build GRCh38
rs773273362 Chr.11: 1754103-1754103 on Build GRCh38
rs769301773 Chr.11: 1753979-1753984 on Build GRCh38
rs911866642 Chr.11: 1754296-1754296 on Build GRCh38
rs911925676 Chr.11: 1754069-1754069 on Build GRCh38
rs760917550 Chr.11: 1754199-1754212 on Build GRCh38
rs987533361 Chr.11: 1754176-1754176 on Build GRCh38
rs1034628151 Chr.11: 1754189-1754189 on Build GRCh38
rs776431760 Chr.11: 1754192-1754192 on Build GRCh38
rs761766384 Chr.11: 1753998-1753998 on Build GRCh38
rs200047861 Chr.11: 1754157-1754157 on Build GRCh38
rs767386720 Chr.11: 1753946-1753946 on Build GRCh38
rs372319807 Chr.11: 1754137-1754137 on Build GRCh38
rs147278302 Chr.11: 1754122-1754122 on Build GRCh38
rs183101544 Chr.11: 1754303-1754303 on Build GRCh38
rs774781089 Chr.11: 1754014-1754014 on Build GRCh38
rs369445669 Chr.11: 1753962-1753962 on Build GRCh38
rs374540411 Chr.11: 1754040-1754040 on Build GRCh38
rs371034011 Chr.11: 1754142-1754142 on Build GRCh38
rs768118630 Chr.11: 1754215-1754215 on Build GRCh38
rs1024726028 Chr.11: 1754293-1754293 on Build GRCh38
rs779000770 Chr.11: 1754158-1754158 on Build GRCh38
rs537939304 Chr.11: 1754183-1754183 on Build GRCh38
rs761119483 Chr.11: 1754231-1754231 on Build GRCh38
rs538647820 Chr.11: 1754212-1754212 on Build GRCh38
rs369229897 Chr.11: 1754009-1754009 on Build GRCh38
rs371747998 Chr.11: 1753987-1753987 on Build GRCh38
rs766338105 Chr.11: 1754013-1754013 on Build GRCh38
rs768684076 Chr.11: 1753935-1753935 on Build GRCh38
rs772151839 Chr.11: 1754003-1754003 on Build GRCh38
rs577674691 Chr.11: 1754208-1754208 on Build GRCh38
rs796052395 Chr.11: 1754151-1754152 on Build GRCh38
rs750994475 Chr.11: 1754053-1754053 on Build GRCh38

View Full Product Details