Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 61349984-61350479 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYB561A3
Entrez Gene ID
220002
Gene Name
cytochrome b561 family member A3
Gene Aliases
CYBASC3, LCYTB
UniGene
Hs.22546
Gene Chromosome Location
Chr.11: 61348748-61362299 on Build GRCh38
Transcripts
NM_001161452.1 NM_001161454.1 NM_001300763.1 NM_153611.4 XM_011544821.1 XM_011544823.1 XM_017017345.1 XM_017017346.1 XM_017017347.1 XM_017017518.1 XM_017017522.1

Gene Symbol
TKFC
Entrez Gene ID
26007
Gene Name
triokinase and FMN cyclase
Gene Aliases
DAK, NET45
UniGene
Hs.6278
Gene Chromosome Location
Chr.11: 61333182-61353426 on Build GRCh38
Transcripts
NM_001161452.1 NM_001161454.1 NM_001300763.1 NM_153611.4 XM_011544821.1 XM_011544823.1 XM_017017345.1 XM_017017346.1 XM_017017347.1 XM_017017518.1 XM_017017522.1

SNP Detail

SNP ID Chromosome Location
rs955909862 Chr.11: 61350444-61350444 on Build GRCh38
rs113025505 Chr.11: 61350177-61350177 on Build GRCh38
rs981467823 Chr.11: 61350218-61350218 on Build GRCh38
rs767292537 Chr.11: 61350360-61350360 on Build GRCh38
rs373552140 Chr.11: 61350318-61350318 on Build GRCh38
rs770375221 Chr.11: 61350425-61350425 on Build GRCh38
rs905967913 Chr.11: 61350391-61350391 on Build GRCh38
rs746571098 Chr.11: 61350244-61350244 on Build GRCh38
rs897008193 Chr.11: 61350255-61350255 on Build GRCh38
rs370802615 Chr.11: 61350317-61350317 on Build GRCh38
rs1012087347 Chr.11: 61350300-61350300 on Build GRCh38
rs748323589 Chr.11: 61350441-61350441 on Build GRCh38
rs955961394 Chr.11: 61350240-61350240 on Build GRCh38
rs957075385 Chr.11: 61350111-61350111 on Build GRCh38
rs974896577 Chr.11: 61350272-61350272 on Build GRCh38
rs549420104 Chr.11: 61350432-61350432 on Build GRCh38
rs756810006 Chr.11: 61350251-61350251 on Build GRCh38
rs531913173 Chr.11: 61350180-61350180 on Build GRCh38
rs376788904 Chr.11: 61350385-61350385 on Build GRCh38
rs940284767 Chr.11: 61350362-61350362 on Build GRCh38
rs780846467 Chr.11: 61350422-61350422 on Build GRCh38
rs574334638 Chr.11: 61350053-61350053 on Build GRCh38
rs540195847 Chr.11: 61350034-61350034 on Build GRCh38
rs113361441 Chr.11: 61350186-61350186 on Build GRCh38
rs11230671 Chr.11: 61350141-61350141 on Build GRCh38
rs1012214889 Chr.11: 61350050-61350050 on Build GRCh38
rs755272722 Chr.11: 61350426-61350426 on Build GRCh38
rs200397941 Chr.11: 61350414-61350414 on Build GRCh38
rs900169920 Chr.11: 61350046-61350046 on Build GRCh38
rs752356292 Chr.11: 61350373-61350373 on Build GRCh38
rs891406272 Chr.11: 61350062-61350062 on Build GRCh38
rs376630188 Chr.11: 61350127-61350127 on Build GRCh38
rs147478513 Chr.11: 61350230-61350230 on Build GRCh38
rs191683631 Chr.11: 61350455-61350455 on Build GRCh38
rs770139442 Chr.11: 61350404-61350404 on Build GRCh38
rs763673265 Chr.11: 61350384-61350384 on Build GRCh38
rs1038665312 Chr.11: 61350033-61350033 on Build GRCh38
rs1051278657 Chr.11: 61350058-61350058 on Build GRCh38
rs932196961 Chr.11: 61350196-61350196 on Build GRCh38
rs1031490842 Chr.11: 61350361-61350361 on Build GRCh38
rs543346644 Chr.11: 61350092-61350092 on Build GRCh38
rs970795594 Chr.11: 61350061-61350061 on Build GRCh38
rs1031208900 Chr.11: 61350098-61350099 on Build GRCh38
rs527560165 Chr.11: 61350013-61350013 on Build GRCh38
rs113572858 Chr.11: 61350204-61350204 on Build GRCh38
rs529558396 Chr.11: 61350085-61350085 on Build GRCh38
rs369390507 Chr.11: 61350419-61350419 on Build GRCh38
rs140016863 Chr.11: 61350260-61350260 on Build GRCh38
rs760284615 Chr.11: 61350375-61350375 on Build GRCh38
rs188092779 Chr.11: 61350116-61350116 on Build GRCh38
rs1023456157 Chr.11: 61350390-61350390 on Build GRCh38
rs111693851 Chr.11: 61350181-61350181 on Build GRCh38
rs79792967 Chr.11: 61350290-61350290 on Build GRCh38
rs372519055 Chr.11: 61350252-61350252 on Build GRCh38
rs1016189842 Chr.11: 61350119-61350119 on Build GRCh38
rs746387728 Chr.11: 61350284-61350284 on Build GRCh38
rs995997994 Chr.11: 61350174-61350174 on Build GRCh38
rs566500967 Chr.11: 61350387-61350387 on Build GRCh38
rs898655347 Chr.11: 61350386-61350386 on Build GRCh38
rs982155769 Chr.11: 61350070-61350070 on Build GRCh38
rs1004313945 Chr.11: 61350114-61350114 on Build GRCh38
rs769913940 Chr.11: 61350454-61350454 on Build GRCh38
rs988635232 Chr.11: 61350243-61350243 on Build GRCh38
rs201730663 Chr.11: 61350098-61350098 on Build GRCh38

View Full Product Details