Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 112275974-112276556 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TIFA
Entrez Gene ID
92610
Gene Name
TRAF interacting protein with forkhead associated domain
Gene Aliases
T2BP, T6BP, TIFAA
UniGene
Hs.310640
Gene Chromosome Location
Chr.4: 112275626-112285903 on Build GRCh38
Transcripts
NM_052864.2

SNP Detail

SNP ID Chromosome Location
rs577913197 Chr.4: 112276403-112276403 on Build GRCh38
rs144770459 Chr.4: 112276010-112276010 on Build GRCh38
rs551142923 Chr.4: 112276188-112276188 on Build GRCh38
rs372245859 Chr.4: 112276434-112276434 on Build GRCh38
rs1037040162 Chr.4: 112276389-112276389 on Build GRCh38
rs971497913 Chr.4: 112276196-112276196 on Build GRCh38
rs770916195 Chr.4: 112276471-112276471 on Build GRCh38
rs545409244 Chr.4: 112276490-112276490 on Build GRCh38
rs1008381215 Chr.4: 112276017-112276017 on Build GRCh38
rs932517104 Chr.4: 112276360-112276360 on Build GRCh38
rs550897234 Chr.4: 112276008-112276008 on Build GRCh38
rs922445773 Chr.4: 112276324-112276324 on Build GRCh38
rs1022993027 Chr.4: 112276531-112276531 on Build GRCh38
rs777699513 Chr.4: 112276325-112276325 on Build GRCh38
rs765594439 Chr.4: 112276365-112276365 on Build GRCh38
rs1046427786 Chr.4: 112276407-112276407 on Build GRCh38
rs970253368 Chr.4: 112276090-112276090 on Build GRCh38
rs1005523691 Chr.4: 112276031-112276031 on Build GRCh38
rs560917625 Chr.4: 112276159-112276160 on Build GRCh38
rs1037664756 Chr.4: 112276034-112276034 on Build GRCh38
rs34001017 Chr.4: 112276526-112276527 on Build GRCh38
rs902228692 Chr.4: 112276425-112276425 on Build GRCh38
rs1012959739 Chr.4: 112276192-112276192 on Build GRCh38
rs932563171 Chr.4: 112276364-112276364 on Build GRCh38
rs779454517 Chr.4: 112276121-112276121 on Build GRCh38
rs375497243 Chr.4: 112276238-112276238 on Build GRCh38
rs957669137 Chr.4: 112276152-112276152 on Build GRCh38
rs3172586 Chr.4: 112276226-112276226 on Build GRCh38
rs1019135246 Chr.4: 112276044-112276044 on Build GRCh38
rs190912041 Chr.4: 112276401-112276401 on Build GRCh38
rs888435337 Chr.4: 112276020-112276020 on Build GRCh38
rs1033271584 Chr.4: 112276103-112276103 on Build GRCh38
rs1022961962 Chr.4: 112276195-112276195 on Build GRCh38
rs573943573 Chr.4: 112276385-112276385 on Build GRCh38
rs186799882 Chr.4: 112276086-112276086 on Build GRCh38
rs981912182 Chr.4: 112276247-112276247 on Build GRCh38
rs746110085 Chr.4: 112276522-112276522 on Build GRCh38
rs753561774 Chr.4: 112276078-112276078 on Build GRCh38
rs781334638 Chr.4: 112276483-112276483 on Build GRCh38
rs1001715516 Chr.4: 112276480-112276480 on Build GRCh38
rs989165003 Chr.4: 112276200-112276200 on Build GRCh38
rs574279072 Chr.4: 112276424-112276424 on Build GRCh38
rs1001934691 Chr.4: 112276108-112276108 on Build GRCh38
rs1014801133 Chr.4: 112276085-112276085 on Build GRCh38
rs908600908 Chr.4: 112276256-112276256 on Build GRCh38
rs999433672 Chr.4: 112276435-112276435 on Build GRCh38
rs566251342 Chr.4: 112276234-112276234 on Build GRCh38
rs531483217 Chr.4: 112276230-112276230 on Build GRCh38
rs147871551 Chr.4: 112276400-112276400 on Build GRCh38
rs534091043 Chr.4: 112276287-112276287 on Build GRCh38
rs368543924 Chr.4: 112276375-112276375 on Build GRCh38
rs368282183 Chr.4: 112276060-112276060 on Build GRCh38
rs748637468 Chr.4: 112276208-112276208 on Build GRCh38
rs1049344609 Chr.4: 112276005-112276005 on Build GRCh38

View Full Product Details