Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 119077798-119078296 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VPS11
Entrez Gene ID
55823
Gene Name
VPS11, CORVET/HOPS core subunit
Gene Aliases
END1, HLD12, PEP5, RNF108, hVPS11
UniGene
Hs.234282
Gene Chromosome Location
Chr.11: 119067752-119081978 on Build GRCh38
Transcripts
NM_001290185.1 NM_021729.5 XM_011542898.1 XM_011542899.2

SNP Detail

SNP ID Chromosome Location
rs782181611 Chr.11: 119078020-119078020 on Build GRCh38
rs1015319447 Chr.11: 119078120-119078120 on Build GRCh38
rs782817112 Chr.11: 119077958-119077958 on Build GRCh38
rs782039718 Chr.11: 119078027-119078027 on Build GRCh38
rs567079585 Chr.11: 119078103-119078103 on Build GRCh38
rs782424603 Chr.11: 119078238-119078238 on Build GRCh38
rs574026427 Chr.11: 119077826-119077826 on Build GRCh38
rs782450511 Chr.11: 119077994-119077994 on Build GRCh38
rs782526268 Chr.11: 119078178-119078178 on Build GRCh38
rs965708934 Chr.11: 119077873-119077873 on Build GRCh38
rs375775913 Chr.11: 119078130-119078130 on Build GRCh38
rs375582928 Chr.11: 119077851-119077851 on Build GRCh38
rs782740204 Chr.11: 119078260-119078260 on Build GRCh38
rs782163667 Chr.11: 119078031-119078031 on Build GRCh38
rs543135449 Chr.11: 119077949-119077949 on Build GRCh38
rs553011376 Chr.11: 119077829-119077829 on Build GRCh38
rs782755733 Chr.11: 119078181-119078182 on Build GRCh38
rs782561371 Chr.11: 119078173-119078173 on Build GRCh38
rs782141817 Chr.11: 119078221-119078221 on Build GRCh38
rs782734510 Chr.11: 119078131-119078131 on Build GRCh38
rs76001104 Chr.11: 119077855-119077855 on Build GRCh38
rs782480928 Chr.11: 119077874-119077874 on Build GRCh38
rs368858377 Chr.11: 119077923-119077923 on Build GRCh38
rs782165270 Chr.11: 119078159-119078159 on Build GRCh38
rs782076833 Chr.11: 119077968-119077968 on Build GRCh38
rs782334357 Chr.11: 119078021-119078021 on Build GRCh38
rs371762955 Chr.11: 119078017-119078017 on Build GRCh38
rs781905193 Chr.11: 119078002-119078002 on Build GRCh38
rs372443553 Chr.11: 119078101-119078101 on Build GRCh38
rs781893691 Chr.11: 119078138-119078138 on Build GRCh38
rs782498761 Chr.11: 119078014-119078014 on Build GRCh38
rs782689264 Chr.11: 119078185-119078185 on Build GRCh38
rs782311347 Chr.11: 119078220-119078220 on Build GRCh38
rs782789112 Chr.11: 119078274-119078274 on Build GRCh38
rs375143893 Chr.11: 119078035-119078035 on Build GRCh38
rs782662938 Chr.11: 119078010-119078010 on Build GRCh38
rs976705036 Chr.11: 119077882-119077882 on Build GRCh38
rs782236473 Chr.11: 119078190-119078190 on Build GRCh38
rs782510678 Chr.11: 119078043-119078043 on Build GRCh38
rs782759975 Chr.11: 119078164-119078164 on Build GRCh38
rs781784085 Chr.11: 119077974-119077974 on Build GRCh38
rs782729648 Chr.11: 119077973-119077973 on Build GRCh38
rs200774813 Chr.11: 119077945-119077945 on Build GRCh38
rs782416448 Chr.11: 119078102-119078102 on Build GRCh38
rs565454827 Chr.11: 119078045-119078045 on Build GRCh38
rs1046660854 Chr.11: 119078059-119078059 on Build GRCh38
rs892472430 Chr.11: 119078024-119078024 on Build GRCh38
rs561501534 Chr.11: 119077996-119077996 on Build GRCh38
rs528675586 Chr.11: 119078013-119078013 on Build GRCh38
rs146122577 Chr.11: 119078214-119078214 on Build GRCh38
rs782207236 Chr.11: 119078016-119078016 on Build GRCh38
rs186792200 Chr.11: 119078195-119078195 on Build GRCh38
rs782137506 Chr.11: 119078108-119078108 on Build GRCh38
rs781965646 Chr.11: 119078124-119078124 on Build GRCh38
rs782532693 Chr.11: 119077905-119077905 on Build GRCh38
rs781818338 Chr.11: 119078165-119078165 on Build GRCh38
rs782570405 Chr.11: 119078089-119078089 on Build GRCh38
rs781935851 Chr.11: 119078229-119078229 on Build GRCh38
rs782324655 Chr.11: 119078030-119078030 on Build GRCh38
rs782133881 Chr.11: 119078250-119078250 on Build GRCh38
rs782391539 Chr.11: 119078213-119078213 on Build GRCh38
rs1002404507 Chr.11: 119078096-119078096 on Build GRCh38
rs781960113 Chr.11: 119078116-119078116 on Build GRCh38
rs782088087 Chr.11: 119078038-119078038 on Build GRCh38
rs905412822 Chr.11: 119078060-119078060 on Build GRCh38
rs782484147 Chr.11: 119078169-119078169 on Build GRCh38
rs782454809 Chr.11: 119078040-119078040 on Build GRCh38
rs781891580 Chr.11: 119077848-119077848 on Build GRCh38
rs374444470 Chr.11: 119077909-119077909 on Build GRCh38
rs781814943 Chr.11: 119077857-119077857 on Build GRCh38
rs781878357 Chr.11: 119078037-119078037 on Build GRCh38
rs527724546 Chr.11: 119078019-119078019 on Build GRCh38
rs373781814 Chr.11: 119077944-119077944 on Build GRCh38
rs781947288 Chr.11: 119077841-119077843 on Build GRCh38
rs371324199 Chr.11: 119077924-119077924 on Build GRCh38
rs370760856 Chr.11: 119077844-119077844 on Build GRCh38
rs962015767 Chr.11: 119078100-119078100 on Build GRCh38
rs781955261 Chr.11: 119077934-119077934 on Build GRCh38
rs781917185 Chr.11: 119078264-119078264 on Build GRCh38
rs781993267 Chr.11: 119077962-119077962 on Build GRCh38
rs369462724 Chr.11: 119078039-119078039 on Build GRCh38
rs1028255023 Chr.11: 119078170-119078170 on Build GRCh38
rs782506288 Chr.11: 119078007-119078007 on Build GRCh38
rs781803339 Chr.11: 119078174-119078174 on Build GRCh38
rs782227762 Chr.11: 119078076-119078076 on Build GRCh38
rs768186101 Chr.11: 119077902-119077902 on Build GRCh38
rs79450906 Chr.11: 119077897-119077897 on Build GRCh38
rs375492582 Chr.11: 119078042-119078042 on Build GRCh38
rs370133447 Chr.11: 119078241-119078241 on Build GRCh38
rs782061826 Chr.11: 119078271-119078271 on Build GRCh38
rs782299183 Chr.11: 119078077-119078077 on Build GRCh38
rs547189232 Chr.11: 119078015-119078015 on Build GRCh38
rs782099869 Chr.11: 119078167-119078168 on Build GRCh38
rs782280365 Chr.11: 119078233-119078233 on Build GRCh38

View Full Product Details