Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40812620-40813113 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZFYVE19
Entrez Gene ID
84936
Gene Name
zinc finger FYVE-type containing 19
Gene Aliases
ANCHR, MPFYVE
UniGene
Hs.121676
Gene Chromosome Location
Chr.15: 40807076-40814569 on Build GRCh38
Transcripts
NM_001077268.1 NM_001258420.1 NM_001258421.1 NM_032850.4 XM_017022683.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
961306 c.949C>T p.R317W Chr.15: 40812821-40812821 on Build GRCh38
4054423 c.1021C>T p.P341S Chr.15: 40812893-40812893 on Build GRCh38
7297462 c.925C>T p.L309L Chr.15: 40812797-40812797 on Build GRCh38
4742733 c.993C>T p.P331P Chr.15: 40812895-40812895 on Build GRCh38
7075633 c.933G>A p.T311T Chr.15: 40812835-40812835 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs17657877 Chr.15: 40812676-40812676 on Build GRCh38
rs772548682 Chr.15: 40812788-40812788 on Build GRCh38
rs544912505 Chr.15: 40812877-40812877 on Build GRCh38
rs774578518 Chr.15: 40812686-40812686 on Build GRCh38
rs761243727 Chr.15: 40812705-40812705 on Build GRCh38
rs371725176 Chr.15: 40812742-40812742 on Build GRCh38
rs746155591 Chr.15: 40812775-40812777 on Build GRCh38
rs762364507 Chr.15: 40812951-40812951 on Build GRCh38
rs765960750 Chr.15: 40812787-40812787 on Build GRCh38
rs749054111 Chr.15: 40812822-40812822 on Build GRCh38
rs138800039 Chr.15: 40812696-40812696 on Build GRCh38
rs564495301 Chr.15: 40812883-40812883 on Build GRCh38
rs7178363 Chr.15: 40812799-40812799 on Build GRCh38
rs554882498 Chr.15: 40812823-40812825 on Build GRCh38
rs746117412 Chr.15: 40812916-40812916 on Build GRCh38
rs367913127 Chr.15: 40812689-40812689 on Build GRCh38
rs952214818 Chr.15: 40813047-40813047 on Build GRCh38
rs547143824 Chr.15: 40812987-40812987 on Build GRCh38
rs751225374 Chr.15: 40812881-40812881 on Build GRCh38
rs902831263 Chr.15: 40812963-40812963 on Build GRCh38
rs770696088 Chr.15: 40812826-40812826 on Build GRCh38
rs192472112 Chr.15: 40812670-40812670 on Build GRCh38
rs1015036474 Chr.15: 40813083-40813083 on Build GRCh38
rs754450033 Chr.15: 40812664-40812664 on Build GRCh38
rs748129559 Chr.15: 40812818-40812818 on Build GRCh38
rs1011656204 Chr.15: 40812761-40812761 on Build GRCh38
rs772453172 Chr.15: 40812919-40812919 on Build GRCh38
rs543909533 Chr.15: 40812899-40812899 on Build GRCh38
rs76824203 Chr.15: 40812748-40812748 on Build GRCh38
rs368873146 Chr.15: 40812821-40812821 on Build GRCh38
rs909027779 Chr.15: 40812944-40812944 on Build GRCh38
rs604250 Chr.15: 40812990-40812990 on Build GRCh38
rs757955058 Chr.15: 40812655-40812655 on Build GRCh38
rs747098424 Chr.15: 40812926-40812926 on Build GRCh38
rs1035202139 Chr.15: 40812969-40812969 on Build GRCh38
rs780664881 Chr.15: 40812815-40812815 on Build GRCh38
rs375252804 Chr.15: 40812912-40812912 on Build GRCh38
rs1012829918 Chr.15: 40812988-40812988 on Build GRCh38
rs764591636 Chr.15: 40812718-40812718 on Build GRCh38
rs747492657 Chr.15: 40812896-40812897 on Build GRCh38
rs985178421 Chr.15: 40812873-40812873 on Build GRCh38
rs376725467 Chr.15: 40812866-40812866 on Build GRCh38
rs978068337 Chr.15: 40812822-40812823 on Build GRCh38
rs984865401 Chr.15: 40813054-40813054 on Build GRCh38
rs896778787 Chr.15: 40812971-40812971 on Build GRCh38
rs371144836 Chr.15: 40812672-40812672 on Build GRCh38
rs552917191 Chr.15: 40812759-40812759 on Build GRCh38
rs574892620 Chr.15: 40812885-40812885 on Build GRCh38
rs772329476 Chr.15: 40812840-40812840 on Build GRCh38
rs750150244 Chr.15: 40812654-40812654 on Build GRCh38
rs999701677 Chr.15: 40812803-40812803 on Build GRCh38
rs780372966 Chr.15: 40812923-40812923 on Build GRCh38
rs375180271 Chr.15: 40812652-40812652 on Build GRCh38
rs777068505 Chr.15: 40812939-40812939 on Build GRCh38
rs775681773 Chr.15: 40812845-40812845 on Build GRCh38
rs777695254 Chr.15: 40812893-40812893 on Build GRCh38
rs186609919 Chr.15: 40812986-40812986 on Build GRCh38
rs923191414 Chr.15: 40812895-40812895 on Build GRCh38
rs781108451 Chr.15: 40812669-40812669 on Build GRCh38
rs748876769 Chr.15: 40813001-40813001 on Build GRCh38
rs779714971 Chr.15: 40812656-40812656 on Build GRCh38
rs952202122 Chr.15: 40812867-40812867 on Build GRCh38
rs957110395 Chr.15: 40812801-40812801 on Build GRCh38
rs779072132 Chr.15: 40812832-40812832 on Build GRCh38
rs779004944 Chr.15: 40812650-40812653 on Build GRCh38
rs768788681 Chr.15: 40812937-40812937 on Build GRCh38
rs776199269 Chr.15: 40812703-40812703 on Build GRCh38
rs746090607 Chr.15: 40812834-40812834 on Build GRCh38
rs746523328 Chr.15: 40812661-40812661 on Build GRCh38
rs965399102 Chr.15: 40812841-40812841 on Build GRCh38
rs975956331 Chr.15: 40812895-40812896 on Build GRCh38
rs767120311 Chr.15: 40812884-40812884 on Build GRCh38
rs896429999 Chr.15: 40813005-40813005 on Build GRCh38
rs754509756 Chr.15: 40812800-40812800 on Build GRCh38
rs555039521 Chr.15: 40812690-40812690 on Build GRCh38
rs940686622 Chr.15: 40812890-40812890 on Build GRCh38
rs369723809 Chr.15: 40812882-40812882 on Build GRCh38
rs769078358 Chr.15: 40812945-40812946 on Build GRCh38
rs756079180 Chr.15: 40812820-40812820 on Build GRCh38
rs753811268 Chr.15: 40812896-40812896 on Build GRCh38
rs183941751 Chr.15: 40812846-40812846 on Build GRCh38
rs777137236 Chr.15: 40812857-40812857 on Build GRCh38
rs769306168 Chr.15: 40812851-40812851 on Build GRCh38
rs988392614 Chr.15: 40812810-40812810 on Build GRCh38
rs1002498243 Chr.15: 40812967-40812967 on Build GRCh38
rs1035251602 Chr.15: 40812816-40812816 on Build GRCh38

View Full Product Details