Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 23817394-23817889 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HMGCL
Entrez Gene ID
3155
Gene Name
3-hydroxymethyl-3-methylglutaryl-CoA lyase
Gene Aliases
HL
UniGene
Hs.533444
Gene Chromosome Location
Chr.1: 23801877-23825459 on Build GRCh38
Transcripts
NM_000191.2 NM_001166059.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1340684 c.204C>A p.L68L Chr.1: 23817524-23817524 on Build GRCh38
4577063 c.233C>T p.S78F Chr.1: 23817495-23817495 on Build GRCh38
4799565 c.223A>G p.S75G Chr.1: 23817505-23817505 on Build GRCh38
7189363 c.145-4A>G p.? Chr.1: 23817587-23817587 on Build GRCh38
3486112 c.150C>T p.I50I Chr.1: 23817578-23817578 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376733438 Chr.1: 23817441-23817441 on Build GRCh38
rs555961548 Chr.1: 23817664-23817664 on Build GRCh38
rs542698771 Chr.1: 23817839-23817839 on Build GRCh38
rs767060526 Chr.1: 23817473-23817473 on Build GRCh38
rs758546371 Chr.1: 23817425-23817425 on Build GRCh38
rs765840549 Chr.1: 23817504-23817504 on Build GRCh38
rs745310996 Chr.1: 23817559-23817559 on Build GRCh38
rs771039622 Chr.1: 23817458-23817458 on Build GRCh38
rs759149770 Chr.1: 23817468-23817468 on Build GRCh38
rs951618867 Chr.1: 23817648-23817648 on Build GRCh38
rs762224192 Chr.1: 23817596-23817596 on Build GRCh38
rs560778852 Chr.1: 23817860-23817860 on Build GRCh38
rs923646391 Chr.1: 23817643-23817643 on Build GRCh38
rs538620811 Chr.1: 23817484-23817484 on Build GRCh38
rs186406939 Chr.1: 23817637-23817637 on Build GRCh38
rs963436984 Chr.1: 23817587-23817587 on Build GRCh38
rs765606573 Chr.1: 23817603-23817603 on Build GRCh38
rs773700977 Chr.1: 23817459-23817459 on Build GRCh38
rs753669003 Chr.1: 23817633-23817633 on Build GRCh38
rs376705029 Chr.1: 23817578-23817578 on Build GRCh38
rs181179069 Chr.1: 23817627-23817627 on Build GRCh38
rs373738626 Chr.1: 23817544-23817544 on Build GRCh38
rs1012852785 Chr.1: 23817728-23817728 on Build GRCh38
rs941376913 Chr.1: 23817457-23817457 on Build GRCh38
rs143981931 Chr.1: 23817546-23817546 on Build GRCh38
rs369286537 Chr.1: 23817591-23817591 on Build GRCh38
rs142942001 Chr.1: 23817577-23817577 on Build GRCh38
rs750944360 Chr.1: 23817609-23817609 on Build GRCh38
rs766502036 Chr.1: 23817621-23817621 on Build GRCh38
rs140840889 Chr.1: 23817629-23817630 on Build GRCh38
rs777837362 Chr.1: 23817754-23817754 on Build GRCh38
rs181467111 Chr.1: 23817721-23817721 on Build GRCh38
rs773244041 Chr.1: 23817448-23817448 on Build GRCh38
rs933966061 Chr.1: 23817743-23817743 on Build GRCh38
rs528406367 Chr.1: 23817435-23817435 on Build GRCh38
rs962917962 Chr.1: 23817685-23817685 on Build GRCh38
rs774401260 Chr.1: 23817549-23817549 on Build GRCh38
rs150122271 Chr.1: 23817747-23817747 on Build GRCh38
rs563631583 Chr.1: 23817817-23817817 on Build GRCh38
rs574268575 Chr.1: 23817710-23817710 on Build GRCh38
rs764468396 Chr.1: 23817492-23817492 on Build GRCh38
rs945207802 Chr.1: 23817575-23817575 on Build GRCh38
rs1029059514 Chr.1: 23817868-23817868 on Build GRCh38
rs754253328 Chr.1: 23817495-23817495 on Build GRCh38
rs575663026 Chr.1: 23817831-23817831 on Build GRCh38
rs121964996 Chr.1: 23817520-23817520 on Build GRCh38
rs190245759 Chr.1: 23817714-23817714 on Build GRCh38
rs780073361 Chr.1: 23817526-23817526 on Build GRCh38
rs755218532 Chr.1: 23817629-23817629 on Build GRCh38
rs781627075 Chr.1: 23817632-23817632 on Build GRCh38
rs895302726 Chr.1: 23817722-23817722 on Build GRCh38
rs774961101 Chr.1: 23817570-23817570 on Build GRCh38
rs1053567971 Chr.1: 23817688-23817688 on Build GRCh38
rs767464363 Chr.1: 23817454-23817454 on Build GRCh38
rs770984830 Chr.1: 23817547-23817547 on Build GRCh38
rs2076344 Chr.1: 23817442-23817442 on Build GRCh38
rs576683525 Chr.1: 23817825-23817826 on Build GRCh38
rs780681704 Chr.1: 23817444-23817448 on Build GRCh38
rs758518472 Chr.1: 23817525-23817525 on Build GRCh38
rs752137615 Chr.1: 23817521-23817522 on Build GRCh38
rs771611643 Chr.1: 23817567-23817567 on Build GRCh38
rs1043499387 Chr.1: 23817789-23817789 on Build GRCh38
rs750457695 Chr.1: 23817517-23817517 on Build GRCh38
rs138581313 Chr.1: 23817753-23817753 on Build GRCh38
rs1014878377 Chr.1: 23817814-23817814 on Build GRCh38
rs755922538 Chr.1: 23817545-23817545 on Build GRCh38
rs994870635 Chr.1: 23817844-23817844 on Build GRCh38
rs985379055 Chr.1: 23817614-23817614 on Build GRCh38
rs550414495 Chr.1: 23817522-23817522 on Build GRCh38
rs775218067 Chr.1: 23817475-23817475 on Build GRCh38
rs936403779 Chr.1: 23817667-23817667 on Build GRCh38
rs934067950 Chr.1: 23817523-23817523 on Build GRCh38
rs912489979 Chr.1: 23817615-23817615 on Build GRCh38
rs201736880 Chr.1: 23817443-23817443 on Build GRCh38

View Full Product Details