Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 69084913-69085458 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA6
Entrez Gene ID
23460
Gene Name
ATP binding cassette subfamily A member 6
Gene Aliases
EST155051
UniGene
Hs.709514
Gene Chromosome Location
Chr.17: 69062044-69142188 on Build GRCh38
Transcripts
NM_080284.2 XM_011524574.2 XM_017024404.1 XM_017024405.1 XM_017024406.1 XM_017024407.1 XM_017024408.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7353500 c.4169G>T p.R1390M Chr.17: 69085043-69085043 on Build GRCh38
707473 c.4030-1G>C p.? Chr.17: 69085183-69085183 on Build GRCh38
1710690 c.4115G>A p.R1372K Chr.17: 69085097-69085097 on Build GRCh38
707474 c.4162G>C p.D1388H Chr.17: 69085050-69085050 on Build GRCh38
2975429 c.4132T>C p.Y1378H Chr.17: 69085080-69085080 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs949824709 Chr.17: 69084939-69084939 on Build GRCh38
rs369645216 Chr.17: 69085103-69085103 on Build GRCh38
rs760791622 Chr.17: 69085051-69085051 on Build GRCh38
rs573638884 Chr.17: 69085386-69085386 on Build GRCh38
rs1038354557 Chr.17: 69085388-69085388 on Build GRCh38
rs182377176 Chr.17: 69085134-69085134 on Build GRCh38
rs769613655 Chr.17: 69085391-69085391 on Build GRCh38
rs531863229 Chr.17: 69085170-69085170 on Build GRCh38
rs981580543 Chr.17: 69084953-69084953 on Build GRCh38
rs950752906 Chr.17: 69085174-69085174 on Build GRCh38
rs748770804 Chr.17: 69085199-69085199 on Build GRCh38
rs1051226880 Chr.17: 69085301-69085301 on Build GRCh38
rs149025377 Chr.17: 69085077-69085077 on Build GRCh38
rs958445145 Chr.17: 69085231-69085231 on Build GRCh38
rs1006600279 Chr.17: 69085410-69085410 on Build GRCh38
rs537984839 Chr.17: 69085416-69085416 on Build GRCh38
rs750663486 Chr.17: 69085193-69085194 on Build GRCh38
rs759910624 Chr.17: 69085124-69085124 on Build GRCh38
rs201746594 Chr.17: 69085150-69085150 on Build GRCh38
rs754263525 Chr.17: 69085022-69085023 on Build GRCh38
rs746661822 Chr.17: 69085001-69085001 on Build GRCh38
rs755716336 Chr.17: 69085188-69085188 on Build GRCh38
rs368756234 Chr.17: 69085117-69085117 on Build GRCh38
rs889878411 Chr.17: 69085338-69085338 on Build GRCh38
rs752314911 Chr.17: 69085186-69085186 on Build GRCh38
rs747592034 Chr.17: 69085015-69085015 on Build GRCh38
rs893060567 Chr.17: 69085019-69085019 on Build GRCh38
rs537077875 Chr.17: 69085017-69085017 on Build GRCh38
rs533510757 Chr.17: 69085299-69085299 on Build GRCh38
rs578247411 Chr.17: 69085107-69085107 on Build GRCh38
rs62082719 Chr.17: 69085020-69085020 on Build GRCh38
rs947010484 Chr.17: 69085003-69085003 on Build GRCh38
rs763423046 Chr.17: 69085021-69085021 on Build GRCh38
rs994407542 Chr.17: 69085429-69085429 on Build GRCh38
rs371622240 Chr.17: 69084998-69084998 on Build GRCh38
rs777131702 Chr.17: 69085191-69085191 on Build GRCh38
rs374878517 Chr.17: 69084986-69084986 on Build GRCh38
rs140310197 Chr.17: 69085169-69085169 on Build GRCh38
rs750257002 Chr.17: 69084990-69084990 on Build GRCh38
rs902155689 Chr.17: 69085025-69085025 on Build GRCh38
rs771167668 Chr.17: 69085038-69085038 on Build GRCh38
rs767787424 Chr.17: 69085046-69085046 on Build GRCh38
rs752826109 Chr.17: 69085048-69085048 on Build GRCh38
rs148658263 Chr.17: 69085012-69085012 on Build GRCh38
rs990946108 Chr.17: 69085227-69085228 on Build GRCh38
rs140239456 Chr.17: 69085265-69085265 on Build GRCh38
rs758110488 Chr.17: 69084994-69084994 on Build GRCh38
rs143317768 Chr.17: 69085039-69085039 on Build GRCh38
rs1018158239 Chr.17: 69085087-69085087 on Build GRCh38
rs751297791 Chr.17: 69085071-69085071 on Build GRCh38
rs780781581 Chr.17: 69085078-69085078 on Build GRCh38
rs914211219 Chr.17: 69085263-69085263 on Build GRCh38
rs559040166 Chr.17: 69085426-69085426 on Build GRCh38
rs544457483 Chr.17: 69085062-69085062 on Build GRCh38
rs746286979 Chr.17: 69085233-69085233 on Build GRCh38
rs757469024 Chr.17: 69085168-69085168 on Build GRCh38
rs759815031 Chr.17: 69085045-69085045 on Build GRCh38
rs142075639 Chr.17: 69085098-69085098 on Build GRCh38
rs927007683 Chr.17: 69084968-69084968 on Build GRCh38
rs764080725 Chr.17: 69085138-69085138 on Build GRCh38
rs1050814609 Chr.17: 69085293-69085293 on Build GRCh38
rs560766436 Chr.17: 69084951-69084951 on Build GRCh38
rs551215842 Chr.17: 69085281-69085281 on Build GRCh38
rs960857291 Chr.17: 69085226-69085226 on Build GRCh38
rs749750215 Chr.17: 69085102-69085102 on Build GRCh38
rs924237397 Chr.17: 69085284-69085284 on Build GRCh38
rs369860134 Chr.17: 69085203-69085203 on Build GRCh38
rs2302132 Chr.17: 69085372-69085372 on Build GRCh38
rs773259704 Chr.17: 69085141-69085141 on Build GRCh38
rs747677895 Chr.17: 69085083-69085083 on Build GRCh38
rs780695660 Chr.17: 69085014-69085014 on Build GRCh38
rs372819223 Chr.17: 69085217-69085217 on Build GRCh38
rs994358188 Chr.17: 69085163-69085163 on Build GRCh38
rs971525783 Chr.17: 69085173-69085173 on Build GRCh38
rs774589393 Chr.17: 69085110-69085110 on Build GRCh38
rs527461596 Chr.17: 69085052-69085052 on Build GRCh38
rs566439343 Chr.17: 69085287-69085287 on Build GRCh38
rs35750521 Chr.17: 69084940-69084941 on Build GRCh38
rs765379105 Chr.17: 69085185-69085185 on Build GRCh38
rs779701262 Chr.17: 69085063-69085063 on Build GRCh38
rs143953811 Chr.17: 69085269-69085269 on Build GRCh38
rs762811326 Chr.17: 69085142-69085142 on Build GRCh38
rs758461168 Chr.17: 69085196-69085197 on Build GRCh38
rs188158113 Chr.17: 69085133-69085133 on Build GRCh38
rs778383822 Chr.17: 69085099-69085099 on Build GRCh38
rs62082718 Chr.17: 69084989-69084989 on Build GRCh38
rs77542162 Chr.17: 69085137-69085137 on Build GRCh38
rs990229500 Chr.17: 69085240-69085240 on Build GRCh38

View Full Product Details