Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 8371102-8371595 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANGPTL4
Entrez Gene ID
51129
Gene Name
angiopoietin like 4
Gene Aliases
ARP4, FIAF, HARP, HFARP, NL2, PGAR, TGQTL, UNQ171, pp1158
UniGene
Hs.9613
Gene Chromosome Location
Chr.19: 8364127-8374375 on Build GRCh38
Transcripts
NM_001039667.2 NM_139314.2 NR_104213.1 XM_005272484.3 XM_005272485.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1305325 c.957C>G p.P319P Chr.19: 8371440-8371440 on Build GRCh38
3836195 c.762G>C p.E254D Chr.19: 8371245-8371245 on Build GRCh38
139115 c.791G>A p.S264N Chr.19: 8371274-8371274 on Build GRCh38
1182734 c.1007G>A p.R336H Chr.19: 8371490-8371490 on Build GRCh38
368299 c.730T>C p.Y244H Chr.19: 8371124-8371124 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766875709 Chr.19: 8371210-8371210 on Build GRCh38
rs138211096 Chr.19: 8371305-8371305 on Build GRCh38
rs769961302 Chr.19: 8371184-8371184 on Build GRCh38
rs531933760 Chr.19: 8371371-8371371 on Build GRCh38
rs555274215 Chr.19: 8371507-8371507 on Build GRCh38
rs752633223 Chr.19: 8371350-8371352 on Build GRCh38
rs139295817 Chr.19: 8371332-8371332 on Build GRCh38
rs761257386 Chr.19: 8371364-8371364 on Build GRCh38
rs761950596 Chr.19: 8371122-8371122 on Build GRCh38
rs10404615 Chr.19: 8371404-8371404 on Build GRCh38
rs779095513 Chr.19: 8371421-8371421 on Build GRCh38
rs756854279 Chr.19: 8371258-8371258 on Build GRCh38
rs753558114 Chr.19: 8371256-8371256 on Build GRCh38
rs35061979 Chr.19: 8371316-8371316 on Build GRCh38
rs202043145 Chr.19: 8371315-8371315 on Build GRCh38
rs766213750 Chr.19: 8371400-8371400 on Build GRCh38
rs758390511 Chr.19: 8371384-8371390 on Build GRCh38
rs368548994 Chr.19: 8371150-8371150 on Build GRCh38
rs777913903 Chr.19: 8371338-8371338 on Build GRCh38
rs199564844 Chr.19: 8371151-8371151 on Build GRCh38
rs369766671 Chr.19: 8371385-8371385 on Build GRCh38
rs561517050 Chr.19: 8371482-8371482 on Build GRCh38
rs771338375 Chr.19: 8371347-8371347 on Build GRCh38
rs146875081 Chr.19: 8371474-8371474 on Build GRCh38
rs774924877 Chr.19: 8371298-8371298 on Build GRCh38
rs767605224 Chr.19: 8371479-8371479 on Build GRCh38
rs200311316 Chr.19: 8371131-8371131 on Build GRCh38
rs754022089 Chr.19: 8371410-8371410 on Build GRCh38
rs747326940 Chr.19: 8371432-8371432 on Build GRCh38
rs919979743 Chr.19: 8371351-8371351 on Build GRCh38
rs866346299 Chr.19: 8371540-8371540 on Build GRCh38
rs1003467490 Chr.19: 8371279-8371279 on Build GRCh38
rs955553633 Chr.19: 8371458-8371458 on Build GRCh38
rs754818405 Chr.19: 8371335-8371335 on Build GRCh38
rs748220039 Chr.19: 8371281-8371281 on Build GRCh38
rs763692209 Chr.19: 8371243-8371243 on Build GRCh38
rs762467580 Chr.19: 8371388-8371388 on Build GRCh38
rs762270935 Chr.19: 8371447-8371447 on Build GRCh38
rs752209196 Chr.19: 8371237-8371237 on Build GRCh38
rs113605673 Chr.19: 8371230-8371230 on Build GRCh38
rs750402576 Chr.19: 8371490-8371490 on Build GRCh38
rs758394227 Chr.19: 8371274-8371274 on Build GRCh38
rs539779534 Chr.19: 8371473-8371473 on Build GRCh38
rs758160705 Chr.19: 8371322-8371322 on Build GRCh38
rs762675357 Chr.19: 8371466-8371466 on Build GRCh38
rs1012348472 Chr.19: 8371285-8371285 on Build GRCh38
rs982859311 Chr.19: 8371562-8371562 on Build GRCh38
rs748671142 Chr.19: 8371531-8371531 on Build GRCh38
rs532896676 Chr.19: 8371318-8371318 on Build GRCh38
rs770680793 Chr.19: 8371168-8371168 on Build GRCh38
rs761945791 Chr.19: 8371181-8371181 on Build GRCh38
rs750108927 Chr.19: 8371377-8371377 on Build GRCh38
rs370389963 Chr.19: 8371525-8371525 on Build GRCh38
rs376708909 Chr.19: 8371267-8371267 on Build GRCh38
rs143620129 Chr.19: 8371311-8371311 on Build GRCh38
rs765430634 Chr.19: 8371123-8371123 on Build GRCh38
rs200918932 Chr.19: 8371453-8371453 on Build GRCh38
rs569902721 Chr.19: 8371477-8371477 on Build GRCh38
rs974089872 Chr.19: 8371340-8371340 on Build GRCh38
rs745824395 Chr.19: 8371173-8371173 on Build GRCh38
rs772780624 Chr.19: 8371542-8371542 on Build GRCh38
rs760125334 Chr.19: 8371227-8371227 on Build GRCh38
rs138696635 Chr.19: 8371242-8371242 on Build GRCh38
rs913940639 Chr.19: 8371452-8371452 on Build GRCh38
rs543375556 Chr.19: 8371472-8371472 on Build GRCh38
rs375651388 Chr.19: 8371180-8371180 on Build GRCh38
rs761295713 Chr.19: 8371372-8371372 on Build GRCh38
rs1018504725 Chr.19: 8371478-8371478 on Build GRCh38
rs1044250 Chr.19: 8371280-8371280 on Build GRCh38
rs763231211 Chr.19: 8371208-8371208 on Build GRCh38
rs1011837076 Chr.19: 8371416-8371416 on Build GRCh38
rs886273998 Chr.19: 8371392-8371392 on Build GRCh38
rs764915999 Chr.19: 8371376-8371376 on Build GRCh38
rs749538025 Chr.19: 8371289-8371289 on Build GRCh38
rs774693998 Chr.19: 8371353-8371353 on Build GRCh38
rs1047608952 Chr.19: 8371199-8371199 on Build GRCh38
rs773121891 Chr.19: 8371288-8371288 on Build GRCh38
rs745591536 Chr.19: 8371271-8371271 on Build GRCh38
rs747940485 Chr.19: 8371419-8371419 on Build GRCh38
rs771211700 Chr.19: 8371297-8371297 on Build GRCh38
rs143596863 Chr.19: 8371444-8371444 on Build GRCh38
rs764437255 Chr.19: 8371368-8371368 on Build GRCh38
rs576267013 Chr.19: 8371152-8371152 on Build GRCh38
rs773454416 Chr.19: 8371191-8371191 on Build GRCh38
rs370978676 Chr.19: 8371135-8371135 on Build GRCh38
rs780121474 Chr.19: 8371497-8371497 on Build GRCh38
rs376328756 Chr.19: 8371145-8371145 on Build GRCh38
rs139998264 Chr.19: 8371405-8371405 on Build GRCh38
rs778638369 Chr.19: 8371171-8371171 on Build GRCh38
rs773846071 Chr.19: 8371569-8371569 on Build GRCh38
rs772128054 Chr.19: 8371425-8371425 on Build GRCh38
rs746114692 Chr.19: 8371422-8371422 on Build GRCh38
rs749083717 Chr.19: 8371166-8371166 on Build GRCh38
rs369127747 Chr.19: 8371306-8371306 on Build GRCh38
rs756065414 Chr.19: 8371483-8371483 on Build GRCh38
rs776032253 Chr.19: 8371300-8371300 on Build GRCh38
rs768868188 Chr.19: 8371516-8371516 on Build GRCh38
rs930002951 Chr.19: 8371556-8371556 on Build GRCh38
rs768135470 Chr.19: 8371231-8371231 on Build GRCh38
rs770222749 Chr.19: 8371539-8371539 on Build GRCh38
rs751249880 Chr.19: 8371402-8371402 on Build GRCh38
rs117456113 Chr.19: 8371554-8371554 on Build GRCh38
rs749374489 Chr.19: 8371346-8371346 on Build GRCh38
rs763357381 Chr.19: 8371132-8371132 on Build GRCh38
rs570859875 Chr.19: 8371461-8371461 on Build GRCh38
rs373081558 Chr.19: 8371138-8371138 on Build GRCh38
rs762357241 Chr.19: 8371547-8371547 on Build GRCh38
rs751916372 Chr.19: 8371218-8371218 on Build GRCh38
rs746993217 Chr.19: 8371333-8371333 on Build GRCh38
rs769261817 Chr.19: 8371443-8371443 on Build GRCh38
rs140744493 Chr.19: 8371489-8371489 on Build GRCh38
rs775667046 Chr.19: 8371360-8371360 on Build GRCh38
rs375060464 Chr.19: 8371375-8371375 on Build GRCh38
rs751688415 Chr.19: 8371558-8371558 on Build GRCh38
rs370364569 Chr.19: 8371149-8371149 on Build GRCh38
rs150000287 Chr.19: 8371354-8371354 on Build GRCh38
rs771935375 Chr.19: 8371178-8371178 on Build GRCh38
rs757379628 Chr.19: 8371411-8371411 on Build GRCh38
rs201723781 Chr.19: 8371427-8371427 on Build GRCh38

View Full Product Details