Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 171391465-171391963 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NPM1
Entrez Gene ID
4869
Gene Name
nucleophosmin
Gene Aliases
B23, NPM
UniGene
Hs.557550
Gene Chromosome Location
Chr.5: 171387648-171410900 on Build GRCh38
Transcripts
NM_001037738.2 NM_002520.6 NM_199185.3 XM_005265920.3 XM_011534564.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3614158 c.317C>T p.S106L Chr.5: 171391764-171391764 on Build GRCh38
6327816 c.301A>C p.R101R Chr.5: 171391748-171391748 on Build GRCh38
482516 c.317C>A p.S106* Chr.5: 171391764-171391764 on Build GRCh38
6327815 c.301A>C p.R101R Chr.5: 171391748-171391748 on Build GRCh38
6330994 c.306G>T p.L102F Chr.5: 171391753-171391753 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758764070 Chr.5: 171391827-171391827 on Build GRCh38
rs886112992 Chr.5: 171391622-171391622 on Build GRCh38
rs770257005 Chr.5: 171391657-171391659 on Build GRCh38
rs563104384 Chr.5: 171391548-171391548 on Build GRCh38
rs3830032 Chr.5: 171391893-171391893 on Build GRCh38
rs965606358 Chr.5: 171391679-171391679 on Build GRCh38
rs752933446 Chr.5: 171391717-171391717 on Build GRCh38
rs945361817 Chr.5: 171391539-171391539 on Build GRCh38
rs756323205 Chr.5: 171391739-171391739 on Build GRCh38
rs752629938 Chr.5: 171391672-171391672 on Build GRCh38
rs1042289188 Chr.5: 171391546-171391546 on Build GRCh38
rs746435229 Chr.5: 171391820-171391820 on Build GRCh38
rs559635693 Chr.5: 171391831-171391831 on Build GRCh38
rs746630707 Chr.5: 171391836-171391836 on Build GRCh38
rs768212080 Chr.5: 171391839-171391839 on Build GRCh38
rs141086340 Chr.5: 171391798-171391798 on Build GRCh38
rs994992891 Chr.5: 171391608-171391608 on Build GRCh38
rs894917989 Chr.5: 171391599-171391599 on Build GRCh38
rs557170584 Chr.5: 171391916-171391916 on Build GRCh38
rs922388986 Chr.5: 171391591-171391591 on Build GRCh38
rs191051017 Chr.5: 171391931-171391931 on Build GRCh38
rs776221894 Chr.5: 171391840-171391840 on Build GRCh38
rs374396635 Chr.5: 171391842-171391842 on Build GRCh38
rs908452741 Chr.5: 171391540-171391540 on Build GRCh38
rs889230726 Chr.5: 171391629-171391629 on Build GRCh38
rs765276443 Chr.5: 171391674-171391674 on Build GRCh38
rs770603961 Chr.5: 171391915-171391915 on Build GRCh38
rs769106810 Chr.5: 171391646-171391646 on Build GRCh38
rs978036156 Chr.5: 171391813-171391813 on Build GRCh38
rs568386550 Chr.5: 171391896-171391896 on Build GRCh38
rs939202809 Chr.5: 171391553-171391553 on Build GRCh38
rs1040988934 Chr.5: 171391549-171391549 on Build GRCh38
rs753931371 Chr.5: 171391777-171391777 on Build GRCh38
rs3830033 Chr.5: 171391902-171391902 on Build GRCh38
rs200782361 Chr.5: 171391699-171391699 on Build GRCh38
rs558618577 Chr.5: 171391936-171391936 on Build GRCh38
rs999029930 Chr.5: 171391832-171391832 on Build GRCh38
rs903428980 Chr.5: 171391821-171391821 on Build GRCh38
rs954187506 Chr.5: 171391743-171391743 on Build GRCh38
rs768178660 Chr.5: 171391655-171391655 on Build GRCh38
rs554854531 Chr.5: 171391493-171391493 on Build GRCh38
rs186305190 Chr.5: 171391616-171391616 on Build GRCh38
rs373155923 Chr.5: 171391845-171391845 on Build GRCh38
rs764187017 Chr.5: 171391741-171391741 on Build GRCh38
rs989533113 Chr.5: 171391523-171391526 on Build GRCh38
rs1054874290 Chr.5: 171391570-171391570 on Build GRCh38
rs762836578 Chr.5: 171391697-171391697 on Build GRCh38
rs989977568 Chr.5: 171391778-171391778 on Build GRCh38
rs201813873 Chr.5: 171391768-171391768 on Build GRCh38
rs115157172 Chr.5: 171391917-171391917 on Build GRCh38
rs543430916 Chr.5: 171391526-171391526 on Build GRCh38
rs755036997 Chr.5: 171391522-171391522 on Build GRCh38
rs1013067255 Chr.5: 171391852-171391852 on Build GRCh38
rs138784682 Chr.5: 171391501-171391501 on Build GRCh38
rs550180782 Chr.5: 171391904-171391904 on Build GRCh38
rs776839632 Chr.5: 171391875-171391875 on Build GRCh38
rs932488342 Chr.5: 171391615-171391615 on Build GRCh38
rs773074541 Chr.5: 171391683-171391683 on Build GRCh38
rs984215391 Chr.5: 171391527-171391527 on Build GRCh38
rs1050474807 Chr.5: 171391621-171391621 on Build GRCh38
rs370955818 Chr.5: 171391668-171391668 on Build GRCh38
rs377566222 Chr.5: 171391804-171391804 on Build GRCh38
rs552947652 Chr.5: 171391589-171391589 on Build GRCh38
rs780733077 Chr.5: 171391691-171391692 on Build GRCh38

View Full Product Details