Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 172688920-172689414 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NEURL1B
Entrez Gene ID
54492
Gene Name
neuralized E3 ubiquitin protein ligase 1B
Gene Aliases
NEURL3, hNeur2, neur2
UniGene
Hs.91521
Gene Chromosome Location
Chr.5: 172641266-172691540 on Build GRCh38
Transcripts
NM_001142651.2 NM_001308177.1 NM_001308178.1

SNP Detail

SNP ID Chromosome Location
rs1006859574 Chr.5: 172688981-172688981 on Build GRCh38
rs897898241 Chr.5: 172689235-172689235 on Build GRCh38
rs553113490 Chr.5: 172689131-172689131 on Build GRCh38
rs1000904677 Chr.5: 172689338-172689338 on Build GRCh38
rs544043136 Chr.5: 172689275-172689275 on Build GRCh38
rs770488232 Chr.5: 172689125-172689125 on Build GRCh38
rs917872023 Chr.5: 172689155-172689155 on Build GRCh38
rs548569377 Chr.5: 172689336-172689336 on Build GRCh38
rs1006583005 Chr.5: 172689112-172689112 on Build GRCh38
rs557514966 Chr.5: 172689277-172689277 on Build GRCh38
rs1031983162 Chr.5: 172689268-172689268 on Build GRCh38
rs535586687 Chr.5: 172689228-172689228 on Build GRCh38
rs777685915 Chr.5: 172689339-172689339 on Build GRCh38
rs979852697 Chr.5: 172689370-172689370 on Build GRCh38
rs953610175 Chr.5: 172689045-172689045 on Build GRCh38
rs762458216 Chr.5: 172689033-172689033 on Build GRCh38
rs1036457719 Chr.5: 172689232-172689232 on Build GRCh38
rs1037271449 Chr.5: 172689143-172689143 on Build GRCh38
rs750918650 Chr.5: 172689113-172689113 on Build GRCh38
rs976030035 Chr.5: 172689017-172689017 on Build GRCh38
rs532704787 Chr.5: 172689209-172689209 on Build GRCh38
rs914272892 Chr.5: 172689052-172689053 on Build GRCh38
rs540114128 Chr.5: 172689126-172689126 on Build GRCh38
rs1030625533 Chr.5: 172689044-172689044 on Build GRCh38
rs768104835 Chr.5: 172689039-172689039 on Build GRCh38
rs1007825808 Chr.5: 172689278-172689278 on Build GRCh38
rs1030783426 Chr.5: 172689375-172689375 on Build GRCh38
rs1022269409 Chr.5: 172689289-172689289 on Build GRCh38
rs551183640 Chr.5: 172689016-172689016 on Build GRCh38
rs60482742 Chr.5: 172689387-172689387 on Build GRCh38
rs991944957 Chr.5: 172689136-172689136 on Build GRCh38
rs1027459380 Chr.5: 172688954-172688954 on Build GRCh38
rs185240225 Chr.5: 172689326-172689326 on Build GRCh38
rs752399983 Chr.5: 172689129-172689129 on Build GRCh38
rs190050404 Chr.5: 172689380-172689380 on Build GRCh38
rs571193656 Chr.5: 172689117-172689117 on Build GRCh38
rs997652774 Chr.5: 172689040-172689040 on Build GRCh38
rs995587639 Chr.5: 172688947-172688947 on Build GRCh38
rs918797860 Chr.5: 172689388-172689388 on Build GRCh38
rs909234744 Chr.5: 172689225-172689225 on Build GRCh38
rs1051917536 Chr.5: 172689253-172689253 on Build GRCh38
rs566822681 Chr.5: 172689160-172689160 on Build GRCh38
rs757983576 Chr.5: 172689229-172689229 on Build GRCh38
rs754559613 Chr.5: 172689122-172689122 on Build GRCh38
rs1019618450 Chr.5: 172689242-172689242 on Build GRCh38
rs903520193 Chr.5: 172689311-172689311 on Build GRCh38
rs897426304 Chr.5: 172689192-172689192 on Build GRCh38
rs889889379 Chr.5: 172688978-172688978 on Build GRCh38
rs931441927 Chr.5: 172689206-172689206 on Build GRCh38
rs555784015 Chr.5: 172689238-172689238 on Build GRCh38
rs531083127 Chr.5: 172689015-172689015 on Build GRCh38
rs180854885 Chr.5: 172689258-172689258 on Build GRCh38

View Full Product Details