Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 107490607-107491104 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SOBP
Entrez Gene ID
55084
Gene Name
sine oculis binding protein homolog
Gene Aliases
JXC1, MRAMS
UniGene
Hs.445244
Gene Chromosome Location
Chr.6: 107490104-107661309 on Build GRCh38
Transcripts
NM_018013.3 XM_005267041.4 XM_005267042.4 XM_011535920.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5709788 c.58G>A p.A20T Chr.6: 107490674-107490674 on Build GRCh38
1665766 c.70delA p.R25fs*7 Chr.6: 107490686-107490686 on Build GRCh38
7250465 c.58G>C p.A20P Chr.6: 107490674-107490674 on Build GRCh38
3828729 c.90G>A p.E30E Chr.6: 107490706-107490706 on Build GRCh38
6342772 c.54G>T p.K18N Chr.6: 107490670-107490670 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs917562978 Chr.6: 107490920-107490920 on Build GRCh38
rs913300796 Chr.6: 107490829-107490829 on Build GRCh38
rs1024101562 Chr.6: 107490905-107490905 on Build GRCh38
rs1040863643 Chr.6: 107490966-107490966 on Build GRCh38
rs979995872 Chr.6: 107490910-107490910 on Build GRCh38
rs769145592 Chr.6: 107490716-107490716 on Build GRCh38
rs779077644 Chr.6: 107490648-107490648 on Build GRCh38
rs772976390 Chr.6: 107490736-107490736 on Build GRCh38
rs943432745 Chr.6: 107491011-107491011 on Build GRCh38
rs903304955 Chr.6: 107490642-107490642 on Build GRCh38
rs1055978251 Chr.6: 107490806-107490806 on Build GRCh38
rs992649290 Chr.6: 107490671-107490671 on Build GRCh38
rs1054553702 Chr.6: 107490991-107490991 on Build GRCh38
rs576365103 Chr.6: 107491001-107491001 on Build GRCh38
rs543813164 Chr.6: 107490919-107490919 on Build GRCh38
rs766270606 Chr.6: 107490744-107490744 on Build GRCh38
rs998963281 Chr.6: 107490848-107490848 on Build GRCh38
rs937637879 Chr.6: 107490805-107490805 on Build GRCh38
rs759273483 Chr.6: 107490764-107490764 on Build GRCh38
rs1049282398 Chr.6: 107490998-107490998 on Build GRCh38
rs769465352 Chr.6: 107490734-107490734 on Build GRCh38
rs757358901 Chr.6: 107490644-107490644 on Build GRCh38
rs879319584 Chr.6: 107490787-107490787 on Build GRCh38
rs755151657 Chr.6: 107491027-107491027 on Build GRCh38
rs746140922 Chr.6: 107490654-107490654 on Build GRCh38
rs893145751 Chr.6: 107490864-107490864 on Build GRCh38
rs988750076 Chr.6: 107490937-107490937 on Build GRCh38
rs555880421 Chr.6: 107490790-107490790 on Build GRCh38
rs935997900 Chr.6: 107490794-107490794 on Build GRCh38
rs1024468495 Chr.6: 107491051-107491051 on Build GRCh38
rs762751918 Chr.6: 107490742-107490742 on Build GRCh38
rs996397022 Chr.6: 107490869-107490869 on Build GRCh38
rs532778457 Chr.6: 107491069-107491069 on Build GRCh38
rs944774689 Chr.6: 107490957-107490957 on Build GRCh38
rs950345755 Chr.6: 107490824-107490824 on Build GRCh38
rs777023662 Chr.6: 107490722-107490722 on Build GRCh38
rs761342894 Chr.6: 107490733-107490733 on Build GRCh38
rs984557788 Chr.6: 107490885-107490885 on Build GRCh38
rs970294960 Chr.6: 107490906-107490906 on Build GRCh38
rs34136144 Chr.6: 107491059-107491059 on Build GRCh38
rs781007270 Chr.6: 107490640-107490640 on Build GRCh38
rs758696302 Chr.6: 107490835-107490835 on Build GRCh38
rs370665434 Chr.6: 107490637-107490637 on Build GRCh38
rs373543978 Chr.6: 107491066-107491066 on Build GRCh38
rs773518817 Chr.6: 107490933-107490933 on Build GRCh38
rs181580175 Chr.6: 107491058-107491058 on Build GRCh38
rs535660723 Chr.6: 107490682-107490682 on Build GRCh38
rs73514976 Chr.6: 107490875-107490875 on Build GRCh38
rs965876610 Chr.6: 107490757-107490757 on Build GRCh38
rs984917251 Chr.6: 107490791-107490791 on Build GRCh38
rs1015285222 Chr.6: 107490886-107490886 on Build GRCh38
rs893260212 Chr.6: 107491006-107491006 on Build GRCh38
rs1026178422 Chr.6: 107490881-107490881 on Build GRCh38
rs922486 Chr.6: 107491023-107491023 on Build GRCh38
rs114521467 Chr.6: 107490970-107490970 on Build GRCh38
rs905642778 Chr.6: 107491054-107491054 on Build GRCh38
rs77282648 Chr.6: 107490849-107490849 on Build GRCh38
rs1017611351 Chr.6: 107490873-107490873 on Build GRCh38
rs751300371 Chr.6: 107490761-107490761 on Build GRCh38
rs780466519 Chr.6: 107490693-107490693 on Build GRCh38
rs1017494787 Chr.6: 107490746-107490746 on Build GRCh38
rs999327231 Chr.6: 107490679-107490679 on Build GRCh38
rs1014428233 Chr.6: 107490904-107490904 on Build GRCh38
rs924822676 Chr.6: 107490941-107490941 on Build GRCh38
rs961122007 Chr.6: 107490903-107490903 on Build GRCh38
rs1005203734 Chr.6: 107490882-107490882 on Build GRCh38
rs747341292 Chr.6: 107490694-107490694 on Build GRCh38
rs766665760 Chr.6: 107491004-107491004 on Build GRCh38
rs754610339 Chr.6: 107491049-107491049 on Build GRCh38
rs919083320 Chr.6: 107490780-107490780 on Build GRCh38
rs957257063 Chr.6: 107491075-107491075 on Build GRCh38
rs753907639 Chr.6: 107490643-107490643 on Build GRCh38
rs566085603 Chr.6: 107490814-107490814 on Build GRCh38
rs908898931 Chr.6: 107490798-107490798 on Build GRCh38
rs944805579 Chr.6: 107490842-107490842 on Build GRCh38
rs541663369 Chr.6: 107491016-107491016 on Build GRCh38
rs916197238 Chr.6: 107490809-107490809 on Build GRCh38
rs975976809 Chr.6: 107490776-107490776 on Build GRCh38
rs925861699 Chr.6: 107490917-107490917 on Build GRCh38
rs932128450 Chr.6: 107490971-107490971 on Build GRCh38
rs913248467 Chr.6: 107490944-107490944 on Build GRCh38
rs888058089 Chr.6: 107490879-107490879 on Build GRCh38
rs755008729 Chr.6: 107490635-107490635 on Build GRCh38
rs1040519335 Chr.6: 107490845-107490845 on Build GRCh38
rs527428740 Chr.6: 107491053-107491053 on Build GRCh38
rs576874917 Chr.6: 107490819-107490819 on Build GRCh38
rs1053080910 Chr.6: 107490797-107490797 on Build GRCh38

View Full Product Details