Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 93129742-93130244 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SAMD9L
Entrez Gene ID
219285
Gene Name
sterile alpha motif domain containing 9 like
Gene Aliases
C7orf6, DRIF2, UEF1
UniGene
Hs.489118
Gene Chromosome Location
Chr.7: 93130054-93148401 on Build GRCh38
Transcripts
NM_001303496.1 NM_001303497.1 NM_001303498.1 NM_001303500.1 NM_152703.3 XM_005250193.3 XM_006715890.1 XM_011515903.2 XM_017011822.1 XM_017011823.1

SNP Detail

SNP ID Chromosome Location
rs972759084 Chr.7: 93130105-93130105 on Build GRCh38
rs572196154 Chr.7: 93129931-93129931 on Build GRCh38
rs1001283971 Chr.7: 93130060-93130060 on Build GRCh38
rs892029048 Chr.7: 93129990-93129990 on Build GRCh38
rs748415379 Chr.7: 93130083-93130083 on Build GRCh38
rs755355733 Chr.7: 93130169-93130169 on Build GRCh38
rs909009845 Chr.7: 93130167-93130167 on Build GRCh38
rs985034426 Chr.7: 93129786-93129786 on Build GRCh38
rs970233805 Chr.7: 93130012-93130012 on Build GRCh38
rs80349270 Chr.7: 93130185-93130185 on Build GRCh38
rs771199634 Chr.7: 93129776-93129776 on Build GRCh38
rs184085127 Chr.7: 93130132-93130132 on Build GRCh38
rs929256877 Chr.7: 93129898-93129898 on Build GRCh38
rs776062873 Chr.7: 93129785-93129785 on Build GRCh38
rs962322981 Chr.7: 93129831-93129831 on Build GRCh38
rs753458812 Chr.7: 93129956-93129956 on Build GRCh38
rs771930266 Chr.7: 93129863-93129863 on Build GRCh38
rs997183204 Chr.7: 93129974-93129974 on Build GRCh38
rs111881813 Chr.7: 93130201-93130201 on Build GRCh38
rs914766997 Chr.7: 93129814-93129814 on Build GRCh38
rs935800851 Chr.7: 93129912-93129912 on Build GRCh38
rs775081975 Chr.7: 93129944-93129944 on Build GRCh38
rs558394764 Chr.7: 93130131-93130131 on Build GRCh38
rs960436267 Chr.7: 93130099-93130099 on Build GRCh38
rs918609369 Chr.7: 93130120-93130120 on Build GRCh38
rs1056751508 Chr.7: 93129864-93129864 on Build GRCh38
rs1006355084 Chr.7: 93130107-93130107 on Build GRCh38
rs527433393 Chr.7: 93129825-93129825 on Build GRCh38
rs553616814 Chr.7: 93129827-93129827 on Build GRCh38
rs891686203 Chr.7: 93129952-93129952 on Build GRCh38
rs1015113593 Chr.7: 93130109-93130109 on Build GRCh38
rs980145042 Chr.7: 93130048-93130048 on Build GRCh38
rs749507531 Chr.7: 93129773-93129773 on Build GRCh38
rs572600136 Chr.7: 93130146-93130146 on Build GRCh38
rs919174491 Chr.7: 93129865-93129865 on Build GRCh38
rs1034135392 Chr.7: 93130072-93130072 on Build GRCh38
rs540006066 Chr.7: 93129991-93129991 on Build GRCh38
rs4267 Chr.7: 93130149-93130149 on Build GRCh38
rs1035786434 Chr.7: 93130070-93130070 on Build GRCh38
rs759257706 Chr.7: 93129836-93129836 on Build GRCh38
rs990331348 Chr.7: 93129797-93129797 on Build GRCh38
rs943212184 Chr.7: 93130182-93130182 on Build GRCh38
rs189107403 Chr.7: 93129911-93129911 on Build GRCh38
rs887535187 Chr.7: 93130089-93130089 on Build GRCh38
rs760711977 Chr.7: 93129922-93129922 on Build GRCh38
rs1038760728 Chr.7: 93129949-93129949 on Build GRCh38
rs1031793311 Chr.7: 93129979-93129979 on Build GRCh38
rs970117838 Chr.7: 93129823-93129823 on Build GRCh38
rs73710961 Chr.7: 93130009-93130009 on Build GRCh38
rs1034499827 Chr.7: 93129784-93129784 on Build GRCh38
rs984689100 Chr.7: 93130154-93130154 on Build GRCh38
rs933089277 Chr.7: 93130213-93130213 on Build GRCh38
rs945999983 Chr.7: 93129966-93129966 on Build GRCh38
rs148957698 Chr.7: 93129953-93129953 on Build GRCh38
rs191654917 Chr.7: 93129930-93129930 on Build GRCh38
rs79801126 Chr.7: 93129832-93129832 on Build GRCh38

View Full Product Details