Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 135032750-135033248 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGBL3
Entrez Gene ID
340351
Gene Name
ATP/GTP binding protein like 3
Gene Aliases
CCP3
UniGene
Hs.648616
Gene Chromosome Location
Chr.7: 134985169-135135782 on Build GRCh38
Transcripts
NM_178563.3 XM_011516122.2 XM_017012136.1 XM_017012137.1 XM_017012138.1 XM_017012139.1 XM_017012140.1 XM_017012141.1 XM_017012142.1 XM_017012143.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5062194 c.422A>G p.Y141C Chr.7: 135032847-135032847 on Build GRCh38
6346947 c.371G>T p.R124L Chr.7: 135032874-135032874 on Build GRCh38
3784997 c.371G>A p.R124Q Chr.7: 135032874-135032874 on Build GRCh38
1085796 c.429G>A p.E143E Chr.7: 135032854-135032854 on Build GRCh38
4359911 c.392C>T p.P131L Chr.7: 135032895-135032895 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs942853277 Chr.7: 135033126-135033126 on Build GRCh38
rs1047072477 Chr.7: 135033183-135033183 on Build GRCh38
rs73725210 Chr.7: 135032777-135032777 on Build GRCh38
rs772358275 Chr.7: 135032923-135032923 on Build GRCh38
rs377744197 Chr.7: 135032989-135032989 on Build GRCh38
rs956504999 Chr.7: 135032978-135032978 on Build GRCh38
rs377179946 Chr.7: 135033044-135033044 on Build GRCh38
rs750695479 Chr.7: 135032873-135032873 on Build GRCh38
rs963684360 Chr.7: 135033082-135033082 on Build GRCh38
rs911357761 Chr.7: 135033094-135033094 on Build GRCh38
rs530113535 Chr.7: 135032827-135032829 on Build GRCh38
rs901299301 Chr.7: 135032889-135032889 on Build GRCh38
rs76055041 Chr.7: 135032928-135032928 on Build GRCh38
rs756567736 Chr.7: 135032877-135032877 on Build GRCh38
rs1035179541 Chr.7: 135032971-135032971 on Build GRCh38
rs765648802 Chr.7: 135033222-135033222 on Build GRCh38
rs922333507 Chr.7: 135033124-135033124 on Build GRCh38
rs968915918 Chr.7: 135033075-135033075 on Build GRCh38
rs369736068 Chr.7: 135032817-135032817 on Build GRCh38
rs377156405 Chr.7: 135032897-135032897 on Build GRCh38
rs1041242055 Chr.7: 135033127-135033127 on Build GRCh38
rs531216809 Chr.7: 135032918-135032918 on Build GRCh38
rs941397046 Chr.7: 135032888-135032888 on Build GRCh38
rs547074424 Chr.7: 135033129-135033129 on Build GRCh38
rs910853527 Chr.7: 135032784-135032784 on Build GRCh38
rs372989688 Chr.7: 135032874-135032874 on Build GRCh38
rs1043175489 Chr.7: 135032858-135032858 on Build GRCh38
rs989771487 Chr.7: 135033029-135033029 on Build GRCh38
rs548196709 Chr.7: 135033093-135033093 on Build GRCh38
rs982967760 Chr.7: 135033186-135033186 on Build GRCh38
rs191982287 Chr.7: 135033182-135033182 on Build GRCh38
rs542032407 Chr.7: 135032850-135032850 on Build GRCh38
rs372654444 Chr.7: 135033190-135033190 on Build GRCh38
rs947668228 Chr.7: 135032802-135032802 on Build GRCh38
rs888452572 Chr.7: 135033225-135033225 on Build GRCh38
rs780847001 Chr.7: 135032866-135032866 on Build GRCh38
rs9656446 Chr.7: 135032905-135032905 on Build GRCh38
rs372990412 Chr.7: 135032960-135032960 on Build GRCh38
rs147068130 Chr.7: 135033019-135033019 on Build GRCh38
rs544646807 Chr.7: 135032919-135032919 on Build GRCh38
rs777982591 Chr.7: 135033129-135033129 on Build GRCh38
rs751185311 Chr.7: 135032816-135032816 on Build GRCh38
rs922668126 Chr.7: 135033167-135033167 on Build GRCh38
rs78700550 Chr.7: 135032932-135032932 on Build GRCh38
rs772562346 Chr.7: 135032981-135032981 on Build GRCh38
rs913466481 Chr.7: 135032791-135032791 on Build GRCh38
rs78253659 Chr.7: 135032930-135032930 on Build GRCh38
rs910000573 Chr.7: 135033227-135033227 on Build GRCh38
rs773938960 Chr.7: 135032895-135032895 on Build GRCh38

View Full Product Details