Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 142018965-142019491 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MGAM
Entrez Gene ID
8972
Gene Name
maltase-glucoamylase
Gene Aliases
MG, MGA
UniGene
Hs.122785
Gene Chromosome Location
Chr.7: 141986517-142106747 on Build GRCh38
Transcripts
NM_004668.2 XM_006716168.2 XM_011516670.2 XM_011516671.2 XM_011516672.2 XM_011516673.2 XM_011516674.2 XM_017012772.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3634412 c.368G>A p.G123E Chr.7: 142019239-142019239 on Build GRCh38
231133 c.368G>A p.G123E Chr.7: 142019239-142019239 on Build GRCh38
5534528 c.399C>T p.S133S Chr.7: 142019270-142019270 on Build GRCh38
4955347 c.372T>A p.A124A Chr.7: 142019243-142019243 on Build GRCh38
5680209 c.377G>A p.S126N Chr.7: 142019248-142019248 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781892890 Chr.7: 142019317-142019317 on Build GRCh38
rs918297383 Chr.7: 142019337-142019337 on Build GRCh38
rs544305672 Chr.7: 142019407-142019407 on Build GRCh38
rs557660275 Chr.7: 142019113-142019113 on Build GRCh38
rs782272356 Chr.7: 142019244-142019244 on Build GRCh38
rs3793158 Chr.7: 142019152-142019152 on Build GRCh38
rs373190169 Chr.7: 142019342-142019342 on Build GRCh38
rs782504315 Chr.7: 142019217-142019217 on Build GRCh38
rs782552577 Chr.7: 142019221-142019221 on Build GRCh38
rs781792206 Chr.7: 142019220-142019220 on Build GRCh38
rs549195092 Chr.7: 142019449-142019449 on Build GRCh38
rs374528123 Chr.7: 142019203-142019203 on Build GRCh38
rs781914209 Chr.7: 142019420-142019420 on Build GRCh38
rs781829209 Chr.7: 142019248-142019249 on Build GRCh38
rs781950351 Chr.7: 142019171-142019171 on Build GRCh38
rs576626552 Chr.7: 142019230-142019230 on Build GRCh38
rs1052451919 Chr.7: 142019445-142019445 on Build GRCh38
rs540337133 Chr.7: 142019330-142019330 on Build GRCh38
rs782768460 Chr.7: 142019199-142019199 on Build GRCh38
rs1358305 Chr.7: 142019151-142019151 on Build GRCh38
rs782246538 Chr.7: 142019166-142019166 on Build GRCh38
rs782003414 Chr.7: 142019294-142019294 on Build GRCh38
rs782730354 Chr.7: 142019214-142019214 on Build GRCh38
rs782141625 Chr.7: 142019318-142019318 on Build GRCh38
rs782204750 Chr.7: 142019256-142019256 on Build GRCh38
rs116863299 Chr.7: 142019433-142019433 on Build GRCh38
rs367663629 Chr.7: 142019333-142019333 on Build GRCh38
rs782682290 Chr.7: 142019336-142019336 on Build GRCh38
rs371564136 Chr.7: 142019312-142019312 on Build GRCh38
rs782421619 Chr.7: 142019324-142019324 on Build GRCh38
rs903848108 Chr.7: 142019410-142019410 on Build GRCh38
rs782145209 Chr.7: 142019185-142019185 on Build GRCh38
rs782771834 Chr.7: 142019319-142019319 on Build GRCh38
rs782659702 Chr.7: 142019340-142019340 on Build GRCh38
rs782365005 Chr.7: 142019167-142019167 on Build GRCh38
rs782531491 Chr.7: 142019359-142019360 on Build GRCh38
rs538932611 Chr.7: 142019106-142019106 on Build GRCh38
rs903878682 Chr.7: 142019095-142019095 on Build GRCh38
rs1018245462 Chr.7: 142019092-142019092 on Build GRCh38
rs575560438 Chr.7: 142019134-142019134 on Build GRCh38
rs782063832 Chr.7: 142019172-142019172 on Build GRCh38
rs370701658 Chr.7: 142019272-142019272 on Build GRCh38
rs782616300 Chr.7: 142019251-142019251 on Build GRCh38
rs782811489 Chr.7: 142019316-142019316 on Build GRCh38
rs782208449 Chr.7: 142019365-142019365 on Build GRCh38
rs782318002 Chr.7: 142019267-142019267 on Build GRCh38
rs571233874 Chr.7: 142019050-142019050 on Build GRCh38
rs929713443 Chr.7: 142019351-142019351 on Build GRCh38
rs782029813 Chr.7: 142019181-142019181 on Build GRCh38
rs200976716 Chr.7: 142019327-142019327 on Build GRCh38
rs973841028 Chr.7: 142019328-142019328 on Build GRCh38
rs969584780 Chr.7: 142019241-142019241 on Build GRCh38
rs782374827 Chr.7: 142019348-142019348 on Build GRCh38
rs782517449 Chr.7: 142019250-142019250 on Build GRCh38
rs782693116 Chr.7: 142019173-142019173 on Build GRCh38
rs782466804 Chr.7: 142019213-142019213 on Build GRCh38
rs201845079 Chr.7: 142019222-142019222 on Build GRCh38
rs782612986 Chr.7: 142019358-142019358 on Build GRCh38
rs376282334 Chr.7: 142019226-142019226 on Build GRCh38
rs552649570 Chr.7: 142019026-142019026 on Build GRCh38
rs184890060 Chr.7: 142019406-142019406 on Build GRCh38
rs188476601 Chr.7: 142019448-142019448 on Build GRCh38
rs782291424 Chr.7: 142019275-142019275 on Build GRCh38
rs912411182 Chr.7: 142019381-142019381 on Build GRCh38
rs114936410 Chr.7: 142019278-142019278 on Build GRCh38
rs368074859 Chr.7: 142019295-142019295 on Build GRCh38
rs981460832 Chr.7: 142019246-142019246 on Build GRCh38
rs782262071 Chr.7: 142019343-142019343 on Build GRCh38

View Full Product Details