Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 18534572-18535109 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSD3
Entrez Gene ID
23362
Gene Name
pleckstrin and Sec7 domain containing 3
Gene Aliases
EFA6D, EFA6R, HCA67
UniGene
Hs.434255
Gene Chromosome Location
Chr.8: 18527303-19086909 on Build GRCh38
Transcripts
NM_015310.3 NM_206909.2 XM_017013258.1 XM_017013259.1 XM_017013260.1 XM_017013261.1 XM_017013265.1

SNP Detail

SNP ID Chromosome Location
rs142539319 Chr.8: 18535014-18535014 on Build GRCh38
rs954963460 Chr.8: 18534920-18534920 on Build GRCh38
rs1023406167 Chr.8: 18534983-18534983 on Build GRCh38
rs778056594 Chr.8: 18535021-18535021 on Build GRCh38
rs917131455 Chr.8: 18535019-18535019 on Build GRCh38
rs764917499 Chr.8: 18534639-18534639 on Build GRCh38
rs529118464 Chr.8: 18534622-18534622 on Build GRCh38
rs951613884 Chr.8: 18534624-18534624 on Build GRCh38
rs569477562 Chr.8: 18534972-18534972 on Build GRCh38
rs555190927 Chr.8: 18534743-18534743 on Build GRCh38
rs868731094 Chr.8: 18534842-18534842 on Build GRCh38
rs933350392 Chr.8: 18534761-18534761 on Build GRCh38
rs534399673 Chr.8: 18535057-18535057 on Build GRCh38
rs935204045 Chr.8: 18534830-18534830 on Build GRCh38
rs970447755 Chr.8: 18534825-18534825 on Build GRCh38
rs1035331332 Chr.8: 18534671-18534671 on Build GRCh38
rs1039825942 Chr.8: 18534872-18534872 on Build GRCh38
rs967276958 Chr.8: 18534690-18534690 on Build GRCh38
rs923255643 Chr.8: 18534757-18534757 on Build GRCh38
rs901211247 Chr.8: 18534771-18534771 on Build GRCh38
rs747953368 Chr.8: 18534876-18534876 on Build GRCh38
rs754928276 Chr.8: 18534869-18534869 on Build GRCh38
rs1031050575 Chr.8: 18534997-18534997 on Build GRCh38
rs1041355150 Chr.8: 18535046-18535046 on Build GRCh38
rs1005725051 Chr.8: 18534951-18534951 on Build GRCh38
rs777701618 Chr.8: 18534821-18534822 on Build GRCh38
rs1032931284 Chr.8: 18534719-18534719 on Build GRCh38
rs772130773 Chr.8: 18534755-18534758 on Build GRCh38
rs964960856 Chr.8: 18534720-18534720 on Build GRCh38
rs560025813 Chr.8: 18534595-18534595 on Build GRCh38
rs148432348 Chr.8: 18534982-18534982 on Build GRCh38
rs746955367 Chr.8: 18534826-18534826 on Build GRCh38
rs761798469 Chr.8: 18534816-18534816 on Build GRCh38
rs927138015 Chr.8: 18534807-18534807 on Build GRCh38
rs569558744 Chr.8: 18534670-18534670 on Build GRCh38
rs1030572909 Chr.8: 18534898-18534898 on Build GRCh38
rs533364945 Chr.8: 18534791-18534791 on Build GRCh38
rs562729204 Chr.8: 18534682-18534682 on Build GRCh38
rs118023047 Chr.8: 18534969-18534969 on Build GRCh38
rs766979103 Chr.8: 18534651-18534651 on Build GRCh38
rs537004653 Chr.8: 18534688-18534688 on Build GRCh38
rs1025926532 Chr.8: 18534800-18534813 on Build GRCh38
rs867671921 Chr.8: 18534989-18534989 on Build GRCh38
rs183248530 Chr.8: 18534785-18534785 on Build GRCh38
rs999927537 Chr.8: 18535060-18535060 on Build GRCh38
rs942120330 Chr.8: 18534778-18534778 on Build GRCh38
rs536177284 Chr.8: 18534867-18534867 on Build GRCh38
rs771994709 Chr.8: 18534818-18534818 on Build GRCh38
rs577413624 Chr.8: 18535083-18535083 on Build GRCh38
rs996370891 Chr.8: 18534887-18534887 on Build GRCh38
rs546985007 Chr.8: 18534820-18534820 on Build GRCh38
rs570951230 Chr.8: 18534788-18534788 on Build GRCh38
rs979583840 Chr.8: 18534718-18534718 on Build GRCh38
rs934032682 Chr.8: 18534877-18534877 on Build GRCh38
rs887374134 Chr.8: 18534905-18534905 on Build GRCh38
rs866570536 Chr.8: 18534841-18534841 on Build GRCh38
rs542478411 Chr.8: 18534672-18534672 on Build GRCh38
rs937222867 Chr.8: 18534810-18534810 on Build GRCh38
rs1009978093 Chr.8: 18534683-18534683 on Build GRCh38
rs780196791 Chr.8: 18534806-18534806 on Build GRCh38
rs555888389 Chr.8: 18534935-18534935 on Build GRCh38
rs973920211 Chr.8: 18534974-18534974 on Build GRCh38
rs904237485 Chr.8: 18534835-18534835 on Build GRCh38
rs865866286 Chr.8: 18534669-18534669 on Build GRCh38
rs552777279 Chr.8: 18535081-18535081 on Build GRCh38
rs907915429 Chr.8: 18534777-18534777 on Build GRCh38
rs945952599 Chr.8: 18535045-18535045 on Build GRCh38
rs982084965 Chr.8: 18534713-18534713 on Build GRCh38
rs983499697 Chr.8: 18534764-18534764 on Build GRCh38
rs923898706 Chr.8: 18534828-18534828 on Build GRCh38
rs905748770 Chr.8: 18534628-18534628 on Build GRCh38
rs970858383 Chr.8: 18534986-18534986 on Build GRCh38
rs907882704 Chr.8: 18534970-18534970 on Build GRCh38
rs187657952 Chr.8: 18534821-18534821 on Build GRCh38
rs780874262 Chr.8: 18534875-18534875 on Build GRCh38
rs367700748 Chr.8: 18534742-18534742 on Build GRCh38
rs916215970 Chr.8: 18534689-18534689 on Build GRCh38
rs915036819 Chr.8: 18534839-18534839 on Build GRCh38
rs776616120 Chr.8: 18535011-18535011 on Build GRCh38
rs895477157 Chr.8: 18534817-18534817 on Build GRCh38
rs1017029002 Chr.8: 18534814-18534814 on Build GRCh38

View Full Product Details