Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 6329435-6329939 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TPD52L3
Entrez Gene ID
89882
Gene Name
tumor protein D52-like 3
Gene Aliases
NYDSP25, hD55
UniGene
Hs.351815
Gene Chromosome Location
Chr.9: 6328349-6331900 on Build GRCh38
Transcripts
NM_001001874.2 NM_001001875.3 NM_033516.5 XM_017015280.1

SNP Detail

SNP ID Chromosome Location
rs1029269039 Chr.9: 6329572-6329575 on Build GRCh38
rs1023275654 Chr.9: 6329884-6329884 on Build GRCh38
rs997817061 Chr.9: 6329571-6329572 on Build GRCh38
rs900604539 Chr.9: 6329670-6329670 on Build GRCh38
rs898673 Chr.9: 6329888-6329888 on Build GRCh38
rs1046969863 Chr.9: 6329497-6329497 on Build GRCh38
rs377002567 Chr.9: 6329656-6329656 on Build GRCh38
rs1040315960 Chr.9: 6329631-6329631 on Build GRCh38
rs796095679 Chr.9: 6329851-6329851 on Build GRCh38
rs929933948 Chr.9: 6329533-6329533 on Build GRCh38
rs183444195 Chr.9: 6329757-6329757 on Build GRCh38
rs149439096 Chr.9: 6329785-6329785 on Build GRCh38
rs907915138 Chr.9: 6329584-6329584 on Build GRCh38
rs1020445350 Chr.9: 6329663-6329663 on Build GRCh38
rs35834483 Chr.9: 6329867-6329867 on Build GRCh38
rs944719633 Chr.9: 6329601-6329601 on Build GRCh38
rs904015921 Chr.9: 6329718-6329718 on Build GRCh38
rs538802882 Chr.9: 6329468-6329468 on Build GRCh38
rs1013276495 Chr.9: 6329798-6329798 on Build GRCh38
rs917251602 Chr.9: 6329745-6329745 on Build GRCh38
rs888267997 Chr.9: 6329499-6329499 on Build GRCh38
rs571679231 Chr.9: 6329459-6329459 on Build GRCh38
rs1005350510 Chr.9: 6329503-6329503 on Build GRCh38
rs995646810 Chr.9: 6329889-6329889 on Build GRCh38
rs1057131663 Chr.9: 6329456-6329456 on Build GRCh38
rs748219425 Chr.9: 6329750-6329751 on Build GRCh38
rs958957464 Chr.9: 6329716-6329716 on Build GRCh38
rs1039509477 Chr.9: 6329826-6329826 on Build GRCh38
rs550690829 Chr.9: 6329715-6329715 on Build GRCh38
rs899723883 Chr.9: 6329570-6329570 on Build GRCh38
rs1052693936 Chr.9: 6329771-6329771 on Build GRCh38
rs927556347 Chr.9: 6329711-6329711 on Build GRCh38
rs1009271112 Chr.9: 6329645-6329645 on Build GRCh38
rs992858432 Chr.9: 6329737-6329737 on Build GRCh38
rs1026859637 Chr.9: 6329898-6329898 on Build GRCh38
rs932032408 Chr.9: 6329700-6329700 on Build GRCh38
rs1043921951 Chr.9: 6329709-6329709 on Build GRCh38
rs930025945 Chr.9: 6329482-6329482 on Build GRCh38
rs532485025 Chr.9: 6329574-6329574 on Build GRCh38
rs1040027098 Chr.9: 6329564-6329564 on Build GRCh38
rs956481613 Chr.9: 6329637-6329637 on Build GRCh38

View Full Product Details