Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136742484-136742979 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LCN10
Entrez Gene ID
414332
Gene Name
lipocalin 10
Gene Aliases
-
UniGene
Hs.98132
Gene Chromosome Location
Chr.9: 136738167-136742959 on Build GRCh38
Transcripts
NM_001001712.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7401559 c.117+3G>A p.? Chr.9: 136742784-136742784 on Build GRCh38
5410762 c.110G>A p.W37* Chr.9: 136742794-136742794 on Build GRCh38
6700021 c.58G>A p.A20T Chr.9: 136742846-136742846 on Build GRCh38
4785882 c.22C>T p.L8L Chr.9: 136742882-136742882 on Build GRCh38
6837594 c.31G>C p.V11L Chr.9: 136742873-136742873 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs918671201 Chr.9: 136742654-136742654 on Build GRCh38
rs746148898 Chr.9: 136742745-136742745 on Build GRCh38
rs780264092 Chr.9: 136742866-136742867 on Build GRCh38
rs1000588224 Chr.9: 136742586-136742586 on Build GRCh38
rs552832322 Chr.9: 136742581-136742581 on Build GRCh38
rs1040365557 Chr.9: 136742526-136742526 on Build GRCh38
rs751437336 Chr.9: 136742930-136742930 on Build GRCh38
rs147718407 Chr.9: 136742869-136742869 on Build GRCh38
rs537766259 Chr.9: 136742588-136742588 on Build GRCh38
rs753220737 Chr.9: 136742895-136742895 on Build GRCh38
rs992269539 Chr.9: 136742594-136742594 on Build GRCh38
rs996068218 Chr.9: 136742542-136742542 on Build GRCh38
rs779380443 Chr.9: 136742888-136742888 on Build GRCh38
rs373886246 Chr.9: 136742826-136742826 on Build GRCh38
rs1006801218 Chr.9: 136742812-136742812 on Build GRCh38
rs886460759 Chr.9: 136742508-136742508 on Build GRCh38
rs779285446 Chr.9: 136742814-136742814 on Build GRCh38
rs199525822 Chr.9: 136742925-136742925 on Build GRCh38
rs752682247 Chr.9: 136742796-136742796 on Build GRCh38
rs999261463 Chr.9: 136742744-136742744 on Build GRCh38
rs112615182 Chr.9: 136742650-136742650 on Build GRCh38
rs769695406 Chr.9: 136742849-136742849 on Build GRCh38
rs951504019 Chr.9: 136742655-136742655 on Build GRCh38
rs754539105 Chr.9: 136742742-136742743 on Build GRCh38
rs984301150 Chr.9: 136742660-136742660 on Build GRCh38
rs537227677 Chr.9: 136742595-136742595 on Build GRCh38
rs937558139 Chr.9: 136742696-136742696 on Build GRCh38
rs148733656 Chr.9: 136742822-136742822 on Build GRCh38
rs940777422 Chr.9: 136742936-136742936 on Build GRCh38
rs1034627139 Chr.9: 136742873-136742873 on Build GRCh38
rs776249843 Chr.9: 136742774-136742774 on Build GRCh38
rs771771769 Chr.9: 136742755-136742755 on Build GRCh38
rs762920275 Chr.9: 136742874-136742874 on Build GRCh38
rs763986466 Chr.9: 136742924-136742924 on Build GRCh38
rs35227454 Chr.9: 136742836-136742837 on Build GRCh38
rs560690833 Chr.9: 136742737-136742737 on Build GRCh38
rs367668289 Chr.9: 136742706-136742706 on Build GRCh38
rs532809000 Chr.9: 136742748-136742748 on Build GRCh38
rs546529777 Chr.9: 136742756-136742756 on Build GRCh38
rs558460253 Chr.9: 136742956-136742956 on Build GRCh38
rs1025176853 Chr.9: 136742566-136742567 on Build GRCh38
rs888718541 Chr.9: 136742513-136742513 on Build GRCh38
rs768785071 Chr.9: 136742877-136742878 on Build GRCh38
rs376143495 Chr.9: 136742795-136742795 on Build GRCh38
rs767635311 Chr.9: 136742801-136742801 on Build GRCh38
rs772982702 Chr.9: 136742757-136742757 on Build GRCh38
rs1000683069 Chr.9: 136742798-136742798 on Build GRCh38
rs940780386 Chr.9: 136742672-136742672 on Build GRCh38
rs758929944 Chr.9: 136742896-136742896 on Build GRCh38
rs757024366 Chr.9: 136742867-136742878 on Build GRCh38
rs780437925 Chr.9: 136742750-136742750 on Build GRCh38
rs752738111 Chr.9: 136742855-136742856 on Build GRCh38
rs760450120 Chr.9: 136742758-136742758 on Build GRCh38
rs758636588 Chr.9: 136742754-136742754 on Build GRCh38
rs770477339 Chr.9: 136742909-136742909 on Build GRCh38
rs762760318 Chr.9: 136742946-136742946 on Build GRCh38
rs754158039 Chr.9: 136742883-136742883 on Build GRCh38
rs776984591 Chr.9: 136742510-136742510 on Build GRCh38
rs766447912 Chr.9: 136742882-136742882 on Build GRCh38
rs750454131 Chr.9: 136742877-136742877 on Build GRCh38
rs368044613 Chr.9: 136742916-136742916 on Build GRCh38
rs538502200 Chr.9: 136742934-136742934 on Build GRCh38
rs980316640 Chr.9: 136742622-136742622 on Build GRCh38
rs907983653 Chr.9: 136742666-136742666 on Build GRCh38
rs764834501 Chr.9: 136742764-136742764 on Build GRCh38
rs1056487527 Chr.9: 136742697-136742697 on Build GRCh38
rs749511624 Chr.9: 136742872-136742873 on Build GRCh38
rs771683531 Chr.9: 136742817-136742817 on Build GRCh38
rs923282749 Chr.9: 136742959-136742959 on Build GRCh38
rs540788442 Chr.9: 136742718-136742718 on Build GRCh38
rs980370452 Chr.9: 136742857-136742857 on Build GRCh38
rs10870117 Chr.9: 136742635-136742635 on Build GRCh38
rs758243348 Chr.9: 136742856-136742857 on Build GRCh38
rs956469036 Chr.9: 136742589-136742589 on Build GRCh38
rs564865038 Chr.9: 136742804-136742804 on Build GRCh38
rs749697591 Chr.9: 136742751-136742751 on Build GRCh38
rs142578827 Chr.9: 136742878-136742878 on Build GRCh38
rs894827032 Chr.9: 136742551-136742551 on Build GRCh38
rs902287013 Chr.9: 136742728-136742728 on Build GRCh38
rs759185196 Chr.9: 136742829-136742829 on Build GRCh38
rs574921181 Chr.9: 136742707-136742707 on Build GRCh38
rs901794064 Chr.9: 136742531-136742531 on Build GRCh38
rs748719536 Chr.9: 136742815-136742815 on Build GRCh38
rs901808065 Chr.9: 136742713-136742713 on Build GRCh38
rs930776931 Chr.9: 136742647-136742647 on Build GRCh38
rs575241223 Chr.9: 136742643-136742643 on Build GRCh38
rs557345932 Chr.9: 136742621-136742621 on Build GRCh38
rs967825686 Chr.9: 136742568-136742568 on Build GRCh38
rs759397579 Chr.9: 136742780-136742780 on Build GRCh38
rs1013332314 Chr.9: 136742563-136742563 on Build GRCh38
rs941646603 Chr.9: 136742515-136742515 on Build GRCh38
rs201151522 Chr.9: 136742805-136742805 on Build GRCh38
rs749561705 Chr.9: 136742808-136742808 on Build GRCh38
rs200886607 Chr.9: 136742885-136742885 on Build GRCh38
rs147075752 Chr.9: 136742820-136742820 on Build GRCh38
rs1054785412 Chr.9: 136742521-136742521 on Build GRCh38
rs777461594 Chr.9: 136742818-136742818 on Build GRCh38
rs1028789582 Chr.9: 136742548-136742548 on Build GRCh38
rs11145858 Chr.9: 136742649-136742649 on Build GRCh38
rs910079094 Chr.9: 136742669-136742669 on Build GRCh38
rs371934266 Chr.9: 136742753-136742753 on Build GRCh38
rs778636349 Chr.9: 136742746-136742746 on Build GRCh38
rs766241890 Chr.9: 136742766-136742766 on Build GRCh38
rs902319907 Chr.9: 136742567-136742567 on Build GRCh38
rs972060231 Chr.9: 136742603-136742603 on Build GRCh38
rs750613268 Chr.9: 136742803-136742803 on Build GRCh38
rs778445885 Chr.9: 136742902-136742902 on Build GRCh38
rs746459804 Chr.9: 136742907-136742907 on Build GRCh38
rs751337850 Chr.9: 136742857-136742862 on Build GRCh38

View Full Product Details