Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127803147-127803651 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FPGS
Entrez Gene ID
2356
Gene Name
folylpolyglutamate synthase
Gene Aliases
-
UniGene
Hs.335084
Gene Chromosome Location
Chr.9: 127802858-127814520 on Build GRCh38
Transcripts
NM_001018078.2 NM_001288803.1 NM_004957.5 NR_110170.1 XM_005251864.3 XM_017014565.1 XM_017014566.1

SNP Detail

SNP ID Chromosome Location
rs78924945 Chr.9: 127803427-127803427 on Build GRCh38
rs114906420 Chr.9: 127803286-127803286 on Build GRCh38
rs1014032496 Chr.9: 127803182-127803182 on Build GRCh38
rs949728893 Chr.9: 127803368-127803368 on Build GRCh38
rs930131149 Chr.9: 127803551-127803551 on Build GRCh38
rs895462892 Chr.9: 127803175-127803176 on Build GRCh38
rs1055503292 Chr.9: 127803175-127803175 on Build GRCh38
rs1037913314 Chr.9: 127803384-127803384 on Build GRCh38
rs761572280 Chr.9: 127803591-127803591 on Build GRCh38
rs545778336 Chr.9: 127803488-127803488 on Build GRCh38
rs986316496 Chr.9: 127803237-127803237 on Build GRCh38
rs977328487 Chr.9: 127803546-127803546 on Build GRCh38
rs967475080 Chr.9: 127803525-127803525 on Build GRCh38
rs967199579 Chr.9: 127803459-127803459 on Build GRCh38
rs182106590 Chr.9: 127803501-127803501 on Build GRCh38
rs910307227 Chr.9: 127803500-127803500 on Build GRCh38
rs747728437 Chr.9: 127803324-127803324 on Build GRCh38
rs1005027355 Chr.9: 127803192-127803192 on Build GRCh38
rs10987742 Chr.9: 127803598-127803598 on Build GRCh38
rs901442563 Chr.9: 127803299-127803299 on Build GRCh38
rs144076306 Chr.9: 127803485-127803485 on Build GRCh38
rs974990425 Chr.9: 127803241-127803241 on Build GRCh38
rs955171 Chr.9: 127803528-127803528 on Build GRCh38
rs769467234 Chr.9: 127803382-127803382 on Build GRCh38
rs150703077 Chr.9: 127803578-127803578 on Build GRCh38
rs916946261 Chr.9: 127803364-127803364 on Build GRCh38
rs910285317 Chr.9: 127803263-127803263 on Build GRCh38
rs555586853 Chr.9: 127803398-127803398 on Build GRCh38
rs963584482 Chr.9: 127803226-127803226 on Build GRCh38
rs547702322 Chr.9: 127803441-127803441 on Build GRCh38
rs537879017 Chr.9: 127803443-127803443 on Build GRCh38
rs1005056804 Chr.9: 127803377-127803377 on Build GRCh38
rs1016534186 Chr.9: 127803213-127803213 on Build GRCh38
rs1029030365 Chr.9: 127803249-127803249 on Build GRCh38
rs540121654 Chr.9: 127803186-127803186 on Build GRCh38
rs1034866368 Chr.9: 127803396-127803396 on Build GRCh38
rs1014509744 Chr.9: 127803430-127803430 on Build GRCh38
rs954745319 Chr.9: 127803482-127803482 on Build GRCh38
rs937291105 Chr.9: 127803325-127803325 on Build GRCh38
rs985793138 Chr.9: 127803499-127803499 on Build GRCh38
rs557716733 Chr.9: 127803484-127803484 on Build GRCh38
rs1005532775 Chr.9: 127803360-127803360 on Build GRCh38
rs888475874 Chr.9: 127803339-127803339 on Build GRCh38
rs1027434816 Chr.9: 127803214-127803214 on Build GRCh38
rs987590453 Chr.9: 127803281-127803281 on Build GRCh38
rs547212842 Chr.9: 127803273-127803273 on Build GRCh38
rs535996340 Chr.9: 127803343-127803343 on Build GRCh38
rs998808320 Chr.9: 127803460-127803460 on Build GRCh38
rs1047324936 Chr.9: 127803554-127803554 on Build GRCh38
rs1047294223 Chr.9: 127803320-127803320 on Build GRCh38
rs930190002 Chr.9: 127803319-127803319 on Build GRCh38
rs886825904 Chr.9: 127803373-127803373 on Build GRCh38
rs568597807 Chr.9: 127803550-127803550 on Build GRCh38
rs1025799335 Chr.9: 127803184-127803184 on Build GRCh38
rs115992881 Chr.9: 127803168-127803168 on Build GRCh38

View Full Product Details