Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127815711-127816203 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENG
Entrez Gene ID
2022
Gene Name
endoglin
Gene Aliases
END, HHT1, ORW1
UniGene
Hs.76753
Gene Chromosome Location
Chr.9: 127815012-127854773 on Build GRCh38
Transcripts
NM_000118.3 NM_001114753.2 NM_001278138.1 NR_136302.1

Gene Symbol
LOC102723566
Entrez Gene ID
102723566
Gene Name
uncharacterized LOC102723566
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.9: 127816066-127822520 on Build GRCh38
Transcripts
NM_000118.3 NM_001114753.2 NM_001278138.1 NR_136302.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5467605 c.1790T>C p.F597S Chr.9: 127816005-127816005 on Build GRCh38
6306459 c.1863C>A p.P621P Chr.9: 127815932-127815932 on Build GRCh38
6370313 c.1780G>T p.G594C Chr.9: 127816015-127816015 on Build GRCh38
5472663 c.1845G>A p.S615S Chr.9: 127815950-127815950 on Build GRCh38
5467606 c.1790T>C p.F597S Chr.9: 127816005-127816005 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776826819 Chr.9: 127816040-127816040 on Build GRCh38
rs779115954 Chr.9: 127815909-127815909 on Build GRCh38
rs756018069 Chr.9: 127815900-127815900 on Build GRCh38
rs747196026 Chr.9: 127816036-127816036 on Build GRCh38
rs762716634 Chr.9: 127815775-127815775 on Build GRCh38
rs908692674 Chr.9: 127815824-127815824 on Build GRCh38
rs748191815 Chr.9: 127816029-127816029 on Build GRCh38
rs777352500 Chr.9: 127815764-127815764 on Build GRCh38
rs144922497 Chr.9: 127815770-127815771 on Build GRCh38
rs891630748 Chr.9: 127816118-127816118 on Build GRCh38
rs371104611 Chr.9: 127815891-127815891 on Build GRCh38
rs780018293 Chr.9: 127815799-127815799 on Build GRCh38
rs148002300 Chr.9: 127815951-127815951 on Build GRCh38
rs766073909 Chr.9: 127816022-127816022 on Build GRCh38
rs758620464 Chr.9: 127816084-127816084 on Build GRCh38
rs201678228 Chr.9: 127816068-127816068 on Build GRCh38
rs373611252 Chr.9: 127816066-127816066 on Build GRCh38
rs1023237192 Chr.9: 127815818-127815818 on Build GRCh38
rs200080694 Chr.9: 127816024-127816024 on Build GRCh38
rs779974705 Chr.9: 127815901-127815901 on Build GRCh38
rs948688834 Chr.9: 127815858-127815858 on Build GRCh38
rs750181050 Chr.9: 127815898-127815898 on Build GRCh38
rs868296721 Chr.9: 127816014-127816014 on Build GRCh38
rs755207132 Chr.9: 127815808-127815809 on Build GRCh38
rs562814353 Chr.9: 127815896-127815896 on Build GRCh38
rs143524008 Chr.9: 127815923-127815923 on Build GRCh38
rs774779781 Chr.9: 127815832-127815832 on Build GRCh38
rs766863961 Chr.9: 127815871-127815871 on Build GRCh38
rs41358947 Chr.9: 127816001-127816001 on Build GRCh38
rs373296026 Chr.9: 127815818-127815823 on Build GRCh38
rs367755236 Chr.9: 127816087-127816087 on Build GRCh38
rs376579767 Chr.9: 127815833-127815833 on Build GRCh38
rs370943570 Chr.9: 127816025-127816025 on Build GRCh38
rs868372047 Chr.9: 127816156-127816156 on Build GRCh38
rs1021923141 Chr.9: 127815935-127815935 on Build GRCh38
rs756003321 Chr.9: 127816021-127816021 on Build GRCh38
rs374591551 Chr.9: 127816045-127816045 on Build GRCh38
rs900458034 Chr.9: 127816133-127816133 on Build GRCh38
rs745843047 Chr.9: 127815783-127815783 on Build GRCh38
rs767008006 Chr.9: 127816065-127816065 on Build GRCh38
rs10760503 Chr.9: 127816125-127816125 on Build GRCh38
rs1040594389 Chr.9: 127815759-127815759 on Build GRCh38
rs529406638 Chr.9: 127815878-127815878 on Build GRCh38
rs201768056 Chr.9: 127816033-127816033 on Build GRCh38
rs147188969 Chr.9: 127815922-127815922 on Build GRCh38
rs964710604 Chr.9: 127815789-127815789 on Build GRCh38
rs938795970 Chr.9: 127816088-127816088 on Build GRCh38
rs780940714 Chr.9: 127815778-127815778 on Build GRCh38
rs186315094 Chr.9: 127815791-127815791 on Build GRCh38
rs1011793561 Chr.9: 127816121-127816121 on Build GRCh38
rs761244549 Chr.9: 127816063-127816063 on Build GRCh38
rs760221843 Chr.9: 127816067-127816067 on Build GRCh38
rs1053103287 Chr.9: 127816095-127816095 on Build GRCh38
rs773861569 Chr.9: 127816050-127816050 on Build GRCh38
rs746414124 Chr.9: 127816044-127816044 on Build GRCh38
rs750187963 Chr.9: 127816002-127816002 on Build GRCh38
rs753864390 Chr.9: 127816023-127816023 on Build GRCh38
rs754989823 Chr.9: 127816072-127816072 on Build GRCh38
rs778898313 Chr.9: 127815809-127815814 on Build GRCh38
rs772588778 Chr.9: 127816086-127816087 on Build GRCh38
rs368472273 Chr.9: 127815815-127815815 on Build GRCh38
rs753536186 Chr.9: 127816069-127816069 on Build GRCh38
rs773680353 Chr.9: 127815847-127815847 on Build GRCh38
rs981674869 Chr.9: 127815821-127815821 on Build GRCh38
rs758012489 Chr.9: 127815755-127815755 on Build GRCh38
rs866067907 Chr.9: 127815745-127815745 on Build GRCh38
rs933049719 Chr.9: 127815819-127815819 on Build GRCh38
rs1031656434 Chr.9: 127815804-127815804 on Build GRCh38
rs747308116 Chr.9: 127816091-127816091 on Build GRCh38
rs768836999 Chr.9: 127815906-127815906 on Build GRCh38
rs150566847 Chr.9: 127816037-127816037 on Build GRCh38
rs765208947 Chr.9: 127816078-127816078 on Build GRCh38
rs749176478 Chr.9: 127815905-127815905 on Build GRCh38
rs527434253 Chr.9: 127815831-127815831 on Build GRCh38
rs764694185 Chr.9: 127815736-127815736 on Build GRCh38
rs898513736 Chr.9: 127815856-127815856 on Build GRCh38
rs922983960 Chr.9: 127815795-127815795 on Build GRCh38
rs796980915 Chr.9: 127815793-127815793 on Build GRCh38
rs1030031854 Chr.9: 127816170-127816170 on Build GRCh38
rs772616157 Chr.9: 127815842-127815842 on Build GRCh38
rs201031046 Chr.9: 127815945-127815945 on Build GRCh38
rs761118628 Chr.9: 127815855-127815855 on Build GRCh38
rs748422950 Chr.9: 127815913-127815913 on Build GRCh38
rs996089843 Chr.9: 127816165-127816165 on Build GRCh38
rs748184867 Chr.9: 127815886-127815887 on Build GRCh38
rs752212042 Chr.9: 127815754-127815754 on Build GRCh38
rs796181085 Chr.9: 127816124-127816125 on Build GRCh38
rs778194854 Chr.9: 127816094-127816095 on Build GRCh38
rs546872552 Chr.9: 127816034-127816034 on Build GRCh38
rs577900032 Chr.9: 127815820-127815820 on Build GRCh38
rs759950184 Chr.9: 127815950-127815950 on Build GRCh38
rs540464910 Chr.9: 127815829-127815829 on Build GRCh38
rs747344646 Chr.9: 127815944-127815944 on Build GRCh38
rs34792295 Chr.9: 127815899-127815900 on Build GRCh38
rs919597400 Chr.9: 127816168-127816168 on Build GRCh38
rs757113987 Chr.9: 127815776-127815776 on Build GRCh38
rs373002544 Chr.9: 127815989-127815989 on Build GRCh38
rs752832223 Chr.9: 127816081-127816081 on Build GRCh38
rs373626498 Chr.9: 127815892-127815892 on Build GRCh38
rs972603389 Chr.9: 127816166-127816166 on Build GRCh38
rs761224492 Chr.9: 127815729-127815729 on Build GRCh38
rs891615717 Chr.9: 127815910-127815910 on Build GRCh38

View Full Product Details