Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127818435-127818931 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENG
Entrez Gene ID
2022
Gene Name
endoglin
Gene Aliases
END, HHT1, ORW1
UniGene
Hs.76753
Gene Chromosome Location
Chr.9: 127815012-127854773 on Build GRCh38
Transcripts
NM_000118.3 NM_001114753.2 NM_001278138.1 NR_136302.1

Gene Symbol
LOC102723566
Entrez Gene ID
102723566
Gene Name
uncharacterized LOC102723566
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.9: 127816066-127822520 on Build GRCh38
Transcripts
NM_000118.3 NM_001114753.2 NM_001278138.1 NR_136302.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6654023 c.1401C>T p.I467I Chr.9: 127818743-127818743 on Build GRCh38
4871437 c.1316A>G p.K439R Chr.9: 127818828-127818828 on Build GRCh38
1105418 c.1316A>G p.K439R Chr.9: 127818828-127818828 on Build GRCh38
4370652 c.1406C>T p.P469L Chr.9: 127818738-127818738 on Build GRCh38
607594 c.1406C>T p.P469L Chr.9: 127818738-127818738 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373269099 Chr.9: 127818877-127818877 on Build GRCh38
rs369020181 Chr.9: 127818851-127818851 on Build GRCh38
rs751310119 Chr.9: 127818684-127818684 on Build GRCh38
rs750412250 Chr.9: 127818830-127818830 on Build GRCh38
rs371513936 Chr.9: 127818690-127818690 on Build GRCh38
rs938107592 Chr.9: 127818588-127818588 on Build GRCh38
rs909202597 Chr.9: 127818587-127818587 on Build GRCh38
rs761685402 Chr.9: 127818589-127818589 on Build GRCh38
rs770902350 Chr.9: 127818845-127818845 on Build GRCh38
rs756263325 Chr.9: 127818736-127818736 on Build GRCh38
rs543098629 Chr.9: 127818609-127818609 on Build GRCh38
rs540809398 Chr.9: 127818466-127818466 on Build GRCh38
rs185549924 Chr.9: 127818661-127818661 on Build GRCh38
rs368250530 Chr.9: 127818791-127818791 on Build GRCh38
rs377760721 Chr.9: 127818872-127818872 on Build GRCh38
rs201684408 Chr.9: 127818844-127818844 on Build GRCh38
rs896325997 Chr.9: 127818603-127818603 on Build GRCh38
rs41302657 Chr.9: 127818737-127818737 on Build GRCh38
rs113436710 Chr.9: 127818639-127818639 on Build GRCh38
rs564304750 Chr.9: 127818498-127818498 on Build GRCh38
rs560856247 Chr.9: 127818473-127818473 on Build GRCh38
rs1049550659 Chr.9: 127818568-127818568 on Build GRCh38
rs541603726 Chr.9: 127818660-127818660 on Build GRCh38
rs34828244 Chr.9: 127818770-127818770 on Build GRCh38
rs863223542 Chr.9: 127818715-127818715 on Build GRCh38
rs375171647 Chr.9: 127818817-127818817 on Build GRCh38
rs764135740 Chr.9: 127818706-127818707 on Build GRCh38
rs950421970 Chr.9: 127818571-127818571 on Build GRCh38
rs763911667 Chr.9: 127818679-127818679 on Build GRCh38
rs965119099 Chr.9: 127818718-127818718 on Build GRCh38
rs761832486 Chr.9: 127818884-127818884 on Build GRCh38
rs377432993 Chr.9: 127818561-127818561 on Build GRCh38
rs983520490 Chr.9: 127818583-127818583 on Build GRCh38
rs1006597317 Chr.9: 127818887-127818887 on Build GRCh38
rs774501582 Chr.9: 127818840-127818841 on Build GRCh38
rs112927907 Chr.9: 127818521-127818521 on Build GRCh38
rs367753784 Chr.9: 127818738-127818738 on Build GRCh38
rs528194730 Chr.9: 127818649-127818649 on Build GRCh38
rs200168633 Chr.9: 127818755-127818755 on Build GRCh38
rs755962839 Chr.9: 127818831-127818831 on Build GRCh38
rs774943840 Chr.9: 127818779-127818779 on Build GRCh38
rs773419231 Chr.9: 127818881-127818881 on Build GRCh38
rs11545662 Chr.9: 127818739-127818739 on Build GRCh38
rs761065794 Chr.9: 127818756-127818756 on Build GRCh38
rs540556638 Chr.9: 127818610-127818610 on Build GRCh38
rs370394954 Chr.9: 127818691-127818691 on Build GRCh38
rs1019290949 Chr.9: 127818903-127818903 on Build GRCh38
rs751203937 Chr.9: 127818821-127818821 on Build GRCh38
rs991998832 Chr.9: 127818502-127818502 on Build GRCh38
rs891374531 Chr.9: 127818681-127818681 on Build GRCh38
rs1055144238 Chr.9: 127818839-127818839 on Build GRCh38
rs757129437 Chr.9: 127818827-127818827 on Build GRCh38
rs921410968 Chr.9: 127818638-127818638 on Build GRCh38
rs1024604850 Chr.9: 127818565-127818573 on Build GRCh38
rs900947822 Chr.9: 127818906-127818906 on Build GRCh38
rs1042762334 Chr.9: 127818696-127818696 on Build GRCh38
rs748457491 Chr.9: 127818754-127818754 on Build GRCh38
rs370554511 Chr.9: 127818742-127818742 on Build GRCh38
rs953670245 Chr.9: 127818666-127818666 on Build GRCh38
rs372540985 Chr.9: 127818807-127818807 on Build GRCh38
rs774869479 Chr.9: 127818743-127818743 on Build GRCh38
rs1042636734 Chr.9: 127818849-127818849 on Build GRCh38
rs561818608 Chr.9: 127818725-127818725 on Build GRCh38
rs1013339091 Chr.9: 127818628-127818628 on Build GRCh38
rs941764619 Chr.9: 127818590-127818590 on Build GRCh38
rs529788586 Chr.9: 127818513-127818513 on Build GRCh38
rs909233489 Chr.9: 127818806-127818806 on Build GRCh38
rs371576360 Chr.9: 127818869-127818869 on Build GRCh38
rs201640809 Chr.9: 127818870-127818870 on Build GRCh38
rs863223536 Chr.9: 127818832-127818832 on Build GRCh38
rs777564305 Chr.9: 127818809-127818809 on Build GRCh38
rs902467754 Chr.9: 127818647-127818647 on Build GRCh38
rs758017547 Chr.9: 127818669-127818669 on Build GRCh38
rs863223543 Chr.9: 127818714-127818714 on Build GRCh38
rs368533266 Chr.9: 127818828-127818828 on Build GRCh38

View Full Product Details