Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 128203135-128203639 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CIZ1
Entrez Gene ID
25792
Gene Name
CDKN1A interacting zinc finger protein 1
Gene Aliases
LSFR1, NP94, ZNF356
UniGene
Hs.212395
Gene Chromosome Location
Chr.9: 128166065-128204383 on Build GRCh38
Transcripts
NM_001131015.1 NM_012127.2 NM_001005336.2 NM_001288737.1 NM_001288738.1 NM_001288739.1 NM_004408.3 XM_005251763.2 XM_005251768.2 XM_005251769.2 XM_006716992.2 XM_006716993.1 XM_011518335.2 XM_011518336.2 XM_017014369.1 XM_017014370.1 XM_017014371.1 XM_017014372.1 XM_017014373.1

Gene Symbol
DNM1
Entrez Gene ID
1759
Gene Name
dynamin 1
Gene Aliases
DNM, EIEE31
UniGene
Hs.522413
Gene Chromosome Location
Chr.9: 128203355-128255248 on Build GRCh38
Transcripts
NM_001131015.1 NM_012127.2 NM_001005336.2 NM_001288737.1 NM_001288738.1 NM_001288739.1 NM_004408.3 XM_005251763.2 XM_005251768.2 XM_005251769.2 XM_006716992.2 XM_006716993.1 XM_011518335.2 XM_011518336.2 XM_017014369.1 XM_017014370.1 XM_017014371.1 XM_017014372.1 XM_017014373.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3654558 c.70G>A p.G24S Chr.9: 128203540-128203540 on Build GRCh38
7093427 c.148A>C p.N50H Chr.9: 128203618-128203618 on Build GRCh38
3664118 c.122G>A p.S41N Chr.9: 128203592-128203592 on Build GRCh38
3664117 c.122G>A p.S41N Chr.9: 128203592-128203592 on Build GRCh38
7093425 c.148A>C p.N50H Chr.9: 128203618-128203618 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368835504 Chr.9: 128203454-128203454 on Build GRCh38
rs895671345 Chr.9: 128203377-128203377 on Build GRCh38
rs750933399 Chr.9: 128203555-128203555 on Build GRCh38
rs1020000605 Chr.9: 128203409-128203409 on Build GRCh38
rs143374823 Chr.9: 128203173-128203173 on Build GRCh38
rs897203537 Chr.9: 128203224-128203224 on Build GRCh38
rs897457074 Chr.9: 128203397-128203397 on Build GRCh38
rs1045283033 Chr.9: 128203389-128203389 on Build GRCh38
rs994400329 Chr.9: 128203256-128203256 on Build GRCh38
rs752123322 Chr.9: 128203568-128203568 on Build GRCh38
rs1009986649 Chr.9: 128203384-128203384 on Build GRCh38
rs746765316 Chr.9: 128203450-128203450 on Build GRCh38
rs869312702 Chr.9: 128203609-128203609 on Build GRCh38
rs754529122 Chr.9: 128203424-128203424 on Build GRCh38
rs1036048662 Chr.9: 128203397-128203398 on Build GRCh38
rs778543086 Chr.9: 128203559-128203559 on Build GRCh38
rs756572497 Chr.9: 128203558-128203558 on Build GRCh38
rs928532911 Chr.9: 128203458-128203458 on Build GRCh38
rs972860852 Chr.9: 128203578-128203578 on Build GRCh38
rs752254892 Chr.9: 128203437-128203438 on Build GRCh38
rs944188764 Chr.9: 128203557-128203557 on Build GRCh38
rs373766853 Chr.9: 128203444-128203444 on Build GRCh38
rs1048915399 Chr.9: 128203271-128203271 on Build GRCh38
rs142432944 Chr.9: 128203587-128203587 on Build GRCh38
rs770857641 Chr.9: 128203594-128203594 on Build GRCh38
rs4240427 Chr.9: 128203260-128203260 on Build GRCh38
rs778217684 Chr.9: 128203436-128203436 on Build GRCh38
rs1018830112 Chr.9: 128203423-128203423 on Build GRCh38
rs994495555 Chr.9: 128203403-128203403 on Build GRCh38
rs923919360 Chr.9: 128203325-128203325 on Build GRCh38
rs763288862 Chr.9: 128203514-128203514 on Build GRCh38
rs762189357 Chr.9: 128203552-128203552 on Build GRCh38
rs1035545174 Chr.9: 128203372-128203372 on Build GRCh38
rs989324181 Chr.9: 128203370-128203370 on Build GRCh38
rs781115362 Chr.9: 128203213-128203213 on Build GRCh38
rs769986411 Chr.9: 128203491-128203491 on Build GRCh38
rs781131754 Chr.9: 128203608-128203608 on Build GRCh38
rs376941388 Chr.9: 128203456-128203456 on Build GRCh38
rs758104991 Chr.9: 128203575-128203575 on Build GRCh38
rs1037134060 Chr.9: 128203203-128203203 on Build GRCh38
rs554692756 Chr.9: 128203428-128203428 on Build GRCh38
rs906272571 Chr.9: 128203238-128203238 on Build GRCh38
rs61757224 Chr.9: 128203516-128203516 on Build GRCh38
rs947930594 Chr.9: 128203388-128203388 on Build GRCh38
rs577437504 Chr.9: 128203554-128203554 on Build GRCh38
rs969698049 Chr.9: 128203508-128203508 on Build GRCh38
rs747901647 Chr.9: 128203437-128203437 on Build GRCh38
rs552269083 Chr.9: 128203174-128203174 on Build GRCh38
rs1007085928 Chr.9: 128203419-128203419 on Build GRCh38
rs915151618 Chr.9: 128203379-128203379 on Build GRCh38
rs771609280 Chr.9: 128203439-128203439 on Build GRCh38
rs1001888605 Chr.9: 128203367-128203367 on Build GRCh38
rs905848137 Chr.9: 128203301-128203301 on Build GRCh38
rs956624769 Chr.9: 128203359-128203359 on Build GRCh38
rs751741699 Chr.9: 128203518-128203518 on Build GRCh38
rs1014993606 Chr.9: 128203241-128203241 on Build GRCh38
rs745880488 Chr.9: 128203614-128203614 on Build GRCh38
rs541580859 Chr.9: 128203307-128203307 on Build GRCh38
rs534786408 Chr.9: 128203390-128203390 on Build GRCh38
rs1003233822 Chr.9: 128203240-128203240 on Build GRCh38
rs1048713864 Chr.9: 128203407-128203407 on Build GRCh38
rs769135520 Chr.9: 128203270-128203270 on Build GRCh38
rs775423550 Chr.9: 128203502-128203502 on Build GRCh38
rs1018381360 Chr.9: 128203298-128203298 on Build GRCh38
rs976791968 Chr.9: 128203323-128203323 on Build GRCh38
rs897427245 Chr.9: 128203254-128203254 on Build GRCh38
rs998638437 Chr.9: 128203433-128203433 on Build GRCh38
rs986129698 Chr.9: 128203480-128203480 on Build GRCh38
rs572264214 Chr.9: 128203294-128203294 on Build GRCh38
rs939072110 Chr.9: 128203392-128203392 on Build GRCh38
rs759483040 Chr.9: 128203483-128203483 on Build GRCh38
rs888680935 Chr.9: 128203410-128203410 on Build GRCh38

View Full Product Details