Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 49611800-49612302 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHAT
Entrez Gene ID
1103
Gene Name
choline O-acetyltransferase
Gene Aliases
CHOACTASE, CMS1A, CMS1A2, CMS6
UniGene
Hs.302002
Gene Chromosome Location
Chr.10: 49609095-49665104 on Build GRCh38
Transcripts
NM_020984.3 NM_003055.2

Gene Symbol
SLC18A3
Entrez Gene ID
6572
Gene Name
solute carrier family 18 member A3
Gene Aliases
VACHT
UniGene
Hs.654374
Gene Chromosome Location
Chr.10: 49610301-49612720 on Build GRCh38
Transcripts
NM_020984.3 NM_003055.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
684124 c.1517G>T p.G506V Chr.10: 49612257-49612257 on Build GRCh38
1675226 c.1196C>T p.T399I Chr.10: 49611936-49611936 on Build GRCh38
5550030 c.1488G>T p.A496A Chr.10: 49612228-49612228 on Build GRCh38
294818 c.1504C>T p.R502C Chr.10: 49612244-49612244 on Build GRCh38
684126 c.1393G>T p.V465F Chr.10: 49612133-49612133 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200368763 Chr.10: 49612240-49612240 on Build GRCh38
rs761075493 Chr.10: 49612265-49612265 on Build GRCh38
rs753705874 Chr.10: 49612112-49612112 on Build GRCh38
rs771643697 Chr.10: 49611921-49611921 on Build GRCh38
rs762952215 Chr.10: 49612049-49612049 on Build GRCh38
rs755315988 Chr.10: 49612147-49612147 on Build GRCh38
rs989233173 Chr.10: 49612032-49612032 on Build GRCh38
rs1057517666 Chr.10: 49611932-49611932 on Build GRCh38
rs774777969 Chr.10: 49612060-49612060 on Build GRCh38
rs762834651 Chr.10: 49612224-49612225 on Build GRCh38
rs368027068 Chr.10: 49612228-49612228 on Build GRCh38
rs920596924 Chr.10: 49612209-49612209 on Build GRCh38
rs757904152 Chr.10: 49612132-49612132 on Build GRCh38
rs761525631 Chr.10: 49612188-49612188 on Build GRCh38
rs764029191 Chr.10: 49612081-49612081 on Build GRCh38
rs757770562 Chr.10: 49612281-49612281 on Build GRCh38
rs765313360 Chr.10: 49612174-49612174 on Build GRCh38
rs149604765 Chr.10: 49611866-49611866 on Build GRCh38
rs760513434 Chr.10: 49612072-49612072 on Build GRCh38
rs776627836 Chr.10: 49611849-49611849 on Build GRCh38
rs748705960 Chr.10: 49612042-49612042 on Build GRCh38
rs769077516 Chr.10: 49612039-49612039 on Build GRCh38
rs754259907 Chr.10: 49612274-49612274 on Build GRCh38
rs747379102 Chr.10: 49611933-49611933 on Build GRCh38
rs974690319 Chr.10: 49612201-49612201 on Build GRCh38
rs771167847 Chr.10: 49612052-49612052 on Build GRCh38
rs745998892 Chr.10: 49611827-49611827 on Build GRCh38
rs761770195 Chr.10: 49612116-49612116 on Build GRCh38
rs747360682 Chr.10: 49612036-49612036 on Build GRCh38
rs759063646 Chr.10: 49612263-49612263 on Build GRCh38
rs748546777 Chr.10: 49612149-49612149 on Build GRCh38
rs139485937 Chr.10: 49612271-49612271 on Build GRCh38
rs780425174 Chr.10: 49611924-49611924 on Build GRCh38
rs760601345 Chr.10: 49611991-49611991 on Build GRCh38
rs762753479 Chr.10: 49612245-49612245 on Build GRCh38
rs555224844 Chr.10: 49611974-49611974 on Build GRCh38
rs886609390 Chr.10: 49612267-49612267 on Build GRCh38
rs35838405 Chr.10: 49611865-49611865 on Build GRCh38
rs775908558 Chr.10: 49612018-49612020 on Build GRCh38
rs559399839 Chr.10: 49612010-49612010 on Build GRCh38
rs749377684 Chr.10: 49612167-49612167 on Build GRCh38
rs545078017 Chr.10: 49612135-49612135 on Build GRCh38
rs761119823 Chr.10: 49611824-49611824 on Build GRCh38
rs558868462 Chr.10: 49612185-49612185 on Build GRCh38
rs756580245 Chr.10: 49612152-49612152 on Build GRCh38
rs778197467 Chr.10: 49612154-49612154 on Build GRCh38
rs766544544 Chr.10: 49611975-49611975 on Build GRCh38
rs769518248 Chr.10: 49612244-49612244 on Build GRCh38
rs142761243 Chr.10: 49612191-49612191 on Build GRCh38
rs557909255 Chr.10: 49611895-49611895 on Build GRCh38
rs753381495 Chr.10: 49611889-49611889 on Build GRCh38
rs751093639 Chr.10: 49612195-49612195 on Build GRCh38
rs563318974 Chr.10: 49612176-49612176 on Build GRCh38
rs867798275 Chr.10: 49612026-49612026 on Build GRCh38
rs767148999 Chr.10: 49612264-49612264 on Build GRCh38
rs770889056 Chr.10: 49612246-49612246 on Build GRCh38
rs773305790 Chr.10: 49612046-49612046 on Build GRCh38
rs765432284 Chr.10: 49611860-49611860 on Build GRCh38
rs753088885 Chr.10: 49612199-49612199 on Build GRCh38
rs374465426 Chr.10: 49611988-49611988 on Build GRCh38
rs950290892 Chr.10: 49612272-49612272 on Build GRCh38
rs777864075 Chr.10: 49611883-49611883 on Build GRCh38
rs371081822 Chr.10: 49611955-49611955 on Build GRCh38
rs765089404 Chr.10: 49612125-49612125 on Build GRCh38
rs1045968094 Chr.10: 49612282-49612282 on Build GRCh38
rs769881006 Chr.10: 49611899-49611899 on Build GRCh38
rs761894527 Chr.10: 49611856-49611856 on Build GRCh38
rs749891211 Chr.10: 49612128-49612128 on Build GRCh38
rs761878159 Chr.10: 49611947-49611947 on Build GRCh38
rs554262479 Chr.10: 49611821-49611821 on Build GRCh38
rs1020790928 Chr.10: 49612065-49612065 on Build GRCh38
rs866122299 Chr.10: 49611968-49611968 on Build GRCh38
rs777182420 Chr.10: 49611936-49611936 on Build GRCh38
rs757804116 Chr.10: 49612224-49612224 on Build GRCh38
rs899224700 Chr.10: 49611862-49611862 on Build GRCh38
rs778310071 Chr.10: 49612202-49612202 on Build GRCh38
rs772362067 Chr.10: 49612232-49612232 on Build GRCh38
rs118107581 Chr.10: 49612016-49612016 on Build GRCh38
rs745582440 Chr.10: 49611906-49611906 on Build GRCh38
rs943388046 Chr.10: 49611823-49611823 on Build GRCh38
rs147348659 Chr.10: 49612227-49612227 on Build GRCh38
rs138708428 Chr.10: 49612184-49612184 on Build GRCh38
rs764121092 Chr.10: 49611993-49611993 on Build GRCh38
rs376743590 Chr.10: 49612193-49612193 on Build GRCh38
rs773286296 Chr.10: 49611957-49611957 on Build GRCh38
rs192440196 Chr.10: 49611934-49611934 on Build GRCh38
rs144340824 Chr.10: 49612181-49612181 on Build GRCh38
rs36060462 Chr.10: 49611972-49611972 on Build GRCh38
rs150494839 Chr.10: 49611832-49611832 on Build GRCh38
rs61846640 Chr.10: 49612250-49612250 on Build GRCh38
rs79468925 Chr.10: 49611830-49611830 on Build GRCh38
rs371922639 Chr.10: 49612200-49612200 on Build GRCh38
rs753863574 Chr.10: 49612005-49612005 on Build GRCh38
rs772526674 Chr.10: 49611820-49611820 on Build GRCh38
rs778397784 Chr.10: 49611904-49611904 on Build GRCh38
rs1047297359 Chr.10: 49611897-49611897 on Build GRCh38
rs767165278 Chr.10: 49612197-49612197 on Build GRCh38
rs765208277 Chr.10: 49612189-49612189 on Build GRCh38
rs1009325288 Chr.10: 49612148-49612148 on Build GRCh38
rs376281494 Chr.10: 49612219-49612219 on Build GRCh38
rs547637544 Chr.10: 49612233-49612233 on Build GRCh38
rs202245540 Chr.10: 49612009-49612009 on Build GRCh38
rs987386343 Chr.10: 49612180-49612180 on Build GRCh38

View Full Product Details