Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 12138149-12138548 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MICAL2
Entrez Gene ID
9645
Gene Name
microtubule associated monooxygenase, calponin and LIM domain containing 2
Gene Aliases
MICAL-2, MICAL2PV1, MICAL2PV2
UniGene
Hs.501928
Gene Chromosome Location
Chr.11: 12110576-12281017 on Build GRCh38
Transcripts
NM_001282663.1 NM_001282664.1 NM_001282665.1 NM_001282668.1 NM_014632.3 XM_017018589.1 XM_017018591.1

SNP Detail

SNP ID Chromosome Location
rs150181763 Chr.11: 12138357-12138357 on Build GRCh38
rs932512729 Chr.11: 12138404-12138404 on Build GRCh38
rs575480163 Chr.11: 12138345-12138345 on Build GRCh38
rs757464524 Chr.11: 12138283-12138283 on Build GRCh38
rs368911881 Chr.11: 12138276-12138276 on Build GRCh38
rs539998437 Chr.11: 12138401-12138401 on Build GRCh38
rs1008759 Chr.11: 12138319-12138319 on Build GRCh38
rs370026521 Chr.11: 12138329-12138329 on Build GRCh38
rs901284 Chr.11: 12138447-12138447 on Build GRCh38
rs919834840 Chr.11: 12138402-12138402 on Build GRCh38
rs1021795837 Chr.11: 12138216-12138216 on Build GRCh38
rs1994319 Chr.11: 12138306-12138306 on Build GRCh38
rs901285 Chr.11: 12138507-12138507 on Build GRCh38
rs756181011 Chr.11: 12138493-12138493 on Build GRCh38
rs553325096 Chr.11: 12138177-12138177 on Build GRCh38
rs892321651 Chr.11: 12138175-12138175 on Build GRCh38
rs190616251 Chr.11: 12138510-12138510 on Build GRCh38
rs992290911 Chr.11: 12138307-12138307 on Build GRCh38
rs371724878 Chr.11: 12138194-12138194 on Build GRCh38
rs577657845 Chr.11: 12138364-12138364 on Build GRCh38
rs890923622 Chr.11: 12138459-12138459 on Build GRCh38
rs939645951 Chr.11: 12138346-12138346 on Build GRCh38
rs868595916 Chr.11: 12138509-12138509 on Build GRCh38
rs908222646 Chr.11: 12138321-12138321 on Build GRCh38
rs891801079 Chr.11: 12138511-12138511 on Build GRCh38
rs998864482 Chr.11: 12138294-12138294 on Build GRCh38
rs1011834026 Chr.11: 12138215-12138215 on Build GRCh38
rs544396247 Chr.11: 12138347-12138347 on Build GRCh38
rs762199816 Chr.11: 12138416-12138416 on Build GRCh38
rs970231583 Chr.11: 12138267-12138267 on Build GRCh38
rs5789715 Chr.11: 12138216-12138216 on Build GRCh38
rs774802887 Chr.11: 12138411-12138411 on Build GRCh38
rs4433551 Chr.11: 12138203-12138203 on Build GRCh38
rs773439371 Chr.11: 12138491-12138491 on Build GRCh38
rs866551478 Chr.11: 12138336-12138336 on Build GRCh38
rs566800761 Chr.11: 12138183-12138183 on Build GRCh38
rs767224301 Chr.11: 12138298-12138298 on Build GRCh38
rs1110007 Chr.11: 12138494-12138494 on Build GRCh38
rs762963250 Chr.11: 12138277-12138277 on Build GRCh38
rs926397243 Chr.11: 12138282-12138282 on Build GRCh38

View Full Product Details