Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 66844733-66845248 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RCE1
Entrez Gene ID
9986
Gene Name
Ras converting CAAX endopeptidase 1
Gene Aliases
FACE2, RCE1A, RCE1B
UniGene
Hs.654972
Gene Chromosome Location
Chr.11: 66843412-66846532 on Build GRCh38
Transcripts
NM_001032279.1 NM_005133.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5373352 c.559T>C p.L187L Chr.11: 66844976-66844976 on Build GRCh38
7288837 c.487C>T p.R163C Chr.11: 66844904-66844904 on Build GRCh38
5051070 c.511G>A p.A171T Chr.11: 66844928-66844928 on Build GRCh38
6259469 c.662G>T p.S221I Chr.11: 66845208-66845208 on Build GRCh38
7095751 c.594C>T p.F198F Chr.11: 66845011-66845011 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs994166554 Chr.11: 66844822-66844822 on Build GRCh38
rs75404994 Chr.11: 66844763-66844763 on Build GRCh38
rs748081348 Chr.11: 66844884-66844884 on Build GRCh38
rs749842586 Chr.11: 66844885-66844885 on Build GRCh38
rs371258472 Chr.11: 66845213-66845213 on Build GRCh38
rs952590843 Chr.11: 66845138-66845138 on Build GRCh38
rs766370063 Chr.11: 66844869-66844869 on Build GRCh38
rs1013006500 Chr.11: 66845114-66845114 on Build GRCh38
rs758898268 Chr.11: 66845223-66845225 on Build GRCh38
rs990398442 Chr.11: 66844907-66844907 on Build GRCh38
rs773493789 Chr.11: 66844913-66844913 on Build GRCh38
rs895729736 Chr.11: 66845092-66845092 on Build GRCh38
rs765958862 Chr.11: 66845088-66845088 on Build GRCh38
rs538803557 Chr.11: 66844809-66844809 on Build GRCh38
rs772883148 Chr.11: 66845127-66845128 on Build GRCh38
rs1029775700 Chr.11: 66844823-66844823 on Build GRCh38
rs77218191 Chr.11: 66845079-66845079 on Build GRCh38
rs377709636 Chr.11: 66844969-66844969 on Build GRCh38
rs778376909 Chr.11: 66845121-66845121 on Build GRCh38
rs867781167 Chr.11: 66844783-66844783 on Build GRCh38
rs761612259 Chr.11: 66844843-66844844 on Build GRCh38
rs766526501 Chr.11: 66844933-66844933 on Build GRCh38
rs760347693 Chr.11: 66845131-66845132 on Build GRCh38
rs374360120 Chr.11: 66844886-66844886 on Build GRCh38
rs972319329 Chr.11: 66844958-66844958 on Build GRCh38
rs750149860 Chr.11: 66844979-66844979 on Build GRCh38
rs766310184 Chr.11: 66845053-66845053 on Build GRCh38
rs959715472 Chr.11: 66844812-66844813 on Build GRCh38
rs752735030 Chr.11: 66844873-66844873 on Build GRCh38
rs748528358 Chr.11: 66844887-66844887 on Build GRCh38
rs748646561 Chr.11: 66845020-66845020 on Build GRCh38
rs1005273662 Chr.11: 66845144-66845144 on Build GRCh38
rs368155242 Chr.11: 66845161-66845161 on Build GRCh38
rs1021418389 Chr.11: 66844786-66844788 on Build GRCh38
rs1056048214 Chr.11: 66844794-66844794 on Build GRCh38
rs554533903 Chr.11: 66844957-66844957 on Build GRCh38
rs753377252 Chr.11: 66845138-66845139 on Build GRCh38
rs201147022 Chr.11: 66845078-66845078 on Build GRCh38
rs774550185 Chr.11: 66844834-66844834 on Build GRCh38
rs117419973 Chr.11: 66844795-66844795 on Build GRCh38
rs201760776 Chr.11: 66844820-66844820 on Build GRCh38
rs747299586 Chr.11: 66845043-66845043 on Build GRCh38
rs371545927 Chr.11: 66844878-66844878 on Build GRCh38
rs758875456 Chr.11: 66845134-66845134 on Build GRCh38
rs772360502 Chr.11: 66844912-66844912 on Build GRCh38
rs202245436 Chr.11: 66845054-66845054 on Build GRCh38
rs779715104 Chr.11: 66844989-66844989 on Build GRCh38
rs760735841 Chr.11: 66845011-66845011 on Build GRCh38
rs760751960 Chr.11: 66844864-66844864 on Build GRCh38
rs547769505 Chr.11: 66844754-66844754 on Build GRCh38
rs541626599 Chr.11: 66845000-66845000 on Build GRCh38
rs755690406 Chr.11: 66844982-66844982 on Build GRCh38
rs773548526 Chr.11: 66845023-66845023 on Build GRCh38
rs374365634 Chr.11: 66845162-66845162 on Build GRCh38
rs370831140 Chr.11: 66844847-66844847 on Build GRCh38
rs760389409 Chr.11: 66845195-66845195 on Build GRCh38
rs199945997 Chr.11: 66845014-66845014 on Build GRCh38
rs761820215 Chr.11: 66845072-66845072 on Build GRCh38
rs1001406266 Chr.11: 66844793-66844793 on Build GRCh38
rs1050413645 Chr.11: 66845013-66845013 on Build GRCh38
rs906946034 Chr.11: 66844789-66844789 on Build GRCh38
rs1011831322 Chr.11: 66844776-66844776 on Build GRCh38
rs753488799 Chr.11: 66845201-66845201 on Build GRCh38
rs556402458 Chr.11: 66845052-66845052 on Build GRCh38
rs947471267 Chr.11: 66845019-66845019 on Build GRCh38
rs747359278 Chr.11: 66845141-66845141 on Build GRCh38
rs367622666 Chr.11: 66845202-66845202 on Build GRCh38
rs772478407 Chr.11: 66845021-66845021 on Build GRCh38
rs772155989 Chr.11: 66845175-66845175 on Build GRCh38
rs752279789 Chr.11: 66845163-66845163 on Build GRCh38
rs545154470 Chr.11: 66845059-66845059 on Build GRCh38
rs776538770 Chr.11: 66844942-66844942 on Build GRCh38
rs200522940 Chr.11: 66844830-66844830 on Build GRCh38
rs377112256 Chr.11: 66844853-66844853 on Build GRCh38
rs61737209 Chr.11: 66844993-66844993 on Build GRCh38
rs928292671 Chr.11: 66844988-66844988 on Build GRCh38
rs769928994 Chr.11: 66845057-66845057 on Build GRCh38
rs763962438 Chr.11: 66844962-66844962 on Build GRCh38
rs536534375 Chr.11: 66844780-66844780 on Build GRCh38
rs773141242 Chr.11: 66845178-66845178 on Build GRCh38
rs985531370 Chr.11: 66844852-66844852 on Build GRCh38
rs778215383 Chr.11: 66845137-66845137 on Build GRCh38
rs767415499 Chr.11: 66844975-66844975 on Build GRCh38
rs1006554557 Chr.11: 66844808-66844808 on Build GRCh38
rs763987813 Chr.11: 66844874-66844874 on Build GRCh38
rs753821049 Chr.11: 66845063-66845063 on Build GRCh38
rs923507733 Chr.11: 66844983-66844983 on Build GRCh38
rs374055495 Chr.11: 66845122-66845122 on Build GRCh38
rs757122849 Chr.11: 66844972-66844972 on Build GRCh38
rs763085245 Chr.11: 66845165-66845165 on Build GRCh38
rs779410043 Chr.11: 66844879-66844879 on Build GRCh38
rs757067541 Chr.11: 66844875-66844875 on Build GRCh38
rs913484564 Chr.11: 66844935-66844935 on Build GRCh38
rs375531678 Chr.11: 66845224-66845224 on Build GRCh38
rs1006049383 Chr.11: 66845058-66845058 on Build GRCh38
rs774208661 Chr.11: 66844821-66844824 on Build GRCh38
rs1014816414 Chr.11: 66844815-66844815 on Build GRCh38
rs781004647 Chr.11: 66844876-66844876 on Build GRCh38
rs1030512897 Chr.11: 66845015-66845028 on Build GRCh38
rs569889090 Chr.11: 66844801-66844801 on Build GRCh38
rs778268309 Chr.11: 66845221-66845221 on Build GRCh38
rs771270478 Chr.11: 66845154-66845154 on Build GRCh38
rs760468168 Chr.11: 66845087-66845087 on Build GRCh38
rs368409311 Chr.11: 66844833-66844833 on Build GRCh38
rs534361272 Chr.11: 66844850-66844850 on Build GRCh38
rs757630151 Chr.11: 66845225-66845225 on Build GRCh38
rs753460902 Chr.11: 66845117-66845117 on Build GRCh38
rs754509961 Chr.11: 66845120-66845120 on Build GRCh38
rs767251347 Chr.11: 66845126-66845129 on Build GRCh38
rs773368830 Chr.11: 66844856-66844856 on Build GRCh38
rs762127880 Chr.11: 66844851-66844851 on Build GRCh38
rs766189288 Chr.11: 66845197-66845197 on Build GRCh38

View Full Product Details