Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 20704002-20704590 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL17D
Entrez Gene ID
53342
Gene Name
interleukin 17D
Gene Aliases
IL-17D
UniGene
Hs.655142
Gene Chromosome Location
Chr.13: 20702775-20723100 on Build GRCh38
Transcripts
NM_138284.1 XM_005266421.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5772697 c.130G>C p.G44R Chr.13: 20704131-20704131 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1050119649 Chr.13: 20704070-20704094 on Build GRCh38
rs111358847 Chr.13: 20704068-20704068 on Build GRCh38
rs1026242349 Chr.13: 20704298-20704298 on Build GRCh38
rs975594451 Chr.13: 20704517-20704517 on Build GRCh38
rs1020815978 Chr.13: 20704475-20704475 on Build GRCh38
rs1039936321 Chr.13: 20704135-20704135 on Build GRCh38
rs890042408 Chr.13: 20704405-20704405 on Build GRCh38
rs1016538343 Chr.13: 20704268-20704268 on Build GRCh38
rs752316602 Chr.13: 20704139-20704139 on Build GRCh38
rs918752957 Chr.13: 20704390-20704390 on Build GRCh38
rs1010300977 Chr.13: 20704470-20704470 on Build GRCh38
rs990424437 Chr.13: 20704557-20704557 on Build GRCh38
rs1024640615 Chr.13: 20704217-20704217 on Build GRCh38
rs942798284 Chr.13: 20704441-20704441 on Build GRCh38
rs199571028 Chr.13: 20704216-20704216 on Build GRCh38
rs1003334698 Chr.13: 20704511-20704511 on Build GRCh38
rs747547288 Chr.13: 20704274-20704274 on Build GRCh38
rs113588022 Chr.13: 20704181-20704181 on Build GRCh38
rs778497617 Chr.13: 20704081-20704081 on Build GRCh38
rs912651530 Chr.13: 20704366-20704366 on Build GRCh38
rs1006859597 Chr.13: 20704410-20704410 on Build GRCh38
rs866321335 Chr.13: 20704552-20704552 on Build GRCh38
rs918604752 Chr.13: 20704078-20704078 on Build GRCh38
rs1001984576 Chr.13: 20704255-20704255 on Build GRCh38
rs972366120 Chr.13: 20704282-20704282 on Build GRCh38
rs1054471150 Chr.13: 20704413-20704413 on Build GRCh38
rs1013212105 Chr.13: 20704197-20704197 on Build GRCh38
rs1037213486 Chr.13: 20704360-20704360 on Build GRCh38
rs909128552 Chr.13: 20704057-20704057 on Build GRCh38
rs909535876 Chr.13: 20704566-20704566 on Build GRCh38
rs893186087 Chr.13: 20704435-20704435 on Build GRCh38
rs757640577 Chr.13: 20704301-20704301 on Build GRCh38
rs1051154300 Chr.13: 20704083-20704083 on Build GRCh38
rs868771622 Chr.13: 20704528-20704528 on Build GRCh38
rs904990229 Chr.13: 20704239-20704239 on Build GRCh38
rs1051342759 Chr.13: 20704399-20704399 on Build GRCh38
rs912661329 Chr.13: 20704094-20704094 on Build GRCh38
rs986803319 Chr.13: 20704352-20704352 on Build GRCh38
rs951328698 Chr.13: 20704314-20704314 on Build GRCh38
rs963305025 Chr.13: 20704276-20704276 on Build GRCh38
rs892102767 Chr.13: 20704191-20704191 on Build GRCh38
rs1034865935 Chr.13: 20704515-20704515 on Build GRCh38
rs966560759 Chr.13: 20704480-20704480 on Build GRCh38
rs934189801 Chr.13: 20704394-20704394 on Build GRCh38
rs576539808 Chr.13: 20704113-20704113 on Build GRCh38
rs932955291 Chr.13: 20704060-20704078 on Build GRCh38
rs925467144 Chr.13: 20704519-20704519 on Build GRCh38
rs1001553182 Chr.13: 20704181-20704182 on Build GRCh38
rs904098138 Chr.13: 20704158-20704158 on Build GRCh38

View Full Product Details