Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 74885483-74885893 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDR59
Entrez Gene ID
79726
Gene Name
WD repeat domain 59
Gene Aliases
FP977
UniGene
Hs.280951
Gene Chromosome Location
Chr.16: 74873568-74985146 on Build GRCh38
Transcripts
NM_030581.3 XM_005256146.3 XM_011523331.2 XM_011523332.2 XM_017023667.1 XM_017023668.1 XM_017023669.1 XM_017023670.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5740738 c.2617T>C p.W873R Chr.16: 74885725-74885725 on Build GRCh38
6821969 c.2689G>T p.E897* Chr.16: 74885653-74885653 on Build GRCh38
4597030 c.2641G>C p.E881Q Chr.16: 74885701-74885701 on Build GRCh38
3370555 c.2647T>G p.L883V Chr.16: 74885695-74885695 on Build GRCh38
3672323 c.2619G>T p.W873C Chr.16: 74885723-74885723 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751986317 Chr.16: 74885724-74885724 on Build GRCh38
rs369583625 Chr.16: 74885604-74885604 on Build GRCh38
rs753769332 Chr.16: 74885759-74885759 on Build GRCh38
rs763404889 Chr.16: 74885717-74885717 on Build GRCh38
rs760183298 Chr.16: 74885766-74885767 on Build GRCh38
rs756003450 Chr.16: 74885659-74885659 on Build GRCh38
rs368220022 Chr.16: 74885799-74885799 on Build GRCh38
rs747162469 Chr.16: 74885695-74885695 on Build GRCh38
rs1038997267 Chr.16: 74885721-74885721 on Build GRCh38
rs201968575 Chr.16: 74885747-74885747 on Build GRCh38
rs764579176 Chr.16: 74885634-74885634 on Build GRCh38
rs371980149 Chr.16: 74885781-74885781 on Build GRCh38
rs781528626 Chr.16: 74885782-74885782 on Build GRCh38
rs151050615 Chr.16: 74885722-74885722 on Build GRCh38
rs143789413 Chr.16: 74885706-74885706 on Build GRCh38
rs778617980 Chr.16: 74885766-74885766 on Build GRCh38
rs760275233 Chr.16: 74885832-74885832 on Build GRCh38
rs758849829 Chr.16: 74885622-74885622 on Build GRCh38
rs749013113 Chr.16: 74885663-74885663 on Build GRCh38
rs187224283 Chr.16: 74885556-74885556 on Build GRCh38
rs374327975 Chr.16: 74885830-74885830 on Build GRCh38
rs1016737984 Chr.16: 74885859-74885859 on Build GRCh38
rs774161710 Chr.16: 74885692-74885692 on Build GRCh38
rs562579301 Chr.16: 74885791-74885791 on Build GRCh38
rs112870564 Chr.16: 74885783-74885783 on Build GRCh38
rs556287386 Chr.16: 74885598-74885598 on Build GRCh38
rs774853289 Chr.16: 74885811-74885811 on Build GRCh38
rs576313871 Chr.16: 74885637-74885637 on Build GRCh38
rs558741480 Chr.16: 74885846-74885846 on Build GRCh38
rs916741498 Chr.16: 74885519-74885519 on Build GRCh38
rs76563127 Chr.16: 74885589-74885589 on Build GRCh38
rs768390298 Chr.16: 74885667-74885667 on Build GRCh38
rs140995093 Chr.16: 74885728-74885728 on Build GRCh38
rs1033485790 Chr.16: 74885576-74885576 on Build GRCh38
rs769329278 Chr.16: 74885768-74885768 on Build GRCh38
rs750042296 Chr.16: 74885831-74885831 on Build GRCh38
rs753251733 Chr.16: 74885770-74885771 on Build GRCh38
rs147964482 Chr.16: 74885654-74885654 on Build GRCh38
rs71392681 Chr.16: 74885537-74885537 on Build GRCh38
rs188695478 Chr.16: 74885732-74885732 on Build GRCh38
rs367616908 Chr.16: 74885707-74885707 on Build GRCh38
rs576664778 Chr.16: 74885566-74885566 on Build GRCh38
rs762310713 Chr.16: 74885828-74885828 on Build GRCh38
rs369345264 Chr.16: 74885738-74885738 on Build GRCh38
rs754910144 Chr.16: 74885763-74885763 on Build GRCh38
rs191535208 Chr.16: 74885701-74885701 on Build GRCh38
rs957735344 Chr.16: 74885797-74885797 on Build GRCh38
rs759726048 Chr.16: 74885702-74885702 on Build GRCh38
rs775193441 Chr.16: 74885748-74885749 on Build GRCh38
rs747992526 Chr.16: 74885775-74885775 on Build GRCh38
rs1009340735 Chr.16: 74885733-74885733 on Build GRCh38
rs533717044 Chr.16: 74885551-74885551 on Build GRCh38
rs763660005 Chr.16: 74885769-74885771 on Build GRCh38
rs747563766 Chr.16: 74885745-74885745 on Build GRCh38
rs963391673 Chr.16: 74885868-74885868 on Build GRCh38
rs776019815 Chr.16: 74885714-74885714 on Build GRCh38
rs148977214 Chr.16: 74885696-74885696 on Build GRCh38
rs771136155 Chr.16: 74885778-74885778 on Build GRCh38
rs375900043 Chr.16: 74885653-74885653 on Build GRCh38
rs372896695 Chr.16: 74885646-74885646 on Build GRCh38
rs763459577 Chr.16: 74885792-74885792 on Build GRCh38
rs8052054 Chr.16: 74885807-74885807 on Build GRCh38
rs930084425 Chr.16: 74885608-74885608 on Build GRCh38
rs183855703 Chr.16: 74885727-74885727 on Build GRCh38

View Full Product Details