Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 125107029-125107521 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AACS
Entrez Gene ID
65985
Gene Name
acetoacetyl-CoA synthetase
Gene Aliases
ACSF1, SUR-5
UniGene
Hs.656073
Gene Chromosome Location
Chr.12: 125065367-125143333 on Build GRCh38
Transcripts
NM_001319839.1 NM_001319840.1 NM_023928.4 XM_011538692.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2227426 c.895T>C p.C299R Chr.12: 125107248-125107248 on Build GRCh38
936808 c.886G>A p.A296T Chr.12: 125107239-125107239 on Build GRCh38
1660327 c.913G>A p.G305R Chr.12: 125107266-125107266 on Build GRCh38
4825799 c.816G>A p.P272P Chr.12: 125107169-125107169 on Build GRCh38
3457880 c.851C>A p.P284Q Chr.12: 125107204-125107204 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1045479820 Chr.12: 125107411-125107411 on Build GRCh38
rs780791114 Chr.12: 125107191-125107191 on Build GRCh38
rs977841990 Chr.12: 125107377-125107377 on Build GRCh38
rs770615943 Chr.12: 125107302-125107303 on Build GRCh38
rs371228445 Chr.12: 125107494-125107494 on Build GRCh38
rs939810134 Chr.12: 125107428-125107428 on Build GRCh38
rs771451812 Chr.12: 125107072-125107072 on Build GRCh38
rs1015487513 Chr.12: 125107286-125107286 on Build GRCh38
rs944530221 Chr.12: 125107150-125107150 on Build GRCh38
rs551327816 Chr.12: 125107354-125107354 on Build GRCh38
rs889584083 Chr.12: 125107495-125107495 on Build GRCh38
rs953921534 Chr.12: 125107325-125107325 on Build GRCh38
rs536905147 Chr.12: 125107392-125107392 on Build GRCh38
rs761869895 Chr.12: 125107167-125107167 on Build GRCh38
rs774559316 Chr.12: 125107165-125107165 on Build GRCh38
rs77154370 Chr.12: 125107178-125107178 on Build GRCh38
rs769722755 Chr.12: 125107197-125107197 on Build GRCh38
rs775168357 Chr.12: 125107217-125107217 on Build GRCh38
rs780509032 Chr.12: 125107116-125107116 on Build GRCh38
rs117939534 Chr.12: 125107477-125107477 on Build GRCh38
rs768410774 Chr.12: 125107278-125107278 on Build GRCh38
rs2297478 Chr.12: 125107298-125107298 on Build GRCh38
rs777512525 Chr.12: 125107187-125107187 on Build GRCh38
rs936238141 Chr.12: 125107382-125107382 on Build GRCh38
rs919118483 Chr.12: 125107437-125107437 on Build GRCh38
rs139116253 Chr.12: 125107149-125107149 on Build GRCh38
rs201678229 Chr.12: 125107103-125107103 on Build GRCh38
rs202000490 Chr.12: 125107090-125107090 on Build GRCh38
rs201450673 Chr.12: 125107112-125107112 on Build GRCh38
rs1004300314 Chr.12: 125107269-125107269 on Build GRCh38
rs555241257 Chr.12: 125107420-125107420 on Build GRCh38
rs144530233 Chr.12: 125107226-125107226 on Build GRCh38
rs770977454 Chr.12: 125107310-125107310 on Build GRCh38
rs776717043 Chr.12: 125107087-125107087 on Build GRCh38
rs369677318 Chr.12: 125107168-125107168 on Build GRCh38
rs760288634 Chr.12: 125107085-125107085 on Build GRCh38
rs373315611 Chr.12: 125107094-125107094 on Build GRCh38
rs1037091312 Chr.12: 125107431-125107431 on Build GRCh38
rs975892611 Chr.12: 125107432-125107432 on Build GRCh38
rs373177690 Chr.12: 125107225-125107225 on Build GRCh38
rs747734924 Chr.12: 125107119-125107119 on Build GRCh38
rs766845698 Chr.12: 125107231-125107231 on Build GRCh38
rs776694452 Chr.12: 125107135-125107135 on Build GRCh38
rs79597395 Chr.12: 125107091-125107091 on Build GRCh38
rs754194640 Chr.12: 125107179-125107179 on Build GRCh38
rs774181764 Chr.12: 125107292-125107292 on Build GRCh38
rs781610796 Chr.12: 125107107-125107107 on Build GRCh38
rs939791943 Chr.12: 125107473-125107482 on Build GRCh38
rs770581705 Chr.12: 125107133-125107133 on Build GRCh38
rs746456550 Chr.12: 125107128-125107128 on Build GRCh38
rs573627976 Chr.12: 125107435-125107435 on Build GRCh38
rs995382898 Chr.12: 125107445-125107445 on Build GRCh38
rs906966085 Chr.12: 125107412-125107412 on Build GRCh38
rs138371145 Chr.12: 125107262-125107262 on Build GRCh38
rs779982775 Chr.12: 125107265-125107265 on Build GRCh38
rs747960058 Chr.12: 125107297-125107297 on Build GRCh38
rs924657302 Chr.12: 125107378-125107378 on Build GRCh38
rs777188135 Chr.12: 125107316-125107316 on Build GRCh38
rs916078902 Chr.12: 125107401-125107401 on Build GRCh38
rs766821572 Chr.12: 125107175-125107175 on Build GRCh38
rs755513494 Chr.12: 125107062-125107062 on Build GRCh38
rs371263015 Chr.12: 125107234-125107234 on Build GRCh38
rs146789000 Chr.12: 125107067-125107067 on Build GRCh38
rs771711554 Chr.12: 125107121-125107121 on Build GRCh38
rs41521044 Chr.12: 125107238-125107238 on Build GRCh38
rs757016707 Chr.12: 125107251-125107251 on Build GRCh38
rs745343464 Chr.12: 125107255-125107255 on Build GRCh38
rs995468117 Chr.12: 125107293-125107293 on Build GRCh38
rs774180700 Chr.12: 125107481-125107481 on Build GRCh38
rs911983995 Chr.12: 125107341-125107341 on Build GRCh38
rs1049043798 Chr.12: 125107182-125107182 on Build GRCh38
rs772910974 Chr.12: 125107080-125107080 on Build GRCh38
rs745765907 Chr.12: 125107196-125107196 on Build GRCh38
rs768451520 Chr.12: 125107211-125107211 on Build GRCh38
rs774553529 Chr.12: 125107212-125107212 on Build GRCh38
rs777604368 Chr.12: 125107122-125107122 on Build GRCh38
rs140472203 Chr.12: 125107188-125107188 on Build GRCh38
rs560864028 Chr.12: 125107444-125107444 on Build GRCh38
rs145595572 Chr.12: 125107250-125107250 on Build GRCh38
rs368086322 Chr.12: 125107069-125107069 on Build GRCh38
rs775319593 Chr.12: 125107200-125107200 on Build GRCh38
rs770955788 Chr.12: 125107086-125107086 on Build GRCh38
rs140471021 Chr.12: 125107181-125107181 on Build GRCh38
rs188567343 Chr.12: 125107252-125107252 on Build GRCh38
rs540917287 Chr.12: 125107095-125107095 on Build GRCh38
rs894247834 Chr.12: 125107049-125107049 on Build GRCh38
rs760753338 Chr.12: 125107303-125107303 on Build GRCh38
rs775400523 Chr.12: 125107147-125107147 on Build GRCh38
rs557257508 Chr.12: 125107206-125107206 on Build GRCh38
rs145117680 Chr.12: 125107169-125107169 on Build GRCh38
rs200174052 Chr.12: 125107111-125107111 on Build GRCh38
rs773314903 Chr.12: 125107301-125107301 on Build GRCh38
rs948867989 Chr.12: 125107409-125107409 on Build GRCh38
rs542969038 Chr.12: 125107389-125107389 on Build GRCh38
rs986141802 Chr.12: 125107329-125107329 on Build GRCh38
rs748786327 Chr.12: 125107165-125107165 on Build GRCh38
rs181063000 Chr.12: 125107390-125107390 on Build GRCh38
rs759581732 Chr.12: 125107139-125107139 on Build GRCh38
rs562410610 Chr.12: 125107235-125107235 on Build GRCh38
rs749291728 Chr.12: 125107267-125107267 on Build GRCh38
rs767563851 Chr.12: 125107222-125107222 on Build GRCh38
rs183531491 Chr.12: 125107148-125107148 on Build GRCh38
rs966041936 Chr.12: 125107374-125107374 on Build GRCh38
rs759192726 Chr.12: 125107089-125107089 on Build GRCh38
rs769440526 Chr.12: 125107143-125107143 on Build GRCh38
rs757356881 Chr.12: 125107106-125107106 on Build GRCh38
rs112666747 Chr.12: 125107379-125107379 on Build GRCh38

View Full Product Details