Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 26951628-26952121 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KDF1
Entrez Gene ID
126695
Gene Name
keratinocyte differentiation factor 1
Gene Aliases
C1orf172
UniGene
Hs.188881
Gene Chromosome Location
Chr.1: 26949556-26960484 on Build GRCh38
Transcripts
NM_152365.2 XM_005245735.2 XM_011540622.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5583331 c.502G>A p.G168S Chr.1: 26951879-26951879 on Build GRCh38
3487624 c.411C>T p.S137S Chr.1: 26951970-26951970 on Build GRCh38
907805 c.487G>A p.D163N Chr.1: 26951894-26951894 on Build GRCh38
1193002 c.521_522delAC p.Y174fs*9 Chr.1: 26951859-26951860 on Build GRCh38
3487622 c.507C>T p.I169I Chr.1: 26951874-26951874 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs541799014 Chr.1: 26952100-26952100 on Build GRCh38
rs765887283 Chr.1: 26952095-26952095 on Build GRCh38
rs899673096 Chr.1: 26951895-26951895 on Build GRCh38
rs766306128 Chr.1: 26951778-26951778 on Build GRCh38
rs754983927 Chr.1: 26951856-26951856 on Build GRCh38
rs780718896 Chr.1: 26951721-26951721 on Build GRCh38
rs768749901 Chr.1: 26952081-26952081 on Build GRCh38
rs566133911 Chr.1: 26951976-26951976 on Build GRCh38
rs777367914 Chr.1: 26951794-26951794 on Build GRCh38
rs550041376 Chr.1: 26951947-26951947 on Build GRCh38
rs148853297 Chr.1: 26952098-26952098 on Build GRCh38
rs781107975 Chr.1: 26951789-26951789 on Build GRCh38
rs547943153 Chr.1: 26951925-26951925 on Build GRCh38
rs754712166 Chr.1: 26951786-26951786 on Build GRCh38
rs924418483 Chr.1: 26951859-26951859 on Build GRCh38
rs755838886 Chr.1: 26951793-26951793 on Build GRCh38
rs550689765 Chr.1: 26952042-26952042 on Build GRCh38
rs758381938 Chr.1: 26951700-26951700 on Build GRCh38
rs371561424 Chr.1: 26952060-26952060 on Build GRCh38
rs771990259 Chr.1: 26951731-26951731 on Build GRCh38
rs770975636 Chr.1: 26951994-26951994 on Build GRCh38
rs199575697 Chr.1: 26951830-26951830 on Build GRCh38
rs3010109 Chr.1: 26952062-26952062 on Build GRCh38
rs756824633 Chr.1: 26951902-26951902 on Build GRCh38
rs746444382 Chr.1: 26951813-26951813 on Build GRCh38
rs767193569 Chr.1: 26951857-26951857 on Build GRCh38
rs749541622 Chr.1: 26952070-26952070 on Build GRCh38
rs753510550 Chr.1: 26951958-26951958 on Build GRCh38
rs752557636 Chr.1: 26951862-26951862 on Build GRCh38
rs774444521 Chr.1: 26951658-26951658 on Build GRCh38
rs17360994 Chr.1: 26952082-26952082 on Build GRCh38
rs371974787 Chr.1: 26951995-26951995 on Build GRCh38
rs761634533 Chr.1: 26951675-26951675 on Build GRCh38
rs751293758 Chr.1: 26951785-26951785 on Build GRCh38
rs377413091 Chr.1: 26951962-26951962 on Build GRCh38
rs150246438 Chr.1: 26951948-26951948 on Build GRCh38
rs772817105 Chr.1: 26951733-26951733 on Build GRCh38
rs368075916 Chr.1: 26951963-26951963 on Build GRCh38
rs369366004 Chr.1: 26951957-26951957 on Build GRCh38
rs144301593 Chr.1: 26952086-26952086 on Build GRCh38
rs572491048 Chr.1: 26951715-26951715 on Build GRCh38
rs749842745 Chr.1: 26952037-26952037 on Build GRCh38
rs748346371 Chr.1: 26951985-26951985 on Build GRCh38
rs147716816 Chr.1: 26951918-26951918 on Build GRCh38
rs752071068 Chr.1: 26952059-26952059 on Build GRCh38
rs767639195 Chr.1: 26951676-26951676 on Build GRCh38
rs139183337 Chr.1: 26952079-26952079 on Build GRCh38
rs534821230 Chr.1: 26951988-26951988 on Build GRCh38
rs1034227152 Chr.1: 26951992-26951992 on Build GRCh38
rs751490034 Chr.1: 26951852-26951852 on Build GRCh38
rs771599585 Chr.1: 26951741-26951741 on Build GRCh38
rs777979399 Chr.1: 26951690-26951690 on Build GRCh38
rs565350870 Chr.1: 26951917-26951917 on Build GRCh38
rs768999890 Chr.1: 26951980-26951980 on Build GRCh38
rs781573827 Chr.1: 26951981-26951981 on Build GRCh38
rs758768470 Chr.1: 26951971-26951971 on Build GRCh38
rs564540710 Chr.1: 26951926-26951926 on Build GRCh38
rs765356214 Chr.1: 26951851-26951851 on Build GRCh38
rs755657572 Chr.1: 26951870-26951870 on Build GRCh38
rs771012694 Chr.1: 26952077-26952077 on Build GRCh38
rs957243169 Chr.1: 26951913-26951913 on Build GRCh38
rs752617278 Chr.1: 26951972-26951973 on Build GRCh38
rs149743404 Chr.1: 26951812-26951812 on Build GRCh38
rs759220131 Chr.1: 26951774-26951775 on Build GRCh38
rs369086580 Chr.1: 26951706-26951706 on Build GRCh38
rs762549436 Chr.1: 26952094-26952094 on Build GRCh38
rs746835693 Chr.1: 26951714-26951714 on Build GRCh38
rs768805849 Chr.1: 26951932-26951932 on Build GRCh38
rs376036605 Chr.1: 26951795-26951795 on Build GRCh38
rs757023061 Chr.1: 26951960-26951960 on Build GRCh38
rs766818287 Chr.1: 26951657-26951657 on Build GRCh38
rs1018990840 Chr.1: 26951824-26951824 on Build GRCh38
rs761892290 Chr.1: 26951834-26951834 on Build GRCh38
rs775975243 Chr.1: 26951820-26951820 on Build GRCh38
rs749007417 Chr.1: 26951898-26951898 on Build GRCh38
rs748813446 Chr.1: 26951801-26951801 on Build GRCh38
rs374964459 Chr.1: 26951698-26951698 on Build GRCh38
rs142040426 Chr.1: 26951829-26951829 on Build GRCh38
rs772927829 Chr.1: 26951750-26951750 on Build GRCh38
rs200494012 Chr.1: 26951742-26951742 on Build GRCh38
rs527397998 Chr.1: 26951880-26951880 on Build GRCh38
rs371743170 Chr.1: 26951677-26951677 on Build GRCh38
rs887047479 Chr.1: 26951848-26951848 on Build GRCh38
rs767411580 Chr.1: 26951800-26951801 on Build GRCh38
rs761001620 Chr.1: 26951749-26951749 on Build GRCh38
rs753001226 Chr.1: 26952103-26952103 on Build GRCh38
rs753712013 Chr.1: 26952019-26952019 on Build GRCh38
rs201518791 Chr.1: 26951933-26951933 on Build GRCh38
rs763108668 Chr.1: 26951991-26951991 on Build GRCh38
rs761519934 Chr.1: 26952023-26952023 on Build GRCh38
rs758651584 Chr.1: 26951671-26951671 on Build GRCh38
rs756134971 Chr.1: 26951687-26951687 on Build GRCh38
rs778611060 Chr.1: 26951916-26951916 on Build GRCh38
rs372588334 Chr.1: 26951745-26951745 on Build GRCh38
rs535449942 Chr.1: 26951952-26951952 on Build GRCh38
rs374676574 Chr.1: 26951759-26951759 on Build GRCh38
rs779779172 Chr.1: 26951702-26951702 on Build GRCh38
rs747525165 Chr.1: 26951978-26951978 on Build GRCh38
rs982450139 Chr.1: 26952076-26952076 on Build GRCh38
rs141390568 Chr.1: 26951724-26951724 on Build GRCh38
rs769935791 Chr.1: 26952067-26952067 on Build GRCh38
rs772911827 Chr.1: 26952093-26952093 on Build GRCh38
rs373738003 Chr.1: 26952012-26952012 on Build GRCh38
rs770025779 Chr.1: 26951986-26951986 on Build GRCh38
rs139585050 Chr.1: 26951736-26951736 on Build GRCh38
rs1009596067 Chr.1: 26951954-26951954 on Build GRCh38
rs755438296 Chr.1: 26952099-26952099 on Build GRCh38
rs34291506 Chr.1: 26951815-26951815 on Build GRCh38
rs939725374 Chr.1: 26951974-26951974 on Build GRCh38
rs199725807 Chr.1: 26951996-26951996 on Build GRCh38
rs367617835 Chr.1: 26952050-26952050 on Build GRCh38
rs370198466 Chr.1: 26951802-26951802 on Build GRCh38
rs747723043 Chr.1: 26951792-26951792 on Build GRCh38
rs141234946 Chr.1: 26952068-26952068 on Build GRCh38
rs765268716 Chr.1: 26951944-26951944 on Build GRCh38
rs761717950 Chr.1: 26952083-26952083 on Build GRCh38
rs760507136 Chr.1: 26951956-26951956 on Build GRCh38
rs779492462 Chr.1: 26952058-26952058 on Build GRCh38
rs145994096 Chr.1: 26951768-26951768 on Build GRCh38
rs757910529 Chr.1: 26952057-26952057 on Build GRCh38
rs138248574 Chr.1: 26952061-26952061 on Build GRCh38
rs760644538 Chr.1: 26952080-26952080 on Build GRCh38
rs894118968 Chr.1: 26951716-26951716 on Build GRCh38
rs767984255 Chr.1: 26951746-26951746 on Build GRCh38
rs3010110 Chr.1: 26952030-26952030 on Build GRCh38

View Full Product Details