Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 11122633-11123130 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HCCS
Entrez Gene ID
3052
Gene Name
holocytochrome c synthase
Gene Aliases
CCHL, LSDMCA1, MCOPS7, MLS
UniGene
Hs.211571
Gene Chromosome Location
Chr.X: 11111286-11123086 on Build GRCh38
Transcripts
NM_001122608.2 NM_001171991.2 NM_005333.4

SNP Detail

SNP ID Chromosome Location
rs796945972 Chr.X: 11122745-11122745 on Build GRCh38
rs982347371 Chr.X: 11122955-11122955 on Build GRCh38
rs1004947178 Chr.X: 11122902-11122902 on Build GRCh38
rs775294797 Chr.X: 11122677-11122677 on Build GRCh38
rs771798364 Chr.X: 11122665-11122665 on Build GRCh38
rs886224674 Chr.X: 11122886-11122886 on Build GRCh38
rs182680236 Chr.X: 11122744-11122744 on Build GRCh38
rs761942801 Chr.X: 11122835-11122835 on Build GRCh38
rs961167198 Chr.X: 11122934-11122934 on Build GRCh38
rs992198103 Chr.X: 11122672-11122672 on Build GRCh38
rs141623095 Chr.X: 11122997-11122997 on Build GRCh38
rs1029205461 Chr.X: 11123076-11123076 on Build GRCh38
rs764569516 Chr.X: 11122869-11122870 on Build GRCh38
rs187244725 Chr.X: 11123043-11123043 on Build GRCh38
rs940442969 Chr.X: 11122889-11122889 on Build GRCh38
rs990782940 Chr.X: 11123004-11123004 on Build GRCh38
rs1040579799 Chr.X: 11122769-11122769 on Build GRCh38
rs766841089 Chr.X: 11123067-11123067 on Build GRCh38
rs1015020089 Chr.X: 11122909-11122909 on Build GRCh38
rs768032763 Chr.X: 11122916-11122916 on Build GRCh38
rs750614217 Chr.X: 11122983-11122983 on Build GRCh38
rs1009485658 Chr.X: 11122760-11122760 on Build GRCh38
rs1042085103 Chr.X: 11123072-11123072 on Build GRCh38
rs768677824 Chr.X: 11122731-11122731 on Build GRCh38
rs1028055769 Chr.X: 11122815-11122815 on Build GRCh38
rs930441964 Chr.X: 11122870-11122874 on Build GRCh38
rs1055198291 Chr.X: 11122689-11122689 on Build GRCh38
rs762125677 Chr.X: 11122903-11122907 on Build GRCh38
rs751839060 Chr.X: 11122893-11122893 on Build GRCh38
rs923683253 Chr.X: 11123088-11123088 on Build GRCh38
rs147898095 Chr.X: 11122802-11122802 on Build GRCh38
rs765579040 Chr.X: 11122945-11122945 on Build GRCh38
rs900747285 Chr.X: 11122774-11122774 on Build GRCh38
rs914934272 Chr.X: 11122712-11122712 on Build GRCh38
rs756323861 Chr.X: 11122730-11122730 on Build GRCh38
rs201397377 Chr.X: 11122880-11122881 on Build GRCh38
rs778496475 Chr.X: 11122767-11122770 on Build GRCh38
rs760658407 Chr.X: 11122728-11122728 on Build GRCh38
rs755130114 Chr.X: 11123048-11123048 on Build GRCh38
rs779991314 Chr.X: 11123027-11123027 on Build GRCh38
rs917908375 Chr.X: 11122753-11122753 on Build GRCh38
rs950038999 Chr.X: 11122771-11122775 on Build GRCh38
rs946457316 Chr.X: 11122719-11122719 on Build GRCh38
rs756188647 Chr.X: 11123011-11123011 on Build GRCh38
rs1048807353 Chr.X: 11122883-11122883 on Build GRCh38
rs956767659 Chr.X: 11122670-11122670 on Build GRCh38
rs980015430 Chr.X: 11122801-11122801 on Build GRCh38

View Full Product Details