Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49928690-49929222 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATF5
Entrez Gene ID
22809
Gene Name
activating transcription factor 5
Gene Aliases
ATFX, HMFN0395
UniGene
Hs.9754
Gene Chromosome Location
Chr.19: 49928702-49933936 on Build GRCh38
Transcripts
NM_001193646.1 NM_012068.5 NM_001258017.1 NM_001258018.1 NM_172374.2 NR_047577.1 NM_001193357.1 NM_012346.4 NM_016553.4 NM_153718.3 NM_153719.3

Gene Symbol
IL4I1
Entrez Gene ID
259307
Gene Name
interleukin 4 induced 1
Gene Aliases
FIG1, LAAO, LAO
UniGene
Hs.574492
Gene Chromosome Location
Chr.19: 49889654-49929539 on Build GRCh38
Transcripts
NM_001193646.1 NM_012068.5 NM_001258017.1 NM_001258018.1 NM_172374.2 NR_047577.1 NM_001193357.1 NM_012346.4 NM_016553.4 NM_153718.3 NM_153719.3

Gene Symbol
NUP62
Entrez Gene ID
23636
Gene Name
nucleoporin 62
Gene Aliases
IBSN, SNDI, p62
UniGene
Hs.574492
Gene Chromosome Location
Chr.19: 49906825-49929731 on Build GRCh38
Transcripts
NM_001193646.1 NM_012068.5 NM_001258017.1 NM_001258018.1 NM_172374.2 NR_047577.1 NM_001193357.1 NM_012346.4 NM_016553.4 NM_153718.3 NM_153719.3

SNP Detail

SNP ID Chromosome Location
rs957693001 Chr.19: 49929001-49929001 on Build GRCh38
rs929049692 Chr.19: 49928894-49928894 on Build GRCh38
rs540655839 Chr.19: 49928917-49928917 on Build GRCh38
rs898785679 Chr.19: 49929119-49929119 on Build GRCh38
rs1045923727 Chr.19: 49928887-49928887 on Build GRCh38
rs987955061 Chr.19: 49928797-49928797 on Build GRCh38
rs759626401 Chr.19: 49929046-49929046 on Build GRCh38
rs747823750 Chr.19: 49929150-49929150 on Build GRCh38
rs998279166 Chr.19: 49928711-49928711 on Build GRCh38
rs1015581292 Chr.19: 49928767-49928767 on Build GRCh38
rs1016926638 Chr.19: 49928745-49928745 on Build GRCh38
rs398079930 Chr.19: 49929192-49929192 on Build GRCh38
rs960749128 Chr.19: 49928962-49928962 on Build GRCh38
rs922946717 Chr.19: 49929075-49929075 on Build GRCh38
rs967985052 Chr.19: 49928842-49928842 on Build GRCh38
rs576212006 Chr.19: 49928777-49928777 on Build GRCh38
rs954186013 Chr.19: 49928947-49928947 on Build GRCh38
rs1004440324 Chr.19: 49928911-49928911 on Build GRCh38
rs905408194 Chr.19: 49929171-49929171 on Build GRCh38
rs1023271777 Chr.19: 49929190-49929190 on Build GRCh38
rs775931068 Chr.19: 49929080-49929080 on Build GRCh38
rs999348086 Chr.19: 49929173-49929173 on Build GRCh38
rs531169565 Chr.19: 49929030-49929030 on Build GRCh38
rs977990368 Chr.19: 49928852-49928852 on Build GRCh38
rs528479000 Chr.19: 49929168-49929168 on Build GRCh38
rs563914335 Chr.19: 49929027-49929027 on Build GRCh38
rs907383308 Chr.19: 49928895-49928895 on Build GRCh38
rs986687291 Chr.19: 49928834-49928834 on Build GRCh38
rs995496481 Chr.19: 49929122-49929123 on Build GRCh38
rs772561002 Chr.19: 49928991-49928991 on Build GRCh38
rs543324007 Chr.19: 49928845-49928845 on Build GRCh38
rs1032206637 Chr.19: 49929178-49929178 on Build GRCh38
rs187416943 Chr.19: 49928771-49928771 on Build GRCh38
rs1047696360 Chr.19: 49928714-49928714 on Build GRCh38
rs969414631 Chr.19: 49928753-49928753 on Build GRCh38
rs985490618 Chr.19: 49928983-49928983 on Build GRCh38
rs908688542 Chr.19: 49929112-49929112 on Build GRCh38
rs139891865 Chr.19: 49929007-49929007 on Build GRCh38
rs553260896 Chr.19: 49928987-49928987 on Build GRCh38
rs567987474 Chr.19: 49929136-49929136 on Build GRCh38
rs1009636685 Chr.19: 49929184-49929184 on Build GRCh38
rs896518693 Chr.19: 49929183-49929183 on Build GRCh38
rs975680702 Chr.19: 49929064-49929064 on Build GRCh38
rs150654811 Chr.19: 49928885-49928885 on Build GRCh38
rs964617085 Chr.19: 49929049-49929049 on Build GRCh38
rs903950851 Chr.19: 49929195-49929195 on Build GRCh38
rs889794801 Chr.19: 49929159-49929159 on Build GRCh38
rs1020785015 Chr.19: 49928981-49928981 on Build GRCh38
rs914373212 Chr.19: 49928867-49928887 on Build GRCh38
rs777856609 Chr.19: 49928870-49928870 on Build GRCh38
rs1047193234 Chr.19: 49928909-49928909 on Build GRCh38
rs922715233 Chr.19: 49928738-49928738 on Build GRCh38
rs1002183501 Chr.19: 49928953-49928953 on Build GRCh38
rs376430172 Chr.19: 49928744-49928744 on Build GRCh38
rs772636098 Chr.19: 49929172-49929172 on Build GRCh38
rs1046585996 Chr.19: 49929059-49929059 on Build GRCh38
rs1013123859 Chr.19: 49928740-49928740 on Build GRCh38
rs898462079 Chr.19: 49928919-49928919 on Build GRCh38
rs397823983 Chr.19: 49929193-49929193 on Build GRCh38
rs1056473078 Chr.19: 49929181-49929181 on Build GRCh38
rs1010101572 Chr.19: 49928974-49928974 on Build GRCh38
rs887327449 Chr.19: 49928743-49928743 on Build GRCh38
rs779454798 Chr.19: 49928944-49928944 on Build GRCh38
rs910201195 Chr.19: 49928857-49928857 on Build GRCh38
rs561691930 Chr.19: 49928865-49928865 on Build GRCh38
rs943068181 Chr.19: 49928862-49928862 on Build GRCh38
rs912449115 Chr.19: 49928839-49928839 on Build GRCh38
rs1152231 Chr.19: 49929194-49929194 on Build GRCh38
rs929061446 Chr.19: 49929085-49929085 on Build GRCh38
rs5828422 Chr.19: 49929190-49929190 on Build GRCh38
rs757746857 Chr.19: 49928935-49928935 on Build GRCh38
rs549695089 Chr.19: 49929055-49929055 on Build GRCh38
rs201781766 Chr.19: 49929130-49929130 on Build GRCh38
rs35018314 Chr.19: 49928933-49928933 on Build GRCh38
rs1028416048 Chr.19: 49928789-49928789 on Build GRCh38
rs966951884 Chr.19: 49928978-49928978 on Build GRCh38
rs960802668 Chr.19: 49928793-49928793 on Build GRCh38
rs570055889 Chr.19: 49928836-49928836 on Build GRCh38
rs1152232 Chr.19: 49929170-49929170 on Build GRCh38
rs1035023973 Chr.19: 49928959-49928959 on Build GRCh38
rs887305919 Chr.19: 49928932-49928932 on Build GRCh38
rs913734969 Chr.19: 49928832-49928832 on Build GRCh38
rs916298050 Chr.19: 49929040-49929040 on Build GRCh38
rs1047973771 Chr.19: 49929094-49929094 on Build GRCh38
rs1054808147 Chr.19: 49928725-49928725 on Build GRCh38
rs779645621 Chr.19: 49929133-49929133 on Build GRCh38

View Full Product Details