Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42838854-42839351 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSME3
Entrez Gene ID
10197
Gene Name
proteasome activator subunit 3
Gene Aliases
HEL-S-283, Ki, PA28-gamma, PA28G, PA28gamma, REG-GAMMA
UniGene
Hs.152978
Gene Chromosome Location
Chr.17: 42833142-42843760 on Build GRCh38
Transcripts
NM_001267045.1 NM_005789.3 NM_176863.2 NR_049772.1 XM_011524177.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5043344 c.647G>T p.R216L Chr.17: 42839304-42839304 on Build GRCh38
6757896 c.541G>A p.V181I Chr.17: 42838972-42838972 on Build GRCh38
163792 c.687C>T p.S229S Chr.17: 42839344-42839344 on Build GRCh38
7150294 c.568G>C p.D190H Chr.17: 42838999-42838999 on Build GRCh38
5458632 c.647G>A p.R216H Chr.17: 42839304-42839304 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777477695 Chr.17: 42839251-42839251 on Build GRCh38
rs754717237 Chr.17: 42839041-42839041 on Build GRCh38
rs762048193 Chr.17: 42839134-42839134 on Build GRCh38
rs927724232 Chr.17: 42839240-42839240 on Build GRCh38
rs146436454 Chr.17: 42839238-42839238 on Build GRCh38
rs758185411 Chr.17: 42839323-42839323 on Build GRCh38
rs558371308 Chr.17: 42839235-42839235 on Build GRCh38
rs961027127 Chr.17: 42839196-42839196 on Build GRCh38
rs376784465 Chr.17: 42839115-42839115 on Build GRCh38
rs780434170 Chr.17: 42839036-42839036 on Build GRCh38
rs373515453 Chr.17: 42839257-42839257 on Build GRCh38
rs771716130 Chr.17: 42839175-42839175 on Build GRCh38
rs183278888 Chr.17: 42839279-42839279 on Build GRCh38
rs776539482 Chr.17: 42838931-42838931 on Build GRCh38
rs766375896 Chr.17: 42839296-42839296 on Build GRCh38
rs745760307 Chr.17: 42839265-42839265 on Build GRCh38
rs562160281 Chr.17: 42839271-42839271 on Build GRCh38
rs777013419 Chr.17: 42839300-42839300 on Build GRCh38
rs777366069 Chr.17: 42839056-42839056 on Build GRCh38
rs959233045 Chr.17: 42839264-42839264 on Build GRCh38
rs745649273 Chr.17: 42839089-42839089 on Build GRCh38
rs373230666 Chr.17: 42838894-42838894 on Build GRCh38
rs770884351 Chr.17: 42838923-42838923 on Build GRCh38
rs900546765 Chr.17: 42839022-42839022 on Build GRCh38
rs755298030 Chr.17: 42839211-42839211 on Build GRCh38
rs944840010 Chr.17: 42838883-42838883 on Build GRCh38
rs1015900999 Chr.17: 42839090-42839090 on Build GRCh38
rs376102848 Chr.17: 42838989-42838989 on Build GRCh38
rs764212450 Chr.17: 42839015-42839015 on Build GRCh38
rs778533249 Chr.17: 42838903-42838903 on Build GRCh38
rs373252756 Chr.17: 42839078-42839078 on Build GRCh38
rs745317701 Chr.17: 42839094-42839096 on Build GRCh38
rs761219791 Chr.17: 42839195-42839195 on Build GRCh38
rs774172014 Chr.17: 42839093-42839097 on Build GRCh38
rs750951881 Chr.17: 42839029-42839029 on Build GRCh38
rs760739410 Chr.17: 42839290-42839290 on Build GRCh38
rs765578029 Chr.17: 42839212-42839212 on Build GRCh38
rs1049209347 Chr.17: 42839039-42839039 on Build GRCh38
rs770972368 Chr.17: 42839070-42839070 on Build GRCh38
rs371079095 Chr.17: 42839217-42839217 on Build GRCh38
rs775654817 Chr.17: 42839082-42839082 on Build GRCh38
rs769566465 Chr.17: 42839098-42839098 on Build GRCh38
rs760037001 Chr.17: 42839306-42839306 on Build GRCh38
rs369233513 Chr.17: 42839040-42839040 on Build GRCh38
rs1031746915 Chr.17: 42838992-42838992 on Build GRCh38
rs536446351 Chr.17: 42839079-42839079 on Build GRCh38
rs772110359 Chr.17: 42838912-42838912 on Build GRCh38
rs766856882 Chr.17: 42839201-42839201 on Build GRCh38
rs770669646 Chr.17: 42839054-42839055 on Build GRCh38
rs772258450 Chr.17: 42839282-42839282 on Build GRCh38
rs780997204 Chr.17: 42839261-42839261 on Build GRCh38
rs763289004 Chr.17: 42838995-42838995 on Build GRCh38
rs760628347 Chr.17: 42838922-42838922 on Build GRCh38
rs746986085 Chr.17: 42839058-42839058 on Build GRCh38
rs773338259 Chr.17: 42839285-42839285 on Build GRCh38
rs966756879 Chr.17: 42839307-42839307 on Build GRCh38
rs747594630 Chr.17: 42838904-42838904 on Build GRCh38
rs1043626055 Chr.17: 42839287-42839287 on Build GRCh38
rs756572650 Chr.17: 42839035-42839035 on Build GRCh38
rs1039225422 Chr.17: 42838924-42838924 on Build GRCh38
rs773285422 Chr.17: 42838915-42838915 on Build GRCh38
rs752311685 Chr.17: 42839215-42839215 on Build GRCh38
rs768431520 Chr.17: 42839034-42839034 on Build GRCh38
rs773243590 Chr.17: 42839160-42839160 on Build GRCh38
rs757685046 Chr.17: 42839222-42839223 on Build GRCh38
rs746509532 Chr.17: 42839252-42839252 on Build GRCh38
rs747764905 Chr.17: 42839050-42839050 on Build GRCh38
rs773945626 Chr.17: 42839204-42839204 on Build GRCh38
rs771718468 Chr.17: 42839054-42839054 on Build GRCh38
rs868306393 Chr.17: 42839109-42839109 on Build GRCh38
rs748409378 Chr.17: 42839281-42839281 on Build GRCh38
rs139649789 Chr.17: 42839311-42839311 on Build GRCh38
rs754292655 Chr.17: 42839206-42839206 on Build GRCh38
rs778422228 Chr.17: 42839046-42839046 on Build GRCh38
rs753086811 Chr.17: 42839317-42839317 on Build GRCh38
rs201575339 Chr.17: 42839021-42839021 on Build GRCh38
rs769501440 Chr.17: 42839270-42839270 on Build GRCh38
rs376387190 Chr.17: 42839020-42839020 on Build GRCh38

View Full Product Details