Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 35869360-35869895 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SRPK1
Entrez Gene ID
6732
Gene Name
SRSF protein kinase 1
Gene Aliases
SFRSK1
UniGene
Hs.443861
Gene Chromosome Location
Chr.6: 35833034-35921180 on Build GRCh38
Transcripts
NM_003137.4 NR_034069.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5960807 c.1303C>T p.Q435* Chr.6: 35869590-35869590 on Build GRCh38
1078436 c.1005C>A p.T335T Chr.6: 35869888-35869888 on Build GRCh38
3075733 c.1065T>G p.N355K Chr.6: 35869828-35869828 on Build GRCh38
5979185 c.1046G>A p.G349D Chr.6: 35869847-35869847 on Build GRCh38
6106761 c.1363T>G p.C455G Chr.6: 35869530-35869530 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372083147 Chr.6: 35869645-35869645 on Build GRCh38
rs1028161687 Chr.6: 35869648-35869648 on Build GRCh38
rs760520593 Chr.6: 35869434-35869434 on Build GRCh38
rs146325361 Chr.6: 35869387-35869387 on Build GRCh38
rs764159064 Chr.6: 35869545-35869545 on Build GRCh38
rs79027141 Chr.6: 35869871-35869871 on Build GRCh38
rs777298905 Chr.6: 35869753-35869753 on Build GRCh38
rs141819656 Chr.6: 35869770-35869770 on Build GRCh38
rs767723804 Chr.6: 35869666-35869666 on Build GRCh38
rs781743706 Chr.6: 35869477-35869477 on Build GRCh38
rs776491550 Chr.6: 35869640-35869640 on Build GRCh38
rs769726012 Chr.6: 35869623-35869623 on Build GRCh38
rs766156743 Chr.6: 35869439-35869439 on Build GRCh38
rs754670580 Chr.6: 35869591-35869591 on Build GRCh38
rs138230979 Chr.6: 35869624-35869624 on Build GRCh38
rs747832186 Chr.6: 35869615-35869615 on Build GRCh38
rs774830131 Chr.6: 35869557-35869557 on Build GRCh38
rs373060394 Chr.6: 35869870-35869870 on Build GRCh38
rs766474058 Chr.6: 35869847-35869847 on Build GRCh38
rs768150388 Chr.6: 35869500-35869500 on Build GRCh38
rs752342368 Chr.6: 35869733-35869733 on Build GRCh38
rs773198874 Chr.6: 35869523-35869523 on Build GRCh38
rs776364168 Chr.6: 35869541-35869541 on Build GRCh38
rs951185216 Chr.6: 35869596-35869596 on Build GRCh38
rs199917638 Chr.6: 35869789-35869789 on Build GRCh38
rs757790254 Chr.6: 35869471-35869471 on Build GRCh38
rs746451428 Chr.6: 35869755-35869755 on Build GRCh38
rs770817432 Chr.6: 35869634-35869634 on Build GRCh38
rs919661212 Chr.6: 35869412-35869412 on Build GRCh38
rs919565817 Chr.6: 35869772-35869772 on Build GRCh38
rs751013450 Chr.6: 35869561-35869561 on Build GRCh38
rs990133048 Chr.6: 35869751-35869751 on Build GRCh38
rs773076358 Chr.6: 35869633-35869633 on Build GRCh38
rs368969962 Chr.6: 35869752-35869752 on Build GRCh38
rs753066315 Chr.6: 35869857-35869857 on Build GRCh38
rs201807680 Chr.6: 35869853-35869853 on Build GRCh38
rs770762353 Chr.6: 35869535-35869535 on Build GRCh38
rs373488236 Chr.6: 35869726-35869726 on Build GRCh38
rs778485680 Chr.6: 35869592-35869592 on Build GRCh38
rs745817501 Chr.6: 35869785-35869785 on Build GRCh38
rs763268578 Chr.6: 35869544-35869544 on Build GRCh38
rs764256849 Chr.6: 35869467-35869467 on Build GRCh38
rs1046514835 Chr.6: 35869779-35869779 on Build GRCh38
rs752958542 Chr.6: 35869456-35869456 on Build GRCh38
rs767705257 Chr.6: 35869559-35869559 on Build GRCh38
rs748408490 Chr.6: 35869806-35869806 on Build GRCh38
rs756636608 Chr.6: 35869572-35869572 on Build GRCh38
rs774507529 Chr.6: 35869647-35869647 on Build GRCh38
rs376417169 Chr.6: 35869652-35869652 on Build GRCh38
rs45481397 Chr.6: 35869799-35869799 on Build GRCh38
rs762509990 Chr.6: 35869865-35869865 on Build GRCh38
rs760065558 Chr.6: 35869850-35869850 on Build GRCh38
rs756583604 Chr.6: 35869487-35869487 on Build GRCh38
rs920945589 Chr.6: 35869383-35869383 on Build GRCh38
rs369251843 Chr.6: 35869699-35869699 on Build GRCh38
rs760575974 Chr.6: 35869533-35869533 on Build GRCh38
rs908225541 Chr.6: 35869382-35869382 on Build GRCh38
rs757203756 Chr.6: 35869763-35869763 on Build GRCh38
rs751676752 Chr.6: 35869548-35869548 on Build GRCh38
rs370339672 Chr.6: 35869479-35869479 on Build GRCh38
rs760280330 Chr.6: 35869738-35869738 on Build GRCh38
rs973835462 Chr.6: 35869405-35869405 on Build GRCh38
rs747553710 Chr.6: 35869505-35869505 on Build GRCh38
rs763065308 Chr.6: 35869457-35869457 on Build GRCh38
rs376600317 Chr.6: 35869744-35869744 on Build GRCh38
rs760893876 Chr.6: 35869834-35869834 on Build GRCh38
rs543559215 Chr.6: 35869825-35869825 on Build GRCh38
rs1015463885 Chr.6: 35869743-35869743 on Build GRCh38
rs772351769 Chr.6: 35869812-35869812 on Build GRCh38
rs186925364 Chr.6: 35869506-35869506 on Build GRCh38
rs755365214 Chr.6: 35869707-35869707 on Build GRCh38
rs773383685 Chr.6: 35869678-35869678 on Build GRCh38
rs761860738 Chr.6: 35869555-35869555 on Build GRCh38
rs777044045 Chr.6: 35869848-35869848 on Build GRCh38
rs754080884 Chr.6: 35869590-35869590 on Build GRCh38
rs1046369089 Chr.6: 35869574-35869574 on Build GRCh38
rs748987695 Chr.6: 35869493-35869493 on Build GRCh38
rs778489915 Chr.6: 35869501-35869501 on Build GRCh38
rs746971962 Chr.6: 35869632-35869632 on Build GRCh38
rs372259775 Chr.6: 35869604-35869604 on Build GRCh38
rs761266237 Chr.6: 35869694-35869694 on Build GRCh38
rs201293031 Chr.6: 35869644-35869644 on Build GRCh38
rs780594079 Chr.6: 35869576-35869576 on Build GRCh38
rs373801550 Chr.6: 35869488-35869488 on Build GRCh38
rs995811456 Chr.6: 35869567-35869567 on Build GRCh38

View Full Product Details