Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 36076453-36076948 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAPK14
Entrez Gene ID
1432
Gene Name
mitogen-activated protein kinase 14
Gene Aliases
CSBP, CSBP1, CSBP2, CSPB1, EXIP, Mxi2, PRKM14, PRKM15, RK, SAPK2A, p38, p38ALPHA
UniGene
Hs.485233
Gene Chromosome Location
Chr.6: 36027635-36122964 on Build GRCh38
Transcripts
NM_001315.2 NM_139012.2 NM_139013.2 NM_139014.2 XM_006714998.3 XM_011514310.2 XM_017010299.1 XM_017010300.1 XM_017010301.1 XM_017010302.1 XM_017010303.1 XM_017010304.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5731987 c.656G>C p.G219A Chr.6: 36076582-36076582 on Build GRCh38
1621578 c.633C>T p.C211C Chr.6: 36076559-36076559 on Build GRCh38
5599894 c.679G>A p.D227N Chr.6: 36076605-36076605 on Build GRCh38
5731988 c.656G>C p.G219A Chr.6: 36076582-36076582 on Build GRCh38
5599893 c.679G>A p.D227N Chr.6: 36076605-36076605 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773348265 Chr.6: 36076527-36076527 on Build GRCh38
rs754861351 Chr.6: 36076562-36076562 on Build GRCh38
rs745992492 Chr.6: 36076675-36076675 on Build GRCh38
rs200890402 Chr.6: 36076634-36076634 on Build GRCh38
rs201154007 Chr.6: 36076506-36076506 on Build GRCh38
rs958007590 Chr.6: 36076502-36076502 on Build GRCh38
rs778959239 Chr.6: 36076491-36076492 on Build GRCh38
rs532912902 Chr.6: 36076885-36076885 on Build GRCh38
rs755087316 Chr.6: 36076661-36076661 on Build GRCh38
rs972194130 Chr.6: 36076709-36076709 on Build GRCh38
rs1052908290 Chr.6: 36076917-36076917 on Build GRCh38
rs763820993 Chr.6: 36076618-36076618 on Build GRCh38
rs751127696 Chr.6: 36076625-36076625 on Build GRCh38
rs867509609 Chr.6: 36076706-36076706 on Build GRCh38
rs758049381 Chr.6: 36076617-36076617 on Build GRCh38
rs765566668 Chr.6: 36076589-36076589 on Build GRCh38
rs746509575 Chr.6: 36076493-36076493 on Build GRCh38
rs916834140 Chr.6: 36076867-36076867 on Build GRCh38
rs544242689 Chr.6: 36076834-36076834 on Build GRCh38
rs756058350 Chr.6: 36076654-36076654 on Build GRCh38
rs200577487 Chr.6: 36076562-36076562 on Build GRCh38
rs750277702 Chr.6: 36076647-36076647 on Build GRCh38
rs757320668 Chr.6: 36076496-36076496 on Build GRCh38
rs911736262 Chr.6: 36076665-36076665 on Build GRCh38
rs199788193 Chr.6: 36076511-36076511 on Build GRCh38
rs201451587 Chr.6: 36076568-36076568 on Build GRCh38
rs201679204 Chr.6: 36076657-36076657 on Build GRCh38
rs756759557 Chr.6: 36076627-36076627 on Build GRCh38
rs561968074 Chr.6: 36076725-36076725 on Build GRCh38
rs994669000 Chr.6: 36076884-36076884 on Build GRCh38
rs774658039 Chr.6: 36076513-36076513 on Build GRCh38
rs940436836 Chr.6: 36076688-36076688 on Build GRCh38
rs540582625 Chr.6: 36076717-36076717 on Build GRCh38
rs7341337 Chr.6: 36076854-36076854 on Build GRCh38
rs546248556 Chr.6: 36076499-36076499 on Build GRCh38
rs374851293 Chr.6: 36076483-36076483 on Build GRCh38
rs748369308 Chr.6: 36076514-36076514 on Build GRCh38
rs760735293 Chr.6: 36076556-36076556 on Build GRCh38
rs144970397 Chr.6: 36076892-36076892 on Build GRCh38
rs369279388 Chr.6: 36076721-36076721 on Build GRCh38
rs375208032 Chr.6: 36076572-36076572 on Build GRCh38
rs201010037 Chr.6: 36076517-36076517 on Build GRCh38
rs948307996 Chr.6: 36076871-36076871 on Build GRCh38
rs150550578 Chr.6: 36076608-36076608 on Build GRCh38
rs905819911 Chr.6: 36076655-36076655 on Build GRCh38
rs930263256 Chr.6: 36076744-36076744 on Build GRCh38
rs1046547499 Chr.6: 36076883-36076883 on Build GRCh38
rs987317274 Chr.6: 36076636-36076636 on Build GRCh38
rs190663927 Chr.6: 36076767-36076767 on Build GRCh38
rs745678445 Chr.6: 36076504-36076504 on Build GRCh38

View Full Product Details