Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 150286137-150286646 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTR3C
Entrez Gene ID
653857
Gene Name
ARP3 actin-related protein 3 homolog C
Gene Aliases
ARP11
UniGene
Hs.664579
Gene Chromosome Location
Chr.7: 149881359-150325523 on Build GRCh38
Transcripts
NM_001164458.1 NM_001164459.1 XM_005250043.4 XM_011516502.2 XM_011516506.2 XM_011516507.2 XM_011516508.2 XM_011516511.2 XM_017012548.1 XM_017012549.1 XM_017012550.1 XM_017012551.1 XM_017012552.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1244193 c.318C>T p.C106C Chr.7: 150286520-150286520 on Build GRCh38
4659172 c.361C>T p.Q121* Chr.7: 150286477-150286477 on Build GRCh38
150416 c.362A>G p.Q121R Chr.7: 150286476-150286476 on Build GRCh38
6332510 c.384G>A p.T128T Chr.7: 150286454-150286454 on Build GRCh38
4950504 c.412T>C p.F138L Chr.7: 150286426-150286426 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751701641 Chr.7: 150286351-150286351 on Build GRCh38
rs760716791 Chr.7: 150286559-150286559 on Build GRCh38
rs575373432 Chr.7: 150286165-150286165 on Build GRCh38
rs201926440 Chr.7: 150286453-150286453 on Build GRCh38
rs745733638 Chr.7: 150286517-150286517 on Build GRCh38
rs746495404 Chr.7: 150286488-150286488 on Build GRCh38
rs796732979 Chr.7: 150286235-150286236 on Build GRCh38
rs776615349 Chr.7: 150286415-150286415 on Build GRCh38
rs769839278 Chr.7: 150286427-150286427 on Build GRCh38
rs200800061 Chr.7: 150286535-150286535 on Build GRCh38
rs397833374 Chr.7: 150286361-150286361 on Build GRCh38
rs550912368 Chr.7: 150286454-150286454 on Build GRCh38
rs1031867297 Chr.7: 150286301-150286301 on Build GRCh38
rs767502283 Chr.7: 150286451-150286451 on Build GRCh38
rs751271103 Chr.7: 150286350-150286355 on Build GRCh38
rs1005138664 Chr.7: 150286611-150286611 on Build GRCh38
rs779281142 Chr.7: 150286381-150286381 on Build GRCh38
rs1011214021 Chr.7: 150286325-150286325 on Build GRCh38
rs747748166 Chr.7: 150286392-150286392 on Build GRCh38
rs751182836 Chr.7: 150286582-150286582 on Build GRCh38
rs200546664 Chr.7: 150286526-150286526 on Build GRCh38
rs775276555 Chr.7: 150286443-150286443 on Build GRCh38
rs780988364 Chr.7: 150286516-150286516 on Build GRCh38
rs754155408 Chr.7: 150286371-150286371 on Build GRCh38
rs770655745 Chr.7: 150286414-150286414 on Build GRCh38
rs759566827 Chr.7: 150286348-150286348 on Build GRCh38
rs777269820 Chr.7: 150286401-150286401 on Build GRCh38
rs922137339 Chr.7: 150286446-150286446 on Build GRCh38
rs769491476 Chr.7: 150286519-150286519 on Build GRCh38
rs559795508 Chr.7: 150286261-150286261 on Build GRCh38
rs777447287 Chr.7: 150286487-150286487 on Build GRCh38
rs141976587 Chr.7: 150286581-150286581 on Build GRCh38
rs544111832 Chr.7: 150286193-150286193 on Build GRCh38
rs754265501 Chr.7: 150286457-150286457 on Build GRCh38
rs773333930 Chr.7: 150286539-150286539 on Build GRCh38
rs776849456 Chr.7: 150286565-150286565 on Build GRCh38
rs766568588 Chr.7: 150286456-150286456 on Build GRCh38
rs1051935331 Chr.7: 150286254-150286254 on Build GRCh38
rs1022804245 Chr.7: 150286340-150286340 on Build GRCh38
rs565470960 Chr.7: 150286372-150286372 on Build GRCh38
rs78661149 Chr.7: 150286477-150286477 on Build GRCh38
rs765062516 Chr.7: 150286349-150286349 on Build GRCh38
rs746689733 Chr.7: 150286406-150286406 on Build GRCh38
rs757008908 Chr.7: 150286504-150286504 on Build GRCh38
rs758897453 Chr.7: 150286486-150286486 on Build GRCh38
rs200584229 Chr.7: 150286370-150286370 on Build GRCh38
rs998605104 Chr.7: 150286284-150286284 on Build GRCh38
rs756814586 Chr.7: 150286585-150286585 on Build GRCh38
rs761967087 Chr.7: 150286442-150286442 on Build GRCh38
rs753145621 Chr.7: 150286484-150286484 on Build GRCh38
rs367599886 Chr.7: 150286250-150286251 on Build GRCh38
rs567305387 Chr.7: 150286455-150286455 on Build GRCh38
rs753055942 Chr.7: 150286573-150286573 on Build GRCh38
rs538017544 Chr.7: 150286569-150286569 on Build GRCh38
rs756266177 Chr.7: 150286364-150286364 on Build GRCh38
rs756895143 Chr.7: 150286538-150286538 on Build GRCh38
rs749217549 Chr.7: 150286528-150286528 on Build GRCh38
rs10258147 Chr.7: 150286570-150286570 on Build GRCh38
rs71531433 Chr.7: 150286476-150286476 on Build GRCh38
rs981363997 Chr.7: 150286366-150286366 on Build GRCh38
rs574072331 Chr.7: 150286236-150286236 on Build GRCh38
rs1016318250 Chr.7: 150286613-150286613 on Build GRCh38
rs28634863 Chr.7: 150286476-150286476 on Build GRCh38
rs1039281688 Chr.7: 150286169-150286169 on Build GRCh38
rs892784087 Chr.7: 150286319-150286319 on Build GRCh38
rs199639052 Chr.7: 150286374-150286374 on Build GRCh38
rs762890577 Chr.7: 150286450-150286450 on Build GRCh38
rs199920648 Chr.7: 150286353-150286353 on Build GRCh38
rs112895300 Chr.7: 150286245-150286246 on Build GRCh38
rs150693647 Chr.7: 150286594-150286594 on Build GRCh38
rs950640251 Chr.7: 150286375-150286375 on Build GRCh38
rs776527336 Chr.7: 150286335-150286335 on Build GRCh38
rs28527325 Chr.7: 150286235-150286235 on Build GRCh38
rs200154560 Chr.7: 150286624-150286624 on Build GRCh38
rs757856792 Chr.7: 150286399-150286399 on Build GRCh38
rs569072888 Chr.7: 150286561-150286561 on Build GRCh38
rs770853733 Chr.7: 150286323-150286323 on Build GRCh38
rs189313027 Chr.7: 150286469-150286469 on Build GRCh38
rs758878790 Chr.7: 150286578-150286578 on Build GRCh38
rs28478124 Chr.7: 150286243-150286243 on Build GRCh38
rs553553115 Chr.7: 150286602-150286602 on Build GRCh38

View Full Product Details