Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 27747952-27748470 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC25
Entrez Gene ID
55246
Gene Name
coiled-coil domain containing 25
Gene Aliases
-
UniGene
Hs.445512
Gene Chromosome Location
Chr.8: 27733316-27772656 on Build GRCh38
Transcripts
NM_001304529.1 NM_001304530.1 NM_001304532.1 NM_018246.2 NR_130761.1 NR_130762.1 XM_011544570.2 XM_011544571.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6622019 c.457C>T p.R153C Chr.8: 27748171-27748171 on Build GRCh38
5409337 c.468G>A p.R156R Chr.8: 27748160-27748160 on Build GRCh38
3731412 c.360delG p.T121fs*6 Chr.8: 27748268-27748268 on Build GRCh38
3768763 c.457C>G p.R153G Chr.8: 27748171-27748171 on Build GRCh38
5417202 c.450C>T p.C150C Chr.8: 27748178-27748178 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747145585 Chr.8: 27748445-27748445 on Build GRCh38
rs745998391 Chr.8: 27748046-27748046 on Build GRCh38
rs976565648 Chr.8: 27748017-27748017 on Build GRCh38
rs370334946 Chr.8: 27748302-27748302 on Build GRCh38
rs150006124 Chr.8: 27748205-27748205 on Build GRCh38
rs775114186 Chr.8: 27748261-27748261 on Build GRCh38
rs751077565 Chr.8: 27748121-27748121 on Build GRCh38
rs775584926 Chr.8: 27748061-27748061 on Build GRCh38
rs779979329 Chr.8: 27748328-27748328 on Build GRCh38
rs145210673 Chr.8: 27748209-27748209 on Build GRCh38
rs201322823 Chr.8: 27748096-27748096 on Build GRCh38
rs762964810 Chr.8: 27748093-27748093 on Build GRCh38
rs148042574 Chr.8: 27748019-27748019 on Build GRCh38
rs752261573 Chr.8: 27748319-27748319 on Build GRCh38
rs773185109 Chr.8: 27748086-27748086 on Build GRCh38
rs770927397 Chr.8: 27748232-27748232 on Build GRCh38
rs907473555 Chr.8: 27748033-27748033 on Build GRCh38
rs770569574 Chr.8: 27748146-27748146 on Build GRCh38
rs780171235 Chr.8: 27748114-27748114 on Build GRCh38
rs202197146 Chr.8: 27748170-27748170 on Build GRCh38
rs76860931 Chr.8: 27748416-27748416 on Build GRCh38
rs758974711 Chr.8: 27748130-27748130 on Build GRCh38
rs986813879 Chr.8: 27748294-27748294 on Build GRCh38
rs750922064 Chr.8: 27748308-27748308 on Build GRCh38
rs758988198 Chr.8: 27748319-27748319 on Build GRCh38
rs929706632 Chr.8: 27748282-27748282 on Build GRCh38
rs772267689 Chr.8: 27748055-27748055 on Build GRCh38
rs544933439 Chr.8: 27747992-27747992 on Build GRCh38
rs374219944 Chr.8: 27748210-27748210 on Build GRCh38
rs555691437 Chr.8: 27748393-27748393 on Build GRCh38
rs768702479 Chr.8: 27748309-27748309 on Build GRCh38
rs757629262 Chr.8: 27748292-27748292 on Build GRCh38
rs572117238 Chr.8: 27748427-27748427 on Build GRCh38
rs1057017176 Chr.8: 27748220-27748220 on Build GRCh38
rs149137907 Chr.8: 27748221-27748221 on Build GRCh38
rs764938057 Chr.8: 27748234-27748234 on Build GRCh38
rs747240444 Chr.8: 27748291-27748291 on Build GRCh38
rs1016075614 Chr.8: 27748191-27748191 on Build GRCh38
rs888073291 Chr.8: 27748436-27748436 on Build GRCh38
rs539637083 Chr.8: 27748150-27748150 on Build GRCh38
rs757755754 Chr.8: 27748171-27748171 on Build GRCh38
rs144489715 Chr.8: 27748015-27748015 on Build GRCh38
rs759148084 Chr.8: 27748113-27748122 on Build GRCh38
rs758425766 Chr.8: 27748204-27748204 on Build GRCh38
rs980999130 Chr.8: 27748198-27748198 on Build GRCh38
rs369943638 Chr.8: 27748147-27748147 on Build GRCh38
rs779289360 Chr.8: 27748178-27748178 on Build GRCh38
rs369502557 Chr.8: 27748070-27748070 on Build GRCh38
rs948873386 Chr.8: 27748356-27748356 on Build GRCh38
rs956091692 Chr.8: 27748304-27748304 on Build GRCh38
rs556940969 Chr.8: 27748326-27748327 on Build GRCh38
rs763525980 Chr.8: 27748332-27748360 on Build GRCh38
rs1018115736 Chr.8: 27748447-27748447 on Build GRCh38
rs142535974 Chr.8: 27748280-27748280 on Build GRCh38
rs755803603 Chr.8: 27748156-27748156 on Build GRCh38
rs139289703 Chr.8: 27748078-27748078 on Build GRCh38
rs867818444 Chr.8: 27748438-27748438 on Build GRCh38
rs1047885663 Chr.8: 27748107-27748107 on Build GRCh38
rs199979471 Chr.8: 27748219-27748219 on Build GRCh38
rs980707227 Chr.8: 27748379-27748379 on Build GRCh38
rs773131630 Chr.8: 27748214-27748214 on Build GRCh38
rs1018149568 Chr.8: 27747998-27747998 on Build GRCh38
rs954821439 Chr.8: 27748276-27748276 on Build GRCh38
rs774169211 Chr.8: 27748238-27748238 on Build GRCh38
rs934783087 Chr.8: 27747987-27747987 on Build GRCh38
rs948947055 Chr.8: 27748024-27748024 on Build GRCh38
rs766905336 Chr.8: 27748244-27748244 on Build GRCh38
rs1044648829 Chr.8: 27748026-27748026 on Build GRCh38
rs778949131 Chr.8: 27748034-27748034 on Build GRCh38
rs1005391107 Chr.8: 27747997-27747997 on Build GRCh38
rs761616875 Chr.8: 27748069-27748069 on Build GRCh38
rs776460710 Chr.8: 27748096-27748098 on Build GRCh38
rs766484443 Chr.8: 27748094-27748094 on Build GRCh38
rs759039261 Chr.8: 27748241-27748241 on Build GRCh38
rs760306103 Chr.8: 27748262-27748262 on Build GRCh38
rs200774099 Chr.8: 27748193-27748193 on Build GRCh38

View Full Product Details