Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 100681088-100681635 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYTL4
Entrez Gene ID
94121
Gene Name
synaptotagmin like 4
Gene Aliases
SLP4
UniGene
Hs.592224
Gene Chromosome Location
Chr.X: 100671964-100732148 on Build GRCh38
Transcripts
NM_001129896.2 NM_001174068.1 NM_080737.2 XM_005262229.2 XM_011531068.2 XM_017029966.1 XM_017029967.1 XM_017029968.1 XM_017029969.1 XM_017029970.1 XM_017029971.1

SNP Detail

SNP ID Chromosome Location
rs770328595 Chr.X: 100681446-100681446 on Build GRCh38
rs562068812 Chr.X: 100681313-100681313 on Build GRCh38
rs986797763 Chr.X: 100681591-100681591 on Build GRCh38
rs184576220 Chr.X: 100681326-100681326 on Build GRCh38
rs930069330 Chr.X: 100681540-100681540 on Build GRCh38
rs759224777 Chr.X: 100681271-100681271 on Build GRCh38
rs960637850 Chr.X: 100681463-100681463 on Build GRCh38
rs960643371 Chr.X: 100681344-100681344 on Build GRCh38
rs776665138 Chr.X: 100681389-100681389 on Build GRCh38
rs900120012 Chr.X: 100681228-100681228 on Build GRCh38
rs990770381 Chr.X: 100681385-100681385 on Build GRCh38
rs190470063 Chr.X: 100681478-100681478 on Build GRCh38
rs773557143 Chr.X: 100681245-100681245 on Build GRCh38
rs902276477 Chr.X: 100681312-100681312 on Build GRCh38
rs778572952 Chr.X: 100681382-100681382 on Build GRCh38
rs925035381 Chr.X: 100681165-100681165 on Build GRCh38
rs12846697 Chr.X: 100681545-100681545 on Build GRCh38
rs775087165 Chr.X: 100681276-100681276 on Build GRCh38
rs772480125 Chr.X: 100681235-100681235 on Build GRCh38
rs745726948 Chr.X: 100681146-100681146 on Build GRCh38
rs772533274 Chr.X: 100681366-100681366 on Build GRCh38
rs754950421 Chr.X: 100681336-100681336 on Build GRCh38
rs778106596 Chr.X: 100681476-100681476 on Build GRCh38
rs773612334 Chr.X: 100681370-100681370 on Build GRCh38
rs181609479 Chr.X: 100681610-100681610 on Build GRCh38
rs748521070 Chr.X: 100681166-100681166 on Build GRCh38
rs979081831 Chr.X: 100681592-100681592 on Build GRCh38
rs762123232 Chr.X: 100681450-100681450 on Build GRCh38
rs370019475 Chr.X: 100681272-100681272 on Build GRCh38
rs750213569 Chr.X: 100681318-100681318 on Build GRCh38
rs913542047 Chr.X: 100681137-100681137 on Build GRCh38
rs12846491 Chr.X: 100681546-100681546 on Build GRCh38
rs867077097 Chr.X: 100681322-100681322 on Build GRCh38
rs757247580 Chr.X: 100681301-100681301 on Build GRCh38
rs749418535 Chr.X: 100681331-100681331 on Build GRCh38
rs962497368 Chr.X: 100681419-100681419 on Build GRCh38
rs762173131 Chr.X: 100681226-100681226 on Build GRCh38
rs4828043 Chr.X: 100681150-100681150 on Build GRCh38
rs775634445 Chr.X: 100681167-100681167 on Build GRCh38
rs774802661 Chr.X: 100681201-100681201 on Build GRCh38
rs762328563 Chr.X: 100681278-100681278 on Build GRCh38
rs966799435 Chr.X: 100681571-100681571 on Build GRCh38
rs769233171 Chr.X: 100681176-100681176 on Build GRCh38
rs747053500 Chr.X: 100681302-100681302 on Build GRCh38
rs1017435056 Chr.X: 100681416-100681416 on Build GRCh38
rs751483395 Chr.X: 100681293-100681293 on Build GRCh38
rs1004193502 Chr.X: 100681411-100681411 on Build GRCh38
rs779409505 Chr.X: 100681352-100681352 on Build GRCh38
rs756408531 Chr.X: 100681319-100681319 on Build GRCh38
rs1002025461 Chr.X: 100681426-100681426 on Build GRCh38
rs898548644 Chr.X: 100681422-100681422 on Build GRCh38
rs748718034 Chr.X: 100681362-100681362 on Build GRCh38
rs111659690 Chr.X: 100681287-100681287 on Build GRCh38

View Full Product Details