Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 75671530-75672128 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCFC2
Entrez Gene ID
6936
Gene Name
GC-rich sequence DNA-binding factor 2
Gene Aliases
C2orf3, DNABF, GCF, TCF9
UniGene
Hs.303808
Gene Chromosome Location
Chr.2: 75662655-75710986 on Build GRCh38
Transcripts
NM_001201334.1 NM_003203.4 XM_005264520.3 XM_011533074.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4095862 c.1909T>A p.S637T Chr.2: 75671997-75671997 on Build GRCh38
4565272 c.1926_1927CC>TT p.Q643* Chr.2: 75671979-75671980 on Build GRCh38
4449271 c.1932_1933delAA p.R645fs*22 Chr.2: 75671973-75671974 on Build GRCh38
5048266 c.1906A>C p.T636P Chr.2: 75672000-75672000 on Build GRCh38
5476401 c.1892C>T p.A631V Chr.2: 75672014-75672014 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs958294455 Chr.2: 75671953-75671953 on Build GRCh38
rs936788099 Chr.2: 75671624-75671624 on Build GRCh38
rs558923997 Chr.2: 75671914-75671914 on Build GRCh38
rs769592113 Chr.2: 75672055-75672055 on Build GRCh38
rs777145216 Chr.2: 75672044-75672045 on Build GRCh38
rs375560842 Chr.2: 75672033-75672033 on Build GRCh38
rs943336338 Chr.2: 75672049-75672049 on Build GRCh38
rs779865744 Chr.2: 75672022-75672023 on Build GRCh38
rs757942471 Chr.2: 75672034-75672034 on Build GRCh38
rs1026786312 Chr.2: 75671868-75671868 on Build GRCh38
rs995279768 Chr.2: 75671854-75671854 on Build GRCh38
rs35772670 Chr.2: 75671752-75671752 on Build GRCh38
rs766589269 Chr.2: 75672012-75672012 on Build GRCh38
rs547133813 Chr.2: 75671616-75671616 on Build GRCh38
rs752918930 Chr.2: 75671764-75671765 on Build GRCh38
rs750598770 Chr.2: 75671982-75671982 on Build GRCh38
rs761925299 Chr.2: 75671972-75671972 on Build GRCh38
rs561510293 Chr.2: 75671919-75671919 on Build GRCh38
rs770800568 Chr.2: 75671905-75671905 on Build GRCh38
rs34342745 Chr.2: 75671765-75671768 on Build GRCh38
rs895365562 Chr.2: 75671820-75671820 on Build GRCh38
rs746797821 Chr.2: 75671738-75671738 on Build GRCh38
rs780841742 Chr.2: 75672048-75672048 on Build GRCh38
rs138640536 Chr.2: 75671610-75671610 on Build GRCh38
rs768081096 Chr.2: 75672035-75672072 on Build GRCh38
rs1061487 Chr.2: 75671880-75671880 on Build GRCh38
rs76454293 Chr.2: 75671801-75671801 on Build GRCh38
rs776497511 Chr.2: 75671907-75671907 on Build GRCh38
rs527662807 Chr.2: 75671890-75671890 on Build GRCh38
rs899733500 Chr.2: 75672082-75672082 on Build GRCh38
rs762903985 Chr.2: 75672065-75672065 on Build GRCh38
rs779524649 Chr.2: 75672032-75672032 on Build GRCh38
rs932585085 Chr.2: 75671593-75671593 on Build GRCh38
rs1052181292 Chr.2: 75671780-75671780 on Build GRCh38
rs1005503642 Chr.2: 75671874-75671874 on Build GRCh38
rs1027961975 Chr.2: 75671962-75671962 on Build GRCh38
rs867007440 Chr.2: 75671950-75671950 on Build GRCh38
rs945510749 Chr.2: 75671769-75671769 on Build GRCh38
rs921144076 Chr.2: 75671581-75671581 on Build GRCh38
rs563434521 Chr.2: 75672078-75672078 on Build GRCh38
rs746460361 Chr.2: 75672033-75672033 on Build GRCh38
rs200089923 Chr.2: 75672041-75672041 on Build GRCh38
rs777608890 Chr.2: 75671899-75671899 on Build GRCh38
rs867687728 Chr.2: 75671665-75671665 on Build GRCh38
rs779885043 Chr.2: 75671738-75671739 on Build GRCh38
rs914098547 Chr.2: 75671986-75671986 on Build GRCh38
rs760905013 Chr.2: 75671994-75671994 on Build GRCh38
rs143276161 Chr.2: 75672080-75672080 on Build GRCh38
rs575344146 Chr.2: 75671808-75671808 on Build GRCh38
rs769833520 Chr.2: 75671929-75671929 on Build GRCh38
rs539408319 Chr.2: 75671819-75671819 on Build GRCh38
rs141752962 Chr.2: 75671975-75671975 on Build GRCh38
rs548980943 Chr.2: 75671902-75671902 on Build GRCh38
rs530858259 Chr.2: 75672079-75672079 on Build GRCh38
rs755428710 Chr.2: 75672024-75672024 on Build GRCh38
rs989693823 Chr.2: 75671660-75671660 on Build GRCh38
rs376497200 Chr.2: 75671765-75671765 on Build GRCh38
rs780037325 Chr.2: 75671597-75671597 on Build GRCh38
rs777447188 Chr.2: 75672035-75672035 on Build GRCh38
rs754298952 Chr.2: 75672022-75672022 on Build GRCh38
rs886593033 Chr.2: 75671869-75671869 on Build GRCh38
rs557811672 Chr.2: 75671870-75671870 on Build GRCh38
rs377425203 Chr.2: 75672084-75672084 on Build GRCh38
rs1049809091 Chr.2: 75671857-75671857 on Build GRCh38
rs1025057513 Chr.2: 75671898-75671898 on Build GRCh38
rs745731374 Chr.2: 75672053-75672053 on Build GRCh38
rs1053961115 Chr.2: 75671746-75671746 on Build GRCh38
rs927995757 Chr.2: 75672076-75672076 on Build GRCh38
rs770710588 Chr.2: 75672042-75672042 on Build GRCh38
rs185239012 Chr.2: 75671802-75671802 on Build GRCh38
rs913959576 Chr.2: 75671747-75671747 on Build GRCh38
rs200603236 Chr.2: 75672011-75672011 on Build GRCh38
rs568180127 Chr.2: 75671791-75671791 on Build GRCh38
rs775533621 Chr.2: 75672060-75672060 on Build GRCh38
rs774279515 Chr.2: 75671955-75671955 on Build GRCh38

View Full Product Details