Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 33739038-33739302 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IP6K3
Entrez Gene ID
117283
Gene Name
inositol hexakisphosphate kinase 3
Gene Aliases
IHPK3, INSP6K3
UniGene
Hs.17253
Gene Chromosome Location
Chr.6: 33721476-33746997 on Build GRCh38
Transcripts
NM_001142883.1 NM_054111.4 XM_005248842.3 XM_011514295.2

SNP Detail

SNP ID Chromosome Location
rs1055846135 Chr.6: 33739105-33739106 on Build GRCh38
rs984298537 Chr.6: 33739213-33739213 on Build GRCh38
rs552967160 Chr.6: 33739267-33739267 on Build GRCh38
rs937255251 Chr.6: 33739083-33739083 on Build GRCh38
rs999613122 Chr.6: 33739192-33739192 on Build GRCh38
rs1014367915 Chr.6: 33739146-33739146 on Build GRCh38
rs902932260 Chr.6: 33739166-33739166 on Build GRCh38
rs28549894 Chr.6: 33739227-33739227 on Build GRCh38
rs987917095 Chr.6: 33739271-33739271 on Build GRCh38
rs1032483272 Chr.6: 33739196-33739196 on Build GRCh38
rs889140530 Chr.6: 33739105-33739105 on Build GRCh38
rs969491970 Chr.6: 33739240-33739240 on Build GRCh38
rs552891110 Chr.6: 33739082-33739082 on Build GRCh38
rs1037543665 Chr.6: 33739065-33739065 on Build GRCh38
rs568023190 Chr.6: 33739267-33739267 on Build GRCh38
rs774428923 Chr.6: 33739211-33739211 on Build GRCh38
rs1011421010 Chr.6: 33739076-33739076 on Build GRCh38
rs6923210 Chr.6: 33739159-33739159 on Build GRCh38
rs747914504 Chr.6: 33739145-33739145 on Build GRCh38
rs117663863 Chr.6: 33739062-33739062 on Build GRCh38
rs1027833890 Chr.6: 33739210-33739210 on Build GRCh38
rs1009139812 Chr.6: 33739115-33739115 on Build GRCh38
rs143320721 Chr.6: 33739228-33739228 on Build GRCh38
rs981990943 Chr.6: 33739265-33739265 on Build GRCh38
rs759732488 Chr.6: 33739215-33739215 on Build GRCh38
rs1052222286 Chr.6: 33739060-33739060 on Build GRCh38
rs571139799 Chr.6: 33739141-33739141 on Build GRCh38

View Full Product Details