Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 99280408-99280998 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAXC
Entrez Gene ID
84553
Gene Name
failed axon connections homolog
Gene Aliases
C6orf168, dJ273F20
UniGene
Hs.573245
Gene Chromosome Location
Chr.6: 99272915-99350081 on Build GRCh38
Transcripts
NM_032511.2 XM_006715582.2 XM_011536188.2

SNP Detail

SNP ID Chromosome Location
rs188656919 Chr.6: 99280889-99280889 on Build GRCh38
rs144808458 Chr.6: 99280467-99280467 on Build GRCh38
rs566034963 Chr.6: 99280663-99280663 on Build GRCh38
rs891654792 Chr.6: 99280542-99280542 on Build GRCh38
rs951824808 Chr.6: 99280960-99280960 on Build GRCh38
rs899270705 Chr.6: 99280747-99280747 on Build GRCh38
rs777742135 Chr.6: 99280448-99280448 on Build GRCh38
rs111918588 Chr.6: 99280962-99280962 on Build GRCh38
rs960841469 Chr.6: 99280677-99280677 on Build GRCh38
rs191353208 Chr.6: 99280441-99280441 on Build GRCh38
rs141031781 Chr.6: 99280458-99280458 on Build GRCh38
rs547643037 Chr.6: 99280629-99280629 on Build GRCh38
rs975255178 Chr.6: 99280878-99280878 on Build GRCh38
rs72921066 Chr.6: 99280498-99280498 on Build GRCh38
rs892147258 Chr.6: 99280668-99280668 on Build GRCh38
rs141896770 Chr.6: 99280613-99280613 on Build GRCh38
rs913602554 Chr.6: 99280949-99280949 on Build GRCh38
rs221538 Chr.6: 99280515-99280515 on Build GRCh38
rs1018622805 Chr.6: 99280560-99280560 on Build GRCh38
rs905409627 Chr.6: 99280698-99280698 on Build GRCh38
rs148024917 Chr.6: 99280450-99280450 on Build GRCh38
rs796799421 Chr.6: 99280736-99280736 on Build GRCh38
rs221539 Chr.6: 99280769-99280769 on Build GRCh38
rs1027307775 Chr.6: 99280953-99280953 on Build GRCh38
rs1046481042 Chr.6: 99280964-99280964 on Build GRCh38
rs912442670 Chr.6: 99280552-99280552 on Build GRCh38
rs537581638 Chr.6: 99280805-99280805 on Build GRCh38
rs904437446 Chr.6: 99280497-99280497 on Build GRCh38
rs762305337 Chr.6: 99280621-99280621 on Build GRCh38
rs221540 Chr.6: 99280812-99280812 on Build GRCh38
rs1002411140 Chr.6: 99280719-99280719 on Build GRCh38
rs1019534500 Chr.6: 99280821-99280821 on Build GRCh38
rs1008705365 Chr.6: 99280548-99280548 on Build GRCh38
rs572417027 Chr.6: 99280919-99280919 on Build GRCh38
rs983730243 Chr.6: 99280632-99280632 on Build GRCh38
rs868579827 Chr.6: 99280597-99280597 on Build GRCh38
rs1496979 Chr.6: 99280566-99280566 on Build GRCh38
rs951404208 Chr.6: 99280511-99280511 on Build GRCh38
rs764705587 Chr.6: 99280693-99280693 on Build GRCh38
rs905380965 Chr.6: 99280433-99280433 on Build GRCh38
rs1025621290 Chr.6: 99280432-99280432 on Build GRCh38
rs964296272 Chr.6: 99280842-99280842 on Build GRCh38
rs1009273533 Chr.6: 99280817-99280817 on Build GRCh38
rs574652457 Chr.6: 99280857-99280857 on Build GRCh38
rs1052944557 Chr.6: 99280853-99280853 on Build GRCh38
rs545465836 Chr.6: 99280936-99280936 on Build GRCh38
rs1026371560 Chr.6: 99280781-99280781 on Build GRCh38
rs745532607 Chr.6: 99280545-99280545 on Build GRCh38
rs183534137 Chr.6: 99280845-99280845 on Build GRCh38
rs770429345 Chr.6: 99280485-99280485 on Build GRCh38
rs558175814 Chr.6: 99280732-99280732 on Build GRCh38
rs948138013 Chr.6: 99280733-99280733 on Build GRCh38
rs539442228 Chr.6: 99280725-99280725 on Build GRCh38
rs908022196 Chr.6: 99280654-99280654 on Build GRCh38
rs763831245 Chr.6: 99280455-99280455 on Build GRCh38
rs925105416 Chr.6: 99280881-99280881 on Build GRCh38
rs542056541 Chr.6: 99280877-99280877 on Build GRCh38
rs949052065 Chr.6: 99280678-99280678 on Build GRCh38

View Full Product Details